Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 24969434-24969929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APMAP
Entrez Gene ID
57136
Gene Name
adipocyte plasma membrane associated protein
Gene Aliases
BSCv, C20orf3
UniGene
Hs.472330
Gene Chromosome Location
Chr.20: 24962925-24992974 on Build GRCh38
Transcripts
NM_020531.2 XM_005260763.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
370060 c.800C>G p.A267G Chr.20: 24969574-24969574 on Build GRCh38
2887244 c.760G>T p.D254Y Chr.20: 24969614-24969614 on Build GRCh38
4608907 c.766C>A p.L256M Chr.20: 24969608-24969608 on Build GRCh38
7262318 c.769C>T p.R257W Chr.20: 24969605-24969605 on Build GRCh38
2887242 c.816G>A p.L272L Chr.20: 24969558-24969558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758417972 Chr.20: 24969615-24969615 on Build GRCh38
rs756375649 Chr.20: 24969476-24969476 on Build GRCh38
rs573257929 Chr.20: 24969479-24969479 on Build GRCh38
rs551970985 Chr.20: 24969691-24969691 on Build GRCh38
rs901329399 Chr.20: 24969772-24969772 on Build GRCh38
rs759039288 Chr.20: 24969762-24969762 on Build GRCh38
rs1015938806 Chr.20: 24969561-24969561 on Build GRCh38
rs754938693 Chr.20: 24969484-24969484 on Build GRCh38
rs369751425 Chr.20: 24969597-24969597 on Build GRCh38
rs772567821 Chr.20: 24969731-24969731 on Build GRCh38
rs750576933 Chr.20: 24969587-24969587 on Build GRCh38
rs199672755 Chr.20: 24969630-24969630 on Build GRCh38
rs773138592 Chr.20: 24969647-24969647 on Build GRCh38
rs147481549 Chr.20: 24969657-24969657 on Build GRCh38
rs746627588 Chr.20: 24969512-24969512 on Build GRCh38
rs767438976 Chr.20: 24969579-24969579 on Build GRCh38
rs1005872426 Chr.20: 24969553-24969553 on Build GRCh38
rs764818369 Chr.20: 24969884-24969884 on Build GRCh38
rs1041626231 Chr.20: 24969496-24969496 on Build GRCh38
rs1028608306 Chr.20: 24969517-24969518 on Build GRCh38
rs901385969 Chr.20: 24969509-24969509 on Build GRCh38
rs180803202 Chr.20: 24969875-24969875 on Build GRCh38
rs779703247 Chr.20: 24969478-24969478 on Build GRCh38
rs565403248 Chr.20: 24969472-24969472 on Build GRCh38
rs774800541 Chr.20: 24969563-24969563 on Build GRCh38
rs537835316 Chr.20: 24969718-24969718 on Build GRCh38
rs915396188 Chr.20: 24969777-24969777 on Build GRCh38
rs554053736 Chr.20: 24969902-24969902 on Build GRCh38
rs915892195 Chr.20: 24969457-24969457 on Build GRCh38
rs771414788 Chr.20: 24969502-24969502 on Build GRCh38
rs762297659 Chr.20: 24969642-24969642 on Build GRCh38
rs1050418157 Chr.20: 24969780-24969780 on Build GRCh38
rs371614701 Chr.20: 24969523-24969523 on Build GRCh38
rs946802034 Chr.20: 24969799-24969799 on Build GRCh38
rs932859224 Chr.20: 24969772-24969772 on Build GRCh38
rs574493444 Chr.20: 24969760-24969760 on Build GRCh38
rs768944644 Chr.20: 24969636-24969636 on Build GRCh38
rs923939910 Chr.20: 24969860-24969860 on Build GRCh38
rs145175270 Chr.20: 24969645-24969645 on Build GRCh38
rs752621572 Chr.20: 24969612-24969612 on Build GRCh38
rs373098055 Chr.20: 24969593-24969593 on Build GRCh38
rs1051591059 Chr.20: 24969541-24969541 on Build GRCh38
rs754776239 Chr.20: 24969598-24969598 on Build GRCh38
rs947407062 Chr.20: 24969458-24969458 on Build GRCh38
rs148287096 Chr.20: 24969594-24969594 on Build GRCh38
rs199515877 Chr.20: 24969637-24969637 on Build GRCh38
rs1042449192 Chr.20: 24969851-24969851 on Build GRCh38
rs756751905 Chr.20: 24969755-24969755 on Build GRCh38
rs560951773 Chr.20: 24969605-24969605 on Build GRCh38
rs982370474 Chr.20: 24969726-24969726 on Build GRCh38
rs950376243 Chr.20: 24969700-24969700 on Build GRCh38
rs373986577 Chr.20: 24969616-24969616 on Build GRCh38
rs959936493 Chr.20: 24969634-24969634 on Build GRCh38
rs35673079 Chr.20: 24969666-24969667 on Build GRCh38
rs971833619 Chr.20: 24969608-24969608 on Build GRCh38
rs769408577 Chr.20: 24969689-24969689 on Build GRCh38
rs780852593 Chr.20: 24969841-24969841 on Build GRCh38
rs991007490 Chr.20: 24969761-24969761 on Build GRCh38
rs777347490 Chr.20: 24969505-24969505 on Build GRCh38
rs754042442 Chr.20: 24969708-24969708 on Build GRCh38
rs35097515 Chr.20: 24969529-24969529 on Build GRCh38
rs530680024 Chr.20: 24969549-24969549 on Build GRCh38
rs775324115 Chr.20: 24969905-24969905 on Build GRCh38
rs1041184012 Chr.20: 24969751-24969751 on Build GRCh38
rs888712519 Chr.20: 24969800-24969800 on Build GRCh38
rs114999734 Chr.20: 24969473-24969473 on Build GRCh38
rs780775339 Chr.20: 24969633-24969633 on Build GRCh38
rs775685510 Chr.20: 24969535-24969535 on Build GRCh38
rs2064404 Chr.20: 24969737-24969737 on Build GRCh38
rs568974661 Chr.20: 24969714-24969714 on Build GRCh38
rs748124283 Chr.20: 24969492-24969492 on Build GRCh38
rs745550814 Chr.20: 24969635-24969635 on Build GRCh38
rs143242757 Chr.20: 24969560-24969560 on Build GRCh38
rs762314042 Chr.20: 24969573-24969573 on Build GRCh38
rs200711893 Chr.20: 24969617-24969617 on Build GRCh38

View Full Product Details