Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 25446184-25446714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS1
Entrez Gene ID
9837
Gene Name
GINS complex subunit 1
Gene Aliases
PSF1
UniGene
Hs.658464
Gene Chromosome Location
Chr.20: 25407470-25448568 on Build GRCh38
Transcripts
NM_021067.4 NR_134574.1 XM_017028162.1

SNP Detail

SNP ID Chromosome Location
rs905247193 Chr.20: 25446330-25446330 on Build GRCh38
rs972305901 Chr.20: 25446616-25446616 on Build GRCh38
rs113033087 Chr.20: 25446324-25446324 on Build GRCh38
rs1036290554 Chr.20: 25446274-25446274 on Build GRCh38
rs568679712 Chr.20: 25446356-25446356 on Build GRCh38
rs182163699 Chr.20: 25446568-25446568 on Build GRCh38
rs1020572298 Chr.20: 25446341-25446341 on Build GRCh38
rs892508337 Chr.20: 25446407-25446407 on Build GRCh38
rs564240409 Chr.20: 25446391-25446391 on Build GRCh38
rs371295950 Chr.20: 25446530-25446530 on Build GRCh38
rs760555617 Chr.20: 25446246-25446246 on Build GRCh38
rs143658928 Chr.20: 25446348-25446348 on Build GRCh38
rs769751144 Chr.20: 25446331-25446331 on Build GRCh38
rs890719795 Chr.20: 25446459-25446459 on Build GRCh38
rs555393974 Chr.20: 25446335-25446335 on Build GRCh38
rs572117297 Chr.20: 25446339-25446339 on Build GRCh38
rs112278657 Chr.20: 25446265-25446265 on Build GRCh38
rs1009758364 Chr.20: 25446435-25446436 on Build GRCh38
rs921728320 Chr.20: 25446243-25446243 on Build GRCh38
rs2387887 Chr.20: 25446445-25446445 on Build GRCh38
rs984542869 Chr.20: 25446511-25446511 on Build GRCh38
rs761025 Chr.20: 25446332-25446332 on Build GRCh38
rs1014280058 Chr.20: 25446296-25446296 on Build GRCh38
rs796226139 Chr.20: 25446213-25446213 on Build GRCh38
rs940466690 Chr.20: 25446260-25446260 on Build GRCh38
rs1026632802 Chr.20: 25446593-25446593 on Build GRCh38
rs1018194919 Chr.20: 25446538-25446538 on Build GRCh38
rs868467756 Chr.20: 25446315-25446315 on Build GRCh38
rs1048816596 Chr.20: 25446252-25446252 on Build GRCh38
rs909065974 Chr.20: 25446254-25446254 on Build GRCh38
rs373208607 Chr.20: 25446333-25446333 on Build GRCh38
rs191965315 Chr.20: 25446210-25446210 on Build GRCh38
rs765476387 Chr.20: 25446669-25446669 on Build GRCh38
rs931709306 Chr.20: 25446245-25446245 on Build GRCh38
rs539638734 Chr.20: 25446286-25446286 on Build GRCh38
rs963781164 Chr.20: 25446557-25446557 on Build GRCh38
rs781012901 Chr.20: 25446235-25446235 on Build GRCh38
rs953003396 Chr.20: 25446476-25446476 on Build GRCh38
rs556157880 Chr.20: 25446304-25446304 on Build GRCh38
rs1010350354 Chr.20: 25446338-25446338 on Build GRCh38
rs905833810 Chr.20: 25446314-25446314 on Build GRCh38
rs541306415 Chr.20: 25446639-25446639 on Build GRCh38
rs531890448 Chr.20: 25446655-25446655 on Build GRCh38
rs6115186 Chr.20: 25446379-25446379 on Build GRCh38
rs949219178 Chr.20: 25446305-25446305 on Build GRCh38

View Full Product Details