Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32316467-32316957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF3B
Entrez Gene ID
9371
Gene Name
kinesin family member 3B
Gene Aliases
FLA8, HH0048, KLP-11
UniGene
Hs.369670
Gene Chromosome Location
Chr.20: 32277640-32335011 on Build GRCh38
Transcripts
NM_004798.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2889189 c.1564G>C p.E522Q Chr.20: 32316584-32316584 on Build GRCh38
4097531 c.1547G>A p.R516Q Chr.20: 32316567-32316567 on Build GRCh38
4097532 c.1582A>C p.S528R Chr.20: 32316602-32316602 on Build GRCh38
477999 c.1543A>G p.S515G Chr.20: 32316563-32316563 on Build GRCh38
3740080 c.1570A>G p.K524E Chr.20: 32316590-32316590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777509379 Chr.20: 32316746-32316746 on Build GRCh38
rs752058628 Chr.20: 32316501-32316501 on Build GRCh38
rs892400572 Chr.20: 32316668-32316668 on Build GRCh38
rs141321389 Chr.20: 32316723-32316723 on Build GRCh38
rs751459525 Chr.20: 32316903-32316903 on Build GRCh38
rs111537871 Chr.20: 32316839-32316839 on Build GRCh38
rs370905283 Chr.20: 32316640-32316640 on Build GRCh38
rs765011704 Chr.20: 32316881-32316881 on Build GRCh38
rs748453666 Chr.20: 32316514-32316514 on Build GRCh38
rs6058640 Chr.20: 32316652-32316652 on Build GRCh38
rs956533968 Chr.20: 32316929-32316929 on Build GRCh38
rs184402202 Chr.20: 32316913-32316913 on Build GRCh38
rs146627265 Chr.20: 32316601-32316601 on Build GRCh38
rs951385697 Chr.20: 32316516-32316516 on Build GRCh38
rs780855149 Chr.20: 32316763-32316763 on Build GRCh38
rs201190319 Chr.20: 32316717-32316717 on Build GRCh38
rs986500532 Chr.20: 32316937-32316937 on Build GRCh38
rs778743312 Chr.20: 32316585-32316585 on Build GRCh38
rs745419678 Chr.20: 32316700-32316700 on Build GRCh38
rs772643174 Chr.20: 32316808-32316808 on Build GRCh38
rs780352862 Chr.20: 32316566-32316566 on Build GRCh38
rs766533251 Chr.20: 32316851-32316851 on Build GRCh38
rs746709623 Chr.20: 32316614-32316614 on Build GRCh38
rs769065135 Chr.20: 32316788-32316788 on Build GRCh38
rs376074399 Chr.20: 32316661-32316661 on Build GRCh38
rs1013473472 Chr.20: 32316715-32316715 on Build GRCh38
rs754070840 Chr.20: 32316712-32316712 on Build GRCh38
rs375473275 Chr.20: 32316893-32316893 on Build GRCh38
rs764599019 Chr.20: 32316496-32316496 on Build GRCh38
rs559306689 Chr.20: 32316644-32316644 on Build GRCh38
rs900031635 Chr.20: 32316836-32316836 on Build GRCh38
rs764115675 Chr.20: 32316898-32316898 on Build GRCh38
rs141218746 Chr.20: 32316803-32316803 on Build GRCh38
rs201693817 Chr.20: 32316670-32316670 on Build GRCh38
rs367985780 Chr.20: 32316752-32316752 on Build GRCh38
rs745491129 Chr.20: 32316777-32316777 on Build GRCh38
rs756707908 Chr.20: 32316758-32316758 on Build GRCh38
rs761832012 Chr.20: 32316529-32316529 on Build GRCh38
rs766727312 Chr.20: 32316708-32316708 on Build GRCh38
rs938706099 Chr.20: 32316602-32316602 on Build GRCh38
rs776208585 Chr.20: 32316625-32316625 on Build GRCh38
rs747881866 Chr.20: 32316784-32316785 on Build GRCh38
rs773345947 Chr.20: 32316689-32316689 on Build GRCh38
rs750666937 Chr.20: 32316534-32316534 on Build GRCh38
rs1047220014 Chr.20: 32316533-32316533 on Build GRCh38
rs760775232 Chr.20: 32316699-32316699 on Build GRCh38
rs779345465 Chr.20: 32316487-32316487 on Build GRCh38
rs995658458 Chr.20: 32316899-32316899 on Build GRCh38
rs757897828 Chr.20: 32316613-32316613 on Build GRCh38
rs145903011 Chr.20: 32316591-32316591 on Build GRCh38
rs756998539 Chr.20: 32316567-32316567 on Build GRCh38
rs767559376 Chr.20: 32316667-32316667 on Build GRCh38
rs370815700 Chr.20: 32316835-32316835 on Build GRCh38
rs750141496 Chr.20: 32316910-32316910 on Build GRCh38
rs756399620 Chr.20: 32316550-32316550 on Build GRCh38
rs769315399 Chr.20: 32316793-32316793 on Build GRCh38
rs192552459 Chr.20: 32316909-32316909 on Build GRCh38
rs777044659 Chr.20: 32316857-32316857 on Build GRCh38
rs760865444 Chr.20: 32316845-32316845 on Build GRCh38
rs752545221 Chr.20: 32316883-32316883 on Build GRCh38
rs767611809 Chr.20: 32316530-32316530 on Build GRCh38
rs772318900 Chr.20: 32316686-32316686 on Build GRCh38
rs373788957 Chr.20: 32316744-32316744 on Build GRCh38
rs770576135 Chr.20: 32316619-32316619 on Build GRCh38
rs771876350 Chr.20: 32316903-32316904 on Build GRCh38
rs368469634 Chr.20: 32316583-32316583 on Build GRCh38
rs756803155 Chr.20: 32316908-32316908 on Build GRCh38
rs765061151 Chr.20: 32316724-32316724 on Build GRCh38
rs752638466 Chr.20: 32316734-32316734 on Build GRCh38

View Full Product Details