Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 34560459-34560970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGU
Entrez Gene ID
128869
Gene Name
phosphatidylinositol glycan anchor biosynthesis class U
Gene Aliases
CDC91L1, GAB1
UniGene
Hs.253319
Gene Chromosome Location
Chr.20: 34560542-34677285 on Build GRCh38
Transcripts
NM_080476.4 XM_011528542.2 XM_017027664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2758083 c.1270G>A p.A424T Chr.20: 34560904-34560904 on Build GRCh38
140858 c.1237G>A p.E413K Chr.20: 34560937-34560937 on Build GRCh38
6458755 c.1214_1215insTTA p.Y405_F406insY Chr.20: 34560959-34560960 on Build GRCh38
1026038 c.1300C>T p.L434F Chr.20: 34560874-34560874 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs543586464 Chr.20: 34560773-34560773 on Build GRCh38
rs1022786558 Chr.20: 34560779-34560779 on Build GRCh38
rs979612592 Chr.20: 34560631-34560631 on Build GRCh38
rs758714375 Chr.20: 34560877-34560877 on Build GRCh38
rs575514146 Chr.20: 34560794-34560794 on Build GRCh38
rs753705724 Chr.20: 34560664-34560664 on Build GRCh38
rs1026039386 Chr.20: 34560506-34560506 on Build GRCh38
rs751288004 Chr.20: 34560834-34560835 on Build GRCh38
rs139695705 Chr.20: 34560917-34560917 on Build GRCh38
rs757521315 Chr.20: 34560943-34560943 on Build GRCh38
rs988551322 Chr.20: 34560648-34560648 on Build GRCh38
rs992624434 Chr.20: 34560532-34560532 on Build GRCh38
rs370636644 Chr.20: 34560934-34560934 on Build GRCh38
rs755414217 Chr.20: 34560853-34560853 on Build GRCh38
rs1015866564 Chr.20: 34560500-34560500 on Build GRCh38
rs1026406269 Chr.20: 34560748-34560748 on Build GRCh38
rs372633825 Chr.20: 34560905-34560905 on Build GRCh38
rs1051373911 Chr.20: 34560739-34560739 on Build GRCh38
rs567167077 Chr.20: 34560901-34560901 on Build GRCh38
rs935473235 Chr.20: 34560720-34560720 on Build GRCh38
rs554142612 Chr.20: 34560586-34560586 on Build GRCh38
rs775126642 Chr.20: 34560834-34560834 on Build GRCh38
rs943612965 Chr.20: 34560703-34560704 on Build GRCh38
rs969908251 Chr.20: 34560780-34560780 on Build GRCh38
rs549880820 Chr.20: 34560615-34560616 on Build GRCh38
rs559972303 Chr.20: 34560810-34560810 on Build GRCh38
rs543195895 Chr.20: 34560597-34560597 on Build GRCh38
rs767058980 Chr.20: 34560926-34560926 on Build GRCh38
rs977196873 Chr.20: 34560921-34560921 on Build GRCh38
rs375918765 Chr.20: 34560836-34560836 on Build GRCh38
rs769689086 Chr.20: 34560823-34560823 on Build GRCh38
rs199852968 Chr.20: 34560911-34560911 on Build GRCh38
rs1040077195 Chr.20: 34560770-34560770 on Build GRCh38
rs766879381 Chr.20: 34560850-34560850 on Build GRCh38
rs141314118 Chr.20: 34560803-34560803 on Build GRCh38
rs772578844 Chr.20: 34560929-34560929 on Build GRCh38
rs958424033 Chr.20: 34560487-34560487 on Build GRCh38
rs116115328 Chr.20: 34560895-34560895 on Build GRCh38
rs397797141 Chr.20: 34560509-34560509 on Build GRCh38
rs145235171 Chr.20: 34560930-34560930 on Build GRCh38
rs781381271 Chr.20: 34560945-34560945 on Build GRCh38
rs754439832 Chr.20: 34560851-34560851 on Build GRCh38
rs969960645 Chr.20: 34560566-34560566 on Build GRCh38
rs182600789 Chr.20: 34560778-34560778 on Build GRCh38
rs914330536 Chr.20: 34560533-34560533 on Build GRCh38
rs763634932 Chr.20: 34560819-34560820 on Build GRCh38
rs773875817 Chr.20: 34560838-34560838 on Build GRCh38
rs933389471 Chr.20: 34560852-34560852 on Build GRCh38
rs527586356 Chr.20: 34560841-34560841 on Build GRCh38
rs1022835989 Chr.20: 34560545-34560546 on Build GRCh38
rs758779773 Chr.20: 34560936-34560936 on Build GRCh38
rs889308762 Chr.20: 34560698-34560698 on Build GRCh38
rs933441891 Chr.20: 34560686-34560686 on Build GRCh38
rs531421778 Chr.20: 34560904-34560904 on Build GRCh38
rs555247065 Chr.20: 34560611-34560611 on Build GRCh38
rs991562033 Chr.20: 34560528-34560528 on Build GRCh38
rs781407604 Chr.20: 34560887-34560887 on Build GRCh38
rs913955003 Chr.20: 34560682-34560682 on Build GRCh38
rs750761873 Chr.20: 34560755-34560755 on Build GRCh38
rs576601997 Chr.20: 34560702-34560702 on Build GRCh38
rs572793348 Chr.20: 34560592-34560592 on Build GRCh38
rs898861826 Chr.20: 34560501-34560501 on Build GRCh38
rs899431997 Chr.20: 34560728-34560728 on Build GRCh38
rs936655908 Chr.20: 34560626-34560626 on Build GRCh38
rs751898046 Chr.20: 34560940-34560940 on Build GRCh38
rs757477980 Chr.20: 34560886-34560886 on Build GRCh38
rs767830601 Chr.20: 34560741-34560741 on Build GRCh38
rs1046893414 Chr.20: 34560830-34560830 on Build GRCh38
rs956159827 Chr.20: 34560809-34560809 on Build GRCh38
rs368860915 Chr.20: 34560878-34560878 on Build GRCh38
rs1051877549 Chr.20: 34560693-34560693 on Build GRCh38
rs548379794 Chr.20: 34560939-34560939 on Build GRCh38
rs376467401 Chr.20: 34560872-34560872 on Build GRCh38
rs3830913 Chr.20: 34560505-34560505 on Build GRCh38
rs924075095 Chr.20: 34560667-34560667 on Build GRCh38

View Full Product Details