Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37944689-37945201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VSTM2L
Entrez Gene ID
128434
Gene Name
V-set and transmembrane domain containing 2 like
Gene Aliases
C20orf102, dJ1118M15.2
UniGene
Hs.517029
Gene Chromosome Location
Chr.20: 37903097-37945350 on Build GRCh38
Transcripts
NM_080607.2 XM_011528530.1

SNP Detail

SNP ID Chromosome Location
rs11550660 Chr.20: 37944928-37944928 on Build GRCh38
rs952184932 Chr.20: 37944920-37944920 on Build GRCh38
rs1049637337 Chr.20: 37945097-37945097 on Build GRCh38
rs116957143 Chr.20: 37944799-37944799 on Build GRCh38
rs1049627545 Chr.20: 37944809-37944809 on Build GRCh38
rs925156163 Chr.20: 37945052-37945052 on Build GRCh38
rs967224432 Chr.20: 37945044-37945044 on Build GRCh38
rs1040160073 Chr.20: 37945032-37945032 on Build GRCh38
rs1016640704 Chr.20: 37944862-37944862 on Build GRCh38
rs1017218313 Chr.20: 37945163-37945163 on Build GRCh38
rs1008273233 Chr.20: 37944872-37944872 on Build GRCh38
rs1043636768 Chr.20: 37944990-37944990 on Build GRCh38
rs979532950 Chr.20: 37945045-37945045 on Build GRCh38
rs534686022 Chr.20: 37945098-37945098 on Build GRCh38
rs1003865816 Chr.20: 37944861-37944861 on Build GRCh38
rs992043371 Chr.20: 37944916-37944916 on Build GRCh38
rs535763031 Chr.20: 37945165-37945165 on Build GRCh38
rs147932496 Chr.20: 37944936-37944936 on Build GRCh38
rs1041723416 Chr.20: 37944733-37944733 on Build GRCh38
rs528097548 Chr.20: 37944876-37944876 on Build GRCh38
rs932445316 Chr.20: 37945074-37945074 on Build GRCh38
rs913941320 Chr.20: 37944734-37944734 on Build GRCh38
rs998759572 Chr.20: 37945120-37945120 on Build GRCh38
rs563526579 Chr.20: 37944758-37944758 on Build GRCh38
rs1043975562 Chr.20: 37944776-37944776 on Build GRCh38
rs567978697 Chr.20: 37944882-37944882 on Build GRCh38
rs1030320224 Chr.20: 37944831-37944831 on Build GRCh38
rs530494546 Chr.20: 37944797-37944797 on Build GRCh38
rs943734531 Chr.20: 37945139-37945139 on Build GRCh38
rs763999923 Chr.20: 37944947-37944947 on Build GRCh38
rs552642257 Chr.20: 37944946-37944946 on Build GRCh38
rs374505306 Chr.20: 37944927-37944927 on Build GRCh38
rs2295379 Chr.20: 37945118-37945118 on Build GRCh38
rs2295378 Chr.20: 37944887-37944887 on Build GRCh38
rs771460731 Chr.20: 37944800-37944800 on Build GRCh38
rs751540980 Chr.20: 37945021-37945021 on Build GRCh38
rs913851780 Chr.20: 37944717-37944718 on Build GRCh38
rs1037130382 Chr.20: 37945141-37945141 on Build GRCh38
rs992258828 Chr.20: 37944729-37944729 on Build GRCh38
rs951702844 Chr.20: 37944854-37944854 on Build GRCh38
rs570655867 Chr.20: 37945067-37945067 on Build GRCh38
rs1039781907 Chr.20: 37944793-37944793 on Build GRCh38
rs887346791 Chr.20: 37945138-37945138 on Build GRCh38
rs1028268738 Chr.20: 37944917-37944917 on Build GRCh38
rs899960111 Chr.20: 37944795-37944795 on Build GRCh38
rs546523323 Chr.20: 37944870-37944870 on Build GRCh38
rs925931910 Chr.20: 37944996-37944996 on Build GRCh38
rs936006031 Chr.20: 37945029-37945029 on Build GRCh38
rs574368219 Chr.20: 37945144-37945144 on Build GRCh38
rs761595798 Chr.20: 37945023-37945023 on Build GRCh38
rs143323233 Chr.20: 37944889-37944889 on Build GRCh38
rs931092480 Chr.20: 37945166-37945166 on Build GRCh38
rs563885184 Chr.20: 37944819-37944819 on Build GRCh38
rs1051296633 Chr.20: 37945134-37945134 on Build GRCh38

View Full Product Details