Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38310448-38310870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPI
Entrez Gene ID
671
Gene Name
bactericidal/permeability-increasing protein
Gene Aliases
BPIFD1, rBPI
UniGene
Hs.529019
Gene Chromosome Location
Chr.20: 38304150-38337503 on Build GRCh38
Transcripts
NM_001725.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5584569 c.495C>T p.S165S Chr.20: 38310599-38310599 on Build GRCh38
1751444 c.533C>G p.S178* Chr.20: 38310637-38310637 on Build GRCh38
370061 c.459C>A p.N153K Chr.20: 38310563-38310563 on Build GRCh38
3389735 c.435G>A p.S145S Chr.20: 38310539-38310539 on Build GRCh38
1736747 c.398G>A p.G133D Chr.20: 38310502-38310502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003836341 Chr.20: 38310806-38310806 on Build GRCh38
rs5741801 Chr.20: 38310476-38310476 on Build GRCh38
rs754318437 Chr.20: 38310518-38310518 on Build GRCh38
rs1036826391 Chr.20: 38310723-38310723 on Build GRCh38
rs770117927 Chr.20: 38310683-38310683 on Build GRCh38
rs781611914 Chr.20: 38310664-38310664 on Build GRCh38
rs144469725 Chr.20: 38310534-38310534 on Build GRCh38
rs112553684 Chr.20: 38310474-38310474 on Build GRCh38
rs757592302 Chr.20: 38310631-38310633 on Build GRCh38
rs763647142 Chr.20: 38310623-38310626 on Build GRCh38
rs1039332883 Chr.20: 38310582-38310582 on Build GRCh38
rs899234728 Chr.20: 38310770-38310770 on Build GRCh38
rs1891032 Chr.20: 38310785-38310785 on Build GRCh38
rs993520879 Chr.20: 38310739-38310739 on Build GRCh38
rs539482020 Chr.20: 38310577-38310577 on Build GRCh38
rs568534730 Chr.20: 38310800-38310800 on Build GRCh38
rs189577107 Chr.20: 38310500-38310500 on Build GRCh38
rs1034862993 Chr.20: 38310820-38310820 on Build GRCh38
rs763371703 Chr.20: 38310689-38310689 on Build GRCh38
rs752215717 Chr.20: 38310626-38310626 on Build GRCh38
rs142530722 Chr.20: 38310569-38310569 on Build GRCh38
rs61738007 Chr.20: 38310627-38310627 on Build GRCh38
rs897828000 Chr.20: 38310724-38310724 on Build GRCh38
rs370476426 Chr.20: 38310703-38310703 on Build GRCh38
rs760534419 Chr.20: 38310704-38310704 on Build GRCh38
rs1004987298 Chr.20: 38310709-38310709 on Build GRCh38
rs764359200 Chr.20: 38310511-38310511 on Build GRCh38
rs745757472 Chr.20: 38310558-38310558 on Build GRCh38
rs777022242 Chr.20: 38310676-38310676 on Build GRCh38
rs370840038 Chr.20: 38310656-38310656 on Build GRCh38
rs5743506 Chr.20: 38310522-38310522 on Build GRCh38
rs990022566 Chr.20: 38310555-38310555 on Build GRCh38
rs372600089 Chr.20: 38310665-38310665 on Build GRCh38
rs759629446 Chr.20: 38310496-38310496 on Build GRCh38
rs148772768 Chr.20: 38310589-38310589 on Build GRCh38
rs778724219 Chr.20: 38310631-38310631 on Build GRCh38
rs5743507 Chr.20: 38310650-38310650 on Build GRCh38
rs757077332 Chr.20: 38310629-38310629 on Build GRCh38
rs185148895 Chr.20: 38310771-38310771 on Build GRCh38
rs888000772 Chr.20: 38310776-38310776 on Build GRCh38
rs775486929 Chr.20: 38310568-38310568 on Build GRCh38
rs745659743 Chr.20: 38310649-38310649 on Build GRCh38
rs760852444 Chr.20: 38310501-38310501 on Build GRCh38
rs956725282 Chr.20: 38310826-38310826 on Build GRCh38
rs201900727 Chr.20: 38310538-38310538 on Build GRCh38
rs375623075 Chr.20: 38310697-38310697 on Build GRCh38
rs946358831 Chr.20: 38310574-38310574 on Build GRCh38
rs748420838 Chr.20: 38310680-38310680 on Build GRCh38
rs780606760 Chr.20: 38310525-38310525 on Build GRCh38
rs778846878 Chr.20: 38310551-38310551 on Build GRCh38
rs770658143 Chr.20: 38310539-38310539 on Build GRCh38
rs558797206 Chr.20: 38310523-38310523 on Build GRCh38
rs754547702 Chr.20: 38310524-38310524 on Build GRCh38
rs891438730 Chr.20: 38310588-38310588 on Build GRCh38
rs773771842 Chr.20: 38310685-38310685 on Build GRCh38
rs184856239 Chr.20: 38310478-38310478 on Build GRCh38
rs772018382 Chr.20: 38310564-38310564 on Build GRCh38
rs550239436 Chr.20: 38310786-38310786 on Build GRCh38
rs765655700 Chr.20: 38310604-38310604 on Build GRCh38
rs193123151 Chr.20: 38310526-38310526 on Build GRCh38
rs546526229 Chr.20: 38310742-38310742 on Build GRCh38
rs546137094 Chr.20: 38310613-38310613 on Build GRCh38
rs147136315 Chr.20: 38310497-38310497 on Build GRCh38
rs575683984 Chr.20: 38310605-38310605 on Build GRCh38
rs763419053 Chr.20: 38310609-38310609 on Build GRCh38
rs534863168 Chr.20: 38310529-38310529 on Build GRCh38
rs751956603 Chr.20: 38310627-38310628 on Build GRCh38
rs373538095 Chr.20: 38310473-38310473 on Build GRCh38
rs762288593 Chr.20: 38310596-38310596 on Build GRCh38

View Full Product Details