Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 42074284-42074793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRT
Entrez Gene ID
11122
Gene Name
protein tyrosine phosphatase, receptor type T
Gene Aliases
RPTPrho
UniGene
Hs.526879
Gene Chromosome Location
Chr.20: 42072752-43189917 on Build GRCh38
Transcripts
NM_007050.5 NM_133170.3 XM_017027611.1 XM_017027612.1 XM_017027613.1

SNP Detail

SNP ID Chromosome Location
rs908988895 Chr.20: 42074638-42074638 on Build GRCh38
rs6102660 Chr.20: 42074383-42074383 on Build GRCh38
rs774730942 Chr.20: 42074504-42074504 on Build GRCh38
rs984575508 Chr.20: 42074408-42074408 on Build GRCh38
rs6072620 Chr.20: 42074678-42074678 on Build GRCh38
rs918198043 Chr.20: 42074325-42074325 on Build GRCh38
rs997775009 Chr.20: 42074600-42074600 on Build GRCh38
rs1018302393 Chr.20: 42074338-42074338 on Build GRCh38
rs766495811 Chr.20: 42074741-42074754 on Build GRCh38
rs971391062 Chr.20: 42074599-42074599 on Build GRCh38
rs940308555 Chr.20: 42074654-42074654 on Build GRCh38
rs570486471 Chr.20: 42074354-42074354 on Build GRCh38
rs78666075 Chr.20: 42074480-42074480 on Build GRCh38
rs965365510 Chr.20: 42074596-42074596 on Build GRCh38
rs1034258251 Chr.20: 42074612-42074612 on Build GRCh38
rs964972021 Chr.20: 42074342-42074342 on Build GRCh38
rs927024066 Chr.20: 42074369-42074369 on Build GRCh38
rs940359092 Chr.20: 42074439-42074439 on Build GRCh38
rs1042798399 Chr.20: 42074563-42074563 on Build GRCh38
rs952950896 Chr.20: 42074615-42074615 on Build GRCh38
rs574483727 Chr.20: 42074590-42074590 on Build GRCh38
rs971739935 Chr.20: 42074698-42074698 on Build GRCh38
rs901769259 Chr.20: 42074458-42074458 on Build GRCh38
rs1009832778 Chr.20: 42074554-42074554 on Build GRCh38
rs981368473 Chr.20: 42074367-42074367 on Build GRCh38
rs776088612 Chr.20: 42074402-42074402 on Build GRCh38
rs950709919 Chr.20: 42074609-42074609 on Build GRCh38
rs6102661 Chr.20: 42074724-42074724 on Build GRCh38
rs908873961 Chr.20: 42074421-42074421 on Build GRCh38
rs1025158294 Chr.20: 42074594-42074594 on Build GRCh38
rs369580892 Chr.20: 42074526-42074526 on Build GRCh38
rs140503676 Chr.20: 42074447-42074447 on Build GRCh38
rs1032413101 Chr.20: 42074528-42074528 on Build GRCh38
rs983438068 Chr.20: 42074618-42074618 on Build GRCh38
rs889172648 Chr.20: 42074484-42074484 on Build GRCh38
rs571947570 Chr.20: 42074478-42074478 on Build GRCh38
rs941887450 Chr.20: 42074445-42074445 on Build GRCh38
rs866175572 Chr.20: 42074565-42074565 on Build GRCh38
rs2144011 Chr.20: 42074487-42074487 on Build GRCh38
rs909268196 Chr.20: 42074432-42074432 on Build GRCh38
rs770309635 Chr.20: 42074327-42074327 on Build GRCh38
rs892453554 Chr.20: 42074544-42074544 on Build GRCh38
rs746400961 Chr.20: 42074316-42074316 on Build GRCh38
rs41278134 Chr.20: 42074606-42074606 on Build GRCh38
rs891463405 Chr.20: 42074503-42074503 on Build GRCh38
rs963493182 Chr.20: 42074687-42074687 on Build GRCh38
rs538572115 Chr.20: 42074364-42074364 on Build GRCh38
rs750488777 Chr.20: 42074650-42074650 on Build GRCh38
rs186579046 Chr.20: 42074723-42074723 on Build GRCh38
rs983554419 Chr.20: 42074371-42074371 on Build GRCh38
rs1056945373 Chr.20: 42074499-42074499 on Build GRCh38
rs937097228 Chr.20: 42074379-42074379 on Build GRCh38
rs923172842 Chr.20: 42074700-42074700 on Build GRCh38
rs573757263 Chr.20: 42074636-42074636 on Build GRCh38
rs904256981 Chr.20: 42074576-42074576 on Build GRCh38

View Full Product Details