Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44054218-44054727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOX2
Entrez Gene ID
84969
Gene Name
TOX high mobility group box family member 2
Gene Aliases
C20orf100, GCX-1, GCX1, dJ1108D11.2, dJ495O3.1
UniGene
Hs.26608
Gene Chromosome Location
Chr.20: 43914852-44069616 on Build GRCh38
Transcripts
NM_001098796.1 NM_001098797.1 NM_001098798.1 NM_032883.2 XM_006723884.1 XM_017028109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833969 c.372G>A p.P124P Chr.20: 44054385-44054385 on Build GRCh38
4455366 c.745A>T p.K249* Chr.20: 44054365-44054365 on Build GRCh38
3911377 c.579G>A p.P193P Chr.20: 44054352-44054352 on Build GRCh38
6806384 c.687C>A p.A229A Chr.20: 44054460-44054460 on Build GRCh38
7394697 c.483G>T p.M161I Chr.20: 44054496-44054496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561015693 Chr.20: 44054303-44054303 on Build GRCh38
rs759987340 Chr.20: 44054506-44054506 on Build GRCh38
rs138123229 Chr.20: 44054317-44054317 on Build GRCh38
rs760389856 Chr.20: 44054679-44054679 on Build GRCh38
rs761895673 Chr.20: 44054340-44054340 on Build GRCh38
rs147266413 Chr.20: 44054466-44054466 on Build GRCh38
rs111568371 Chr.20: 44054351-44054351 on Build GRCh38
rs766856606 Chr.20: 44054352-44054352 on Build GRCh38
rs759953916 Chr.20: 44054286-44054289 on Build GRCh38
rs149594311 Chr.20: 44054326-44054326 on Build GRCh38
rs768057191 Chr.20: 44054508-44054508 on Build GRCh38
rs867839104 Chr.20: 44054357-44054357 on Build GRCh38
rs528065786 Chr.20: 44054502-44054502 on Build GRCh38
rs775632756 Chr.20: 44054334-44054334 on Build GRCh38
rs1027631199 Chr.20: 44054240-44054240 on Build GRCh38
rs778414163 Chr.20: 44054539-44054539 on Build GRCh38
rs752969865 Chr.20: 44054515-44054515 on Build GRCh38
rs748871682 Chr.20: 44054312-44054312 on Build GRCh38
rs765545392 Chr.20: 44054343-44054343 on Build GRCh38
rs1028815281 Chr.20: 44054659-44054659 on Build GRCh38
rs758159018 Chr.20: 44054563-44054563 on Build GRCh38
rs138861548 Chr.20: 44054472-44054472 on Build GRCh38
rs778822885 Chr.20: 44054263-44054263 on Build GRCh38
rs758809703 Chr.20: 44054267-44054267 on Build GRCh38
rs572066015 Chr.20: 44054242-44054242 on Build GRCh38
rs773572720 Chr.20: 44054412-44054412 on Build GRCh38
rs74593867 Chr.20: 44054341-44054341 on Build GRCh38
rs771870520 Chr.20: 44054327-44054327 on Build GRCh38
rs750539790 Chr.20: 44054264-44054264 on Build GRCh38
rs775691925 Chr.20: 44054537-44054540 on Build GRCh38
rs763917185 Chr.20: 44054337-44054337 on Build GRCh38
rs200547615 Chr.20: 44054484-44054484 on Build GRCh38
rs996882728 Chr.20: 44054633-44054633 on Build GRCh38
rs768841808 Chr.20: 44054320-44054320 on Build GRCh38
rs755479356 Chr.20: 44054384-44054384 on Build GRCh38
rs201327310 Chr.20: 44054339-44054339 on Build GRCh38
rs376544017 Chr.20: 44054546-44054546 on Build GRCh38
rs752003699 Chr.20: 44054378-44054378 on Build GRCh38
rs779665607 Chr.20: 44054570-44054570 on Build GRCh38
rs138839996 Chr.20: 44054248-44054248 on Build GRCh38
rs955724515 Chr.20: 44054359-44054364 on Build GRCh38
rs563538915 Chr.20: 44054385-44054385 on Build GRCh38
rs73908516 Chr.20: 44054532-44054532 on Build GRCh38
rs202217962 Chr.20: 44054433-44054433 on Build GRCh38
rs757509566 Chr.20: 44054255-44054255 on Build GRCh38
rs370568497 Chr.20: 44054434-44054434 on Build GRCh38
rs746906395 Chr.20: 44054418-44054418 on Build GRCh38
rs150404582 Chr.20: 44054304-44054304 on Build GRCh38
rs768739987 Chr.20: 44054426-44054426 on Build GRCh38
rs531702660 Chr.20: 44054325-44054325 on Build GRCh38
rs756833543 Chr.20: 44054400-44054400 on Build GRCh38
rs1019598872 Chr.20: 44054580-44054580 on Build GRCh38
rs775113508 Chr.20: 44054535-44054535 on Build GRCh38
rs750439413 Chr.20: 44054348-44054348 on Build GRCh38
rs890548469 Chr.20: 44054656-44054656 on Build GRCh38
rs182587475 Chr.20: 44054406-44054406 on Build GRCh38
rs780580996 Chr.20: 44054292-44054292 on Build GRCh38
rs552399859 Chr.20: 44054407-44054407 on Build GRCh38
rs951637927 Chr.20: 44054241-44054241 on Build GRCh38
rs568147423 Chr.20: 44054607-44054607 on Build GRCh38
rs769954018 Chr.20: 44054351-44054352 on Build GRCh38
rs760704342 Chr.20: 44054335-44054335 on Build GRCh38
rs1048590426 Chr.20: 44054505-44054505 on Build GRCh38
rs1039170990 Chr.20: 44054700-44054700 on Build GRCh38
rs748564309 Chr.20: 44054399-44054399 on Build GRCh38
rs748617791 Chr.20: 44054391-44054391 on Build GRCh38
rs777906775 Chr.20: 44054435-44054435 on Build GRCh38
rs919237867 Chr.20: 44054576-44054576 on Build GRCh38
rs79564181 Chr.20: 44054614-44054614 on Build GRCh38
rs771652887 Chr.20: 44054411-44054411 on Build GRCh38

View Full Product Details