Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44280546-44281085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDAP1L1
Entrez Gene ID
78997
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Gene Aliases
dJ881L22.1, dJ995J12.1.1
UniGene
Hs.517059
Gene Chromosome Location
Chr.20: 44247099-44280917 on Build GRCh38
Transcripts
NM_001256737.1 NM_001256738.1 NM_001256739.1 NM_001256740.1 NM_024034.4 NR_046353.1

SNP Detail

SNP ID Chromosome Location
rs930699830 Chr.20: 44280621-44280621 on Build GRCh38
rs548287003 Chr.20: 44280647-44280647 on Build GRCh38
rs73107796 Chr.20: 44280749-44280749 on Build GRCh38
rs537503465 Chr.20: 44280699-44280699 on Build GRCh38
rs1009939060 Chr.20: 44280765-44280769 on Build GRCh38
rs148014853 Chr.20: 44280930-44280930 on Build GRCh38
rs1047946005 Chr.20: 44280662-44280662 on Build GRCh38
rs1002139855 Chr.20: 44280925-44280925 on Build GRCh38
rs577085341 Chr.20: 44280737-44280737 on Build GRCh38
rs920659509 Chr.20: 44280620-44280620 on Build GRCh38
rs867450633 Chr.20: 44280863-44280863 on Build GRCh38
rs768566525 Chr.20: 44280814-44280814 on Build GRCh38
rs1055634790 Chr.20: 44280612-44280612 on Build GRCh38
rs747862965 Chr.20: 44281037-44281037 on Build GRCh38
rs557208369 Chr.20: 44280706-44280706 on Build GRCh38
rs939928313 Chr.20: 44280692-44280692 on Build GRCh38
rs776615590 Chr.20: 44280901-44280901 on Build GRCh38
rs952865808 Chr.20: 44281004-44281004 on Build GRCh38
rs906526985 Chr.20: 44280909-44280909 on Build GRCh38
rs1009164063 Chr.20: 44280674-44280674 on Build GRCh38
rs568571492 Chr.20: 44280693-44280693 on Build GRCh38
rs892911435 Chr.20: 44280762-44280762 on Build GRCh38
rs892759406 Chr.20: 44280851-44280851 on Build GRCh38
rs1040747600 Chr.20: 44280698-44280698 on Build GRCh38
rs1010226262 Chr.20: 44280862-44280862 on Build GRCh38
rs1001048347 Chr.20: 44280769-44280769 on Build GRCh38
rs1034245951 Chr.20: 44280825-44280825 on Build GRCh38
rs1054325098 Chr.20: 44280815-44280815 on Build GRCh38
rs1040592658 Chr.20: 44280686-44280686 on Build GRCh38
rs758008484 Chr.20: 44280664-44280664 on Build GRCh38
rs1033740250 Chr.20: 44280930-44280930 on Build GRCh38
rs971169317 Chr.20: 44280813-44280813 on Build GRCh38
rs1015832989 Chr.20: 44281053-44281053 on Build GRCh38
rs1020108202 Chr.20: 44280793-44280793 on Build GRCh38
rs1024961303 Chr.20: 44280905-44280905 on Build GRCh38
rs908456265 Chr.20: 44280690-44280690 on Build GRCh38
rs937987000 Chr.20: 44280579-44280579 on Build GRCh38

View Full Product Details