Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44753495-44754005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RIMS4
Entrez Gene ID
140730
Gene Name
regulating synaptic membrane exocytosis 4
Gene Aliases
C20orf190, RIM 4, RIM-4, RIM4, RIM4-gamma, RIM4gamma
UniGene
Hs.517065
Gene Chromosome Location
Chr.20: 44751804-44810338 on Build GRCh38
Transcripts
NM_001205317.1 NM_182970.3

SNP Detail

SNP ID Chromosome Location
rs551377453 Chr.20: 44753742-44753742 on Build GRCh38
rs866563961 Chr.20: 44753866-44753866 on Build GRCh38
rs911042207 Chr.20: 44753678-44753678 on Build GRCh38
rs536036504 Chr.20: 44753888-44753888 on Build GRCh38
rs188407189 Chr.20: 44753902-44753902 on Build GRCh38
rs1010792473 Chr.20: 44753826-44753826 on Build GRCh38
rs575688475 Chr.20: 44753533-44753533 on Build GRCh38
rs146873042 Chr.20: 44753801-44753801 on Build GRCh38
rs564847021 Chr.20: 44753774-44753774 on Build GRCh38
rs557843149 Chr.20: 44753970-44753970 on Build GRCh38
rs914335729 Chr.20: 44753974-44753974 on Build GRCh38
rs1040776748 Chr.20: 44753797-44753797 on Build GRCh38
rs776634955 Chr.20: 44753833-44753835 on Build GRCh38
rs752810542 Chr.20: 44753541-44753541 on Build GRCh38
rs549345331 Chr.20: 44753901-44753901 on Build GRCh38
rs1009864379 Chr.20: 44753714-44753714 on Build GRCh38
rs757774430 Chr.20: 44753862-44753862 on Build GRCh38
rs141334674 Chr.20: 44753680-44753680 on Build GRCh38
rs764441832 Chr.20: 44753755-44753755 on Build GRCh38
rs1051390980 Chr.20: 44753674-44753674 on Build GRCh38
rs902790375 Chr.20: 44753721-44753721 on Build GRCh38
rs145116784 Chr.20: 44753703-44753703 on Build GRCh38
rs918516666 Chr.20: 44753641-44753641 on Build GRCh38
rs962756438 Chr.20: 44753845-44753845 on Build GRCh38
rs756330627 Chr.20: 44753614-44753614 on Build GRCh38
rs369045336 Chr.20: 44753523-44753523 on Build GRCh38
rs185213483 Chr.20: 44753813-44753813 on Build GRCh38
rs189314670 Chr.20: 44753613-44753613 on Build GRCh38
rs557757569 Chr.20: 44753598-44753598 on Build GRCh38
rs995399313 Chr.20: 44753892-44753895 on Build GRCh38
rs140649597 Chr.20: 44753892-44753893 on Build GRCh38
rs972691082 Chr.20: 44753621-44753621 on Build GRCh38
rs573731320 Chr.20: 44753655-44753655 on Build GRCh38
rs1041335237 Chr.20: 44753719-44753719 on Build GRCh38
rs958636397 Chr.20: 44753891-44753892 on Build GRCh38
rs1035171505 Chr.20: 44753569-44753569 on Build GRCh38
rs537920063 Chr.20: 44753964-44753964 on Build GRCh38
rs966898185 Chr.20: 44753538-44753538 on Build GRCh38
rs115145073 Chr.20: 44753617-44753617 on Build GRCh38
rs537832145 Chr.20: 44753537-44753537 on Build GRCh38
rs893328723 Chr.20: 44753814-44753814 on Build GRCh38
rs1004132275 Chr.20: 44753552-44753552 on Build GRCh38
rs181976237 Chr.20: 44753704-44753704 on Build GRCh38
rs932208544 Chr.20: 44753805-44753805 on Build GRCh38
rs1025628655 Chr.20: 44753852-44753852 on Build GRCh38
rs185175486 Chr.20: 44753609-44753609 on Build GRCh38
rs981474162 Chr.20: 44753578-44753578 on Build GRCh38
rs1035625824 Chr.20: 44753743-44753743 on Build GRCh38
rs537814967 Chr.20: 44753853-44753853 on Build GRCh38
rs779329696 Chr.20: 44753930-44753930 on Build GRCh38
rs959744953 Chr.20: 44753579-44753579 on Build GRCh38
rs1011150476 Chr.20: 44753525-44753525 on Build GRCh38
rs982984888 Chr.20: 44753929-44753929 on Build GRCh38
rs549298764 Chr.20: 44753642-44753642 on Build GRCh38
rs911083787 Chr.20: 44753895-44753895 on Build GRCh38
rs972743683 Chr.20: 44753851-44753851 on Build GRCh38
rs777783425 Chr.20: 44753630-44753630 on Build GRCh38
rs990907214 Chr.20: 44753582-44753582 on Build GRCh38

View Full Product Details