Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46013513-46014054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMP9
Entrez Gene ID
4318
Gene Name
matrix metallopeptidase 9
Gene Aliases
CLG4B, GELB, MANDP2, MMP-9
UniGene
Hs.297413
Gene Chromosome Location
Chr.20: 46008908-46016561 on Build GRCh38
Transcripts
NM_004994.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558830 c.1610G>T p.G537V Chr.20: 46013534-46013534 on Build GRCh38
3294223 c.1637delG p.S548fs*66 Chr.20: 46013683-46013683 on Build GRCh38
6051637 c.1596T>G p.Y532* Chr.20: 46013520-46013520 on Build GRCh38
5513633 c.1686G>A p.A562A Chr.20: 46013732-46013732 on Build GRCh38
4608927 c.1610+3G>T p.? Chr.20: 46013537-46013537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754955508 Chr.20: 46013668-46013668 on Build GRCh38
rs200548880 Chr.20: 46013667-46013667 on Build GRCh38
rs138685704 Chr.20: 46013725-46013725 on Build GRCh38
rs892533981 Chr.20: 46013870-46013870 on Build GRCh38
rs752829288 Chr.20: 46013609-46013609 on Build GRCh38
rs767178225 Chr.20: 46013860-46013860 on Build GRCh38
rs185167031 Chr.20: 46013631-46013631 on Build GRCh38
rs754829019 Chr.20: 46013763-46013763 on Build GRCh38
rs3918259 Chr.20: 46013871-46013871 on Build GRCh38
rs775387047 Chr.20: 46013721-46013721 on Build GRCh38
rs771415685 Chr.20: 46013716-46013716 on Build GRCh38
rs756255359 Chr.20: 46013618-46013618 on Build GRCh38
rs3918258 Chr.20: 46013565-46013565 on Build GRCh38
rs2250889 Chr.20: 46013767-46013767 on Build GRCh38
rs199657984 Chr.20: 46013797-46013797 on Build GRCh38
rs557525204 Chr.20: 46013644-46013644 on Build GRCh38
rs758597128 Chr.20: 46013791-46013792 on Build GRCh38
rs780500313 Chr.20: 46013671-46013671 on Build GRCh38
rs200866285 Chr.20: 46013717-46013717 on Build GRCh38
rs779383755 Chr.20: 46013775-46013775 on Build GRCh38
rs1025251522 Chr.20: 46013632-46013632 on Build GRCh38
rs573847655 Chr.20: 46013821-46013821 on Build GRCh38
rs765341372 Chr.20: 46013611-46013612 on Build GRCh38
rs927750224 Chr.20: 46013911-46013911 on Build GRCh38
rs758158592 Chr.20: 46013635-46013635 on Build GRCh38
rs763671512 Chr.20: 46014022-46014022 on Build GRCh38
rs970028758 Chr.20: 46013681-46013681 on Build GRCh38
rs990449222 Chr.20: 46013928-46013928 on Build GRCh38
rs3918260 Chr.20: 46013893-46013893 on Build GRCh38
rs180783469 Chr.20: 46014033-46014033 on Build GRCh38
rs556160913 Chr.20: 46013550-46013550 on Build GRCh38
rs200196442 Chr.20: 46013630-46013630 on Build GRCh38
rs926968908 Chr.20: 46013704-46013704 on Build GRCh38
rs748984817 Chr.20: 46013699-46013699 on Build GRCh38
rs989313438 Chr.20: 46013673-46013673 on Build GRCh38
rs757535139 Chr.20: 46013752-46013752 on Build GRCh38
rs201880215 Chr.20: 46013559-46013559 on Build GRCh38
rs201887847 Chr.20: 46013817-46013817 on Build GRCh38
rs1017491592 Chr.20: 46013869-46013869 on Build GRCh38
rs989429349 Chr.20: 46013875-46013875 on Build GRCh38
rs961763071 Chr.20: 46013575-46013575 on Build GRCh38
rs772374825 Chr.20: 46013785-46013785 on Build GRCh38
rs955534468 Chr.20: 46013857-46013857 on Build GRCh38
rs201312492 Chr.20: 46013772-46013772 on Build GRCh38
rs533464744 Chr.20: 46013828-46013828 on Build GRCh38
rs981016109 Chr.20: 46013683-46013683 on Build GRCh38
rs41410049 Chr.20: 46013848-46013848 on Build GRCh38
rs761153280 Chr.20: 46013801-46013801 on Build GRCh38
rs767440904 Chr.20: 46013568-46013568 on Build GRCh38
rs553302870 Chr.20: 46014031-46014031 on Build GRCh38
rs535088896 Chr.20: 46013918-46013918 on Build GRCh38
rs973709572 Chr.20: 46013940-46013940 on Build GRCh38
rs189827770 Chr.20: 46013766-46013766 on Build GRCh38
rs369346801 Chr.20: 46013553-46013553 on Build GRCh38
rs1038500196 Chr.20: 46013776-46013776 on Build GRCh38
rs951121455 Chr.20: 46014018-46014018 on Build GRCh38
rs747724013 Chr.20: 46013684-46013684 on Build GRCh38
rs141966515 Chr.20: 46013727-46013727 on Build GRCh38
rs1005636653 Chr.20: 46013773-46013773 on Build GRCh38
rs553464370 Chr.20: 46013593-46013593 on Build GRCh38
rs941610243 Chr.20: 46014012-46014012 on Build GRCh38
rs150224695 Chr.20: 46013719-46013719 on Build GRCh38
rs35691798 Chr.20: 46013757-46013757 on Build GRCh38
rs994304932 Chr.20: 46013591-46013591 on Build GRCh38
rs543335411 Chr.20: 46013740-46013740 on Build GRCh38
rs769280932 Chr.20: 46013691-46013691 on Build GRCh38
rs549431429 Chr.20: 46013885-46013885 on Build GRCh38
rs747719903 Chr.20: 46013600-46013600 on Build GRCh38
rs752994774 Chr.20: 46013671-46013672 on Build GRCh38
rs1050867679 Chr.20: 46013818-46013818 on Build GRCh38
rs200663446 Chr.20: 46013826-46013826 on Build GRCh38
rs1022534576 Chr.20: 46013884-46013884 on Build GRCh38
rs781210808 Chr.20: 46013765-46013765 on Build GRCh38
rs746270965 Chr.20: 46013783-46013783 on Build GRCh38
rs201602753 Chr.20: 46013683-46013683 on Build GRCh38
rs201797233 Chr.20: 46013638-46013638 on Build GRCh38
rs772972610 Chr.20: 46013694-46013694 on Build GRCh38
rs200375269 Chr.20: 46013741-46013741 on Build GRCh38
rs770714851 Chr.20: 46013771-46013771 on Build GRCh38
rs201266379 Chr.20: 46013560-46013560 on Build GRCh38
rs760437446 Chr.20: 46014004-46014004 on Build GRCh38
rs751263874 Chr.20: 46013825-46013825 on Build GRCh38
rs200572037 Chr.20: 46013837-46013837 on Build GRCh38
rs200924143 Chr.20: 46013804-46013804 on Build GRCh38
rs759896847 Chr.20: 46013718-46013718 on Build GRCh38
rs1031056760 Chr.20: 46013622-46013622 on Build GRCh38
rs199907705 Chr.20: 46013820-46013820 on Build GRCh38
rs146961494 Chr.20: 46013756-46013756 on Build GRCh38

View Full Product Details