Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47015839-47016354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
UniGene
Hs.472877
Gene Chromosome Location
Chr.20: 46894624-47188844 on Build GRCh38
Transcripts
NM_005244.4 NM_172110.3 XM_005260327.2 XM_017027721.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5699862 c.313G>C p.D105H Chr.20: 47016195-47016195 on Build GRCh38
3963628 c.356A>G p.Y119C Chr.20: 47016238-47016238 on Build GRCh38
7103982 c.310G>A p.E104K Chr.20: 47016192-47016192 on Build GRCh38
5673666 c.400_415+6del22 p.? Chr.20: 47016282-47016303 on Build GRCh38
1412222 c.358G>T p.G120C Chr.20: 47016240-47016240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057492340 Chr.20: 47016061-47016061 on Build GRCh38
rs765160974 Chr.20: 47016199-47016199 on Build GRCh38
rs770297353 Chr.20: 47016147-47016147 on Build GRCh38
rs972000985 Chr.20: 47015928-47015928 on Build GRCh38
rs942713561 Chr.20: 47016048-47016048 on Build GRCh38
rs370908032 Chr.20: 47015893-47015893 on Build GRCh38
rs201099338 Chr.20: 47016246-47016246 on Build GRCh38
rs775687789 Chr.20: 47016325-47016325 on Build GRCh38
rs1012662634 Chr.20: 47016184-47016184 on Build GRCh38
rs759717805 Chr.20: 47016291-47016291 on Build GRCh38
rs931751936 Chr.20: 47015892-47015892 on Build GRCh38
rs575100947 Chr.20: 47015955-47015955 on Build GRCh38
rs778332979 Chr.20: 47016266-47016266 on Build GRCh38
rs1027047175 Chr.20: 47016118-47016118 on Build GRCh38
rs914577861 Chr.20: 47016083-47016083 on Build GRCh38
rs771105381 Chr.20: 47016279-47016279 on Build GRCh38
rs954552992 Chr.20: 47015961-47015961 on Build GRCh38
rs367617095 Chr.20: 47016173-47016173 on Build GRCh38
rs759880122 Chr.20: 47016177-47016177 on Build GRCh38
rs749880494 Chr.20: 47016274-47016274 on Build GRCh38
rs868813774 Chr.20: 47016002-47016002 on Build GRCh38
rs755771791 Chr.20: 47015978-47015978 on Build GRCh38
rs1041961029 Chr.20: 47016109-47016109 on Build GRCh38
rs1041943225 Chr.20: 47015972-47015972 on Build GRCh38
rs770068078 Chr.20: 47016262-47016262 on Build GRCh38
rs550862358 Chr.20: 47016162-47016162 on Build GRCh38
rs778937707 Chr.20: 47016162-47016163 on Build GRCh38
rs13347174 Chr.20: 47016217-47016217 on Build GRCh38
rs1030134691 Chr.20: 47016207-47016207 on Build GRCh38
rs771197035 Chr.20: 47016165-47016165 on Build GRCh38
rs775584894 Chr.20: 47016294-47016294 on Build GRCh38
rs563132318 Chr.20: 47015881-47015881 on Build GRCh38
rs138752804 Chr.20: 47016295-47016295 on Build GRCh38
rs766324822 Chr.20: 47016309-47016309 on Build GRCh38
rs763962033 Chr.20: 47016181-47016181 on Build GRCh38
rs552714929 Chr.20: 47016140-47016140 on Build GRCh38
rs781655445 Chr.20: 47016141-47016141 on Build GRCh38
rs748485679 Chr.20: 47016144-47016144 on Build GRCh38
rs757898980 Chr.20: 47016218-47016218 on Build GRCh38
rs542413714 Chr.20: 47016052-47016052 on Build GRCh38
rs1026587935 Chr.20: 47015949-47015949 on Build GRCh38
rs778060469 Chr.20: 47016022-47016022 on Build GRCh38
rs898811147 Chr.20: 47016206-47016206 on Build GRCh38
rs752350674 Chr.20: 47016317-47016317 on Build GRCh38
rs77478939 Chr.20: 47016085-47016085 on Build GRCh38
rs771134776 Chr.20: 47016129-47016129 on Build GRCh38
rs764647733 Chr.20: 47016071-47016071 on Build GRCh38
rs767140019 Chr.20: 47016311-47016311 on Build GRCh38
rs371678825 Chr.20: 47016308-47016308 on Build GRCh38
rs748478693 Chr.20: 47016260-47016260 on Build GRCh38
rs1016381387 Chr.20: 47016193-47016193 on Build GRCh38
rs146537659 Chr.20: 47016183-47016183 on Build GRCh38
rs754490646 Chr.20: 47016310-47016310 on Build GRCh38
rs749664461 Chr.20: 47016024-47016024 on Build GRCh38
rs867826246 Chr.20: 47016223-47016223 on Build GRCh38
rs547961765 Chr.20: 47016331-47016331 on Build GRCh38
rs772311315 Chr.20: 47016282-47016282 on Build GRCh38
rs113021703 Chr.20: 47016073-47016073 on Build GRCh38
rs923011433 Chr.20: 47015870-47015870 on Build GRCh38
rs779577193 Chr.20: 47016238-47016238 on Build GRCh38
rs531965679 Chr.20: 47016151-47016151 on Build GRCh38
rs567374244 Chr.20: 47016155-47016155 on Build GRCh38
rs761515314 Chr.20: 47016195-47016195 on Build GRCh38
rs779576777 Chr.20: 47016221-47016221 on Build GRCh38
rs115924568 Chr.20: 47016258-47016258 on Build GRCh38
rs768498940 Chr.20: 47016154-47016155 on Build GRCh38
rs746329857 Chr.20: 47016261-47016261 on Build GRCh38
rs534576492 Chr.20: 47015939-47015939 on Build GRCh38
rs746729679 Chr.20: 47016134-47016134 on Build GRCh38
rs986867817 Chr.20: 47016010-47016010 on Build GRCh38
rs546319918 Chr.20: 47016120-47016120 on Build GRCh38
rs765007512 Chr.20: 47016296-47016296 on Build GRCh38
rs570725372 Chr.20: 47016128-47016128 on Build GRCh38
rs544889103 Chr.20: 47015869-47015869 on Build GRCh38

View Full Product Details