Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 49063075-49063616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSE1L
Entrez Gene ID
1434
Gene Name
chromosome segregation 1 like
Gene Aliases
CAS, CSE1, XPO2
UniGene
Hs.90073
Gene Chromosome Location
Chr.20: 49046246-49096960 on Build GRCh38
Transcripts
NM_001256135.1 NM_001316.3 NR_045796.1 XM_011528599.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1249165 c.222G>A p.W74* Chr.20: 49063338-49063338 on Build GRCh38
4474026 c.188C>T p.S63L Chr.20: 49063304-49063304 on Build GRCh38
3727323 c.202A>C p.N68H Chr.20: 49063318-49063318 on Build GRCh38
2761669 c.131T>G p.L44R Chr.20: 49063247-49063247 on Build GRCh38
4099183 c.139T>A p.L47M Chr.20: 49063255-49063255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759411890 Chr.20: 49063119-49063121 on Build GRCh38
rs112333627 Chr.20: 49063325-49063325 on Build GRCh38
rs36109051 Chr.20: 49063125-49063125 on Build GRCh38
rs142935832 Chr.20: 49063547-49063547 on Build GRCh38
rs758027401 Chr.20: 49063169-49063169 on Build GRCh38
rs746886192 Chr.20: 49063275-49063275 on Build GRCh38
rs566273276 Chr.20: 49063136-49063136 on Build GRCh38
rs10714453 Chr.20: 49063127-49063127 on Build GRCh38
rs987449591 Chr.20: 49063189-49063189 on Build GRCh38
rs77912819 Chr.20: 49063563-49063563 on Build GRCh38
rs900138418 Chr.20: 49063288-49063288 on Build GRCh38
rs762345348 Chr.20: 49063308-49063308 on Build GRCh38
rs370198144 Chr.20: 49063171-49063171 on Build GRCh38
rs61452497 Chr.20: 49063160-49063160 on Build GRCh38
rs992843122 Chr.20: 49063132-49063132 on Build GRCh38
rs776940831 Chr.20: 49063121-49063121 on Build GRCh38
rs982895976 Chr.20: 49063566-49063566 on Build GRCh38
rs771766492 Chr.20: 49063142-49063142 on Build GRCh38
rs772660162 Chr.20: 49063307-49063307 on Build GRCh38
rs542485836 Chr.20: 49063504-49063504 on Build GRCh38
rs753751089 Chr.20: 49063544-49063544 on Build GRCh38
rs750076249 Chr.20: 49063161-49063161 on Build GRCh38
rs748608785 Chr.20: 49063209-49063209 on Build GRCh38
rs372846963 Chr.20: 49063242-49063242 on Build GRCh38
rs761669365 Chr.20: 49063292-49063292 on Build GRCh38
rs968779550 Chr.20: 49063126-49063127 on Build GRCh38
rs114388644 Chr.20: 49063375-49063375 on Build GRCh38
rs527491080 Chr.20: 49063500-49063500 on Build GRCh38
rs1049954551 Chr.20: 49063535-49063535 on Build GRCh38
rs755265305 Chr.20: 49063173-49063173 on Build GRCh38
rs6019621 Chr.20: 49063426-49063426 on Build GRCh38
rs939318106 Chr.20: 49063408-49063408 on Build GRCh38
rs926550802 Chr.20: 49063406-49063406 on Build GRCh38
rs764135974 Chr.20: 49063152-49063152 on Build GRCh38
rs760367206 Chr.20: 49063151-49063151 on Build GRCh38
rs995936372 Chr.20: 49063332-49063332 on Build GRCh38
rs201335585 Chr.20: 49063126-49063126 on Build GRCh38
rs781779719 Chr.20: 49063193-49063193 on Build GRCh38
rs757283076 Chr.20: 49063385-49063385 on Build GRCh38
rs766094789 Chr.20: 49063311-49063311 on Build GRCh38
rs537727778 Chr.20: 49063323-49063323 on Build GRCh38
rs751159883 Chr.20: 49063324-49063324 on Build GRCh38
rs996991575 Chr.20: 49063135-49063135 on Build GRCh38
rs148567834 Chr.20: 49063439-49063439 on Build GRCh38
rs145805814 Chr.20: 49063343-49063343 on Build GRCh38
rs1037792170 Chr.20: 49063279-49063279 on Build GRCh38
rs776558691 Chr.20: 49063291-49063291 on Build GRCh38
rs749428437 Chr.20: 49063382-49063382 on Build GRCh38
rs551647183 Chr.20: 49063524-49063524 on Build GRCh38
rs191560041 Chr.20: 49063127-49063127 on Build GRCh38
rs1004007057 Chr.20: 49063120-49063120 on Build GRCh38
rs6095418 Chr.20: 49063097-49063097 on Build GRCh38
rs140201989 Chr.20: 49063558-49063558 on Build GRCh38
rs545334710 Chr.20: 49063236-49063236 on Build GRCh38
rs1010063182 Chr.20: 49063398-49063398 on Build GRCh38
rs1000092651 Chr.20: 49063461-49063461 on Build GRCh38
rs543446236 Chr.20: 49063102-49063102 on Build GRCh38
rs11479646 Chr.20: 49063119-49063119 on Build GRCh38
rs979155245 Chr.20: 49063127-49063128 on Build GRCh38
rs939672203 Chr.20: 49063462-49063462 on Build GRCh38
rs780437710 Chr.20: 49063530-49063530 on Build GRCh38
rs577663851 Chr.20: 49063411-49063411 on Build GRCh38
rs751097526 Chr.20: 49063278-49063278 on Build GRCh38
rs941729274 Chr.20: 49063266-49063266 on Build GRCh38
rs774408375 Chr.20: 49063259-49063259 on Build GRCh38
rs369371888 Chr.20: 49063218-49063218 on Build GRCh38
rs2734901 Chr.20: 49063109-49063109 on Build GRCh38
rs1030184099 Chr.20: 49063109-49063110 on Build GRCh38
rs770000216 Chr.20: 49063216-49063216 on Build GRCh38
rs898695851 Chr.20: 49063130-49063130 on Build GRCh38
rs756643059 Chr.20: 49063359-49063359 on Build GRCh38
rs567220880 Chr.20: 49063357-49063357 on Build GRCh38
rs980534430 Chr.20: 49063376-49063376 on Build GRCh38
rs2664535 Chr.20: 49063219-49063219 on Build GRCh38
rs753767865 Chr.20: 49063153-49063153 on Build GRCh38
rs2664556 Chr.20: 49063315-49063315 on Build GRCh38
rs764757989 Chr.20: 49063305-49063305 on Build GRCh38
rs886768797 Chr.20: 49063117-49063117 on Build GRCh38
rs955573543 Chr.20: 49063181-49063181 on Build GRCh38
rs2734900 Chr.20: 49063107-49063107 on Build GRCh38
rs533402440 Chr.20: 49063147-49063147 on Build GRCh38
rs552831409 Chr.20: 49063389-49063389 on Build GRCh38
rs761368891 Chr.20: 49063155-49063155 on Build GRCh38
rs200630338 Chr.20: 49063561-49063561 on Build GRCh38
rs767926801 Chr.20: 49063345-49063345 on Build GRCh38
rs547440630 Chr.20: 49063129-49063129 on Build GRCh38

View Full Product Details