Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 50596000-50596516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM65C
Entrez Gene ID
140876
Gene Name
family with sequence similarity 65 member C
Gene Aliases
C20orf175, C20orf176
UniGene
Hs.372578
Gene Chromosome Location
Chr.20: 50586108-50692004 on Build GRCh38
Transcripts
NM_001290268.1 NM_080829.3 NR_110890.1 XM_005260294.3 XM_006723713.3 XM_011528578.2 XM_011528579.2 XM_011528580.2 XM_011528581.2 XM_011528584.2 XM_011528585.2 XM_011528586.2 XM_017027681.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6236149 c.1786C>T p.R596W Chr.20: 50596256-50596256 on Build GRCh38
5443050 c.1818G>C p.A606A Chr.20: 50596224-50596224 on Build GRCh38
5040097 c.1790C>T p.P597L Chr.20: 50596252-50596252 on Build GRCh38
1307516 c.1866G>A p.L622L Chr.20: 50596176-50596176 on Build GRCh38
6660035 c.1871T>C p.V624A Chr.20: 50596171-50596171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570701658 Chr.20: 50596389-50596389 on Build GRCh38
rs989780222 Chr.20: 50596381-50596381 on Build GRCh38
rs978231501 Chr.20: 50596354-50596354 on Build GRCh38
rs377276576 Chr.20: 50596243-50596243 on Build GRCh38
rs970324353 Chr.20: 50596102-50596102 on Build GRCh38
rs950721000 Chr.20: 50596463-50596463 on Build GRCh38
rs770229488 Chr.20: 50596250-50596250 on Build GRCh38
rs747296395 Chr.20: 50596269-50596273 on Build GRCh38
rs759748109 Chr.20: 50596271-50596271 on Build GRCh38
rs1001778974 Chr.20: 50596385-50596385 on Build GRCh38
rs993935175 Chr.20: 50596258-50596258 on Build GRCh38
rs6063535 Chr.20: 50596384-50596384 on Build GRCh38
rs771258806 Chr.20: 50596281-50596282 on Build GRCh38
rs778280983 Chr.20: 50596119-50596119 on Build GRCh38
rs577491042 Chr.20: 50596107-50596107 on Build GRCh38
rs372472260 Chr.20: 50596224-50596224 on Build GRCh38
rs556905789 Chr.20: 50596090-50596090 on Build GRCh38
rs773729956 Chr.20: 50596251-50596251 on Build GRCh38
rs755953536 Chr.20: 50596203-50596203 on Build GRCh38
rs1016799507 Chr.20: 50596475-50596475 on Build GRCh38
rs189232949 Chr.20: 50596225-50596225 on Build GRCh38
rs149518874 Chr.20: 50596476-50596476 on Build GRCh38
rs771635715 Chr.20: 50596139-50596139 on Build GRCh38
rs150387614 Chr.20: 50596067-50596067 on Build GRCh38
rs891561427 Chr.20: 50596081-50596081 on Build GRCh38
rs930067160 Chr.20: 50596445-50596445 on Build GRCh38
rs1049240095 Chr.20: 50596046-50596046 on Build GRCh38
rs766540580 Chr.20: 50596190-50596190 on Build GRCh38
rs976562747 Chr.20: 50596065-50596065 on Build GRCh38
rs549738284 Chr.20: 50596439-50596439 on Build GRCh38
rs1036057713 Chr.20: 50596169-50596169 on Build GRCh38
rs373141664 Chr.20: 50596240-50596240 on Build GRCh38
rs1042648261 Chr.20: 50596075-50596075 on Build GRCh38
rs777729833 Chr.20: 50596105-50596105 on Build GRCh38
rs761546880 Chr.20: 50596297-50596297 on Build GRCh38
rs747842460 Chr.20: 50596225-50596226 on Build GRCh38
rs769107803 Chr.20: 50596158-50596158 on Build GRCh38
rs544985389 Chr.20: 50596032-50596032 on Build GRCh38
rs994059815 Chr.20: 50596452-50596452 on Build GRCh38
rs555201747 Chr.20: 50596033-50596033 on Build GRCh38
rs373812762 Chr.20: 50596196-50596196 on Build GRCh38
rs376576553 Chr.20: 50596431-50596431 on Build GRCh38
rs373756359 Chr.20: 50596101-50596102 on Build GRCh38
rs76945848 Chr.20: 50596210-50596210 on Build GRCh38
rs1029442258 Chr.20: 50596346-50596346 on Build GRCh38
rs541419603 Chr.20: 50596035-50596035 on Build GRCh38
rs535448403 Chr.20: 50596485-50596485 on Build GRCh38
rs765713430 Chr.20: 50596184-50596184 on Build GRCh38
rs747405056 Chr.20: 50596151-50596151 on Build GRCh38
rs1005433558 Chr.20: 50596290-50596290 on Build GRCh38
rs372985228 Chr.20: 50596239-50596239 on Build GRCh38
rs757881867 Chr.20: 50596236-50596236 on Build GRCh38
rs929340081 Chr.20: 50596045-50596045 on Build GRCh38
rs544572870 Chr.20: 50596109-50596109 on Build GRCh38
rs181547850 Chr.20: 50596383-50596383 on Build GRCh38
rs888314154 Chr.20: 50596477-50596477 on Build GRCh38
rs1005423888 Chr.20: 50596484-50596484 on Build GRCh38
rs1052679026 Chr.20: 50596079-50596079 on Build GRCh38
rs888325507 Chr.20: 50596282-50596282 on Build GRCh38
rs368476611 Chr.20: 50596302-50596302 on Build GRCh38
rs948126174 Chr.20: 50596188-50596188 on Build GRCh38
rs964101492 Chr.20: 50596059-50596059 on Build GRCh38
rs748469823 Chr.20: 50596249-50596249 on Build GRCh38
rs780079472 Chr.20: 50596378-50596378 on Build GRCh38
rs997176187 Chr.20: 50596073-50596073 on Build GRCh38
rs771568258 Chr.20: 50596240-50596242 on Build GRCh38
rs199573464 Chr.20: 50596370-50596370 on Build GRCh38
rs752458733 Chr.20: 50596091-50596091 on Build GRCh38
rs116264080 Chr.20: 50596261-50596261 on Build GRCh38
rs909345823 Chr.20: 50596058-50596058 on Build GRCh38
rs564876086 Chr.20: 50596202-50596202 on Build GRCh38
rs369019040 Chr.20: 50596208-50596208 on Build GRCh38
rs117579288 Chr.20: 50596098-50596098 on Build GRCh38
rs765113769 Chr.20: 50596301-50596301 on Build GRCh38
rs936870366 Chr.20: 50596369-50596369 on Build GRCh38
rs1022992298 Chr.20: 50596469-50596477 on Build GRCh38
rs967132536 Chr.20: 50596347-50596347 on Build GRCh38
rs376015124 Chr.20: 50596293-50596293 on Build GRCh38
rs756817507 Chr.20: 50596117-50596117 on Build GRCh38
rs765333419 Chr.20: 50596327-50596327 on Build GRCh38
rs766461315 Chr.20: 50596255-50596255 on Build GRCh38
rs763041379 Chr.20: 50596185-50596185 on Build GRCh38
rs773648496 Chr.20: 50596242-50596243 on Build GRCh38
rs6126047 Chr.20: 50596140-50596140 on Build GRCh38
rs767937393 Chr.20: 50596069-50596069 on Build GRCh38
rs767658021 Chr.20: 50596284-50596284 on Build GRCh38
rs762407181 Chr.20: 50596171-50596171 on Build GRCh38
rs924106219 Chr.20: 50596366-50596366 on Build GRCh38
rs376080684 Chr.20: 50596226-50596226 on Build GRCh38
rs745318441 Chr.20: 50596127-50596127 on Build GRCh38
rs369926463 Chr.20: 50596166-50596166 on Build GRCh38
rs1025546671 Chr.20: 50596457-50596457 on Build GRCh38

View Full Product Details