Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 57508522-57509020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTCFL
Entrez Gene ID
140690
Gene Name
CCCTC-binding factor like
Gene Aliases
BORIS, CT27, CTCF-T, HMGB1L1, dJ579F20.2
UniGene
Hs.131543
Gene Chromosome Location
Chr.20: 57495965-57525652 on Build GRCh38
Transcripts
NM_001269040.1 NM_001269041.1 NM_001269042.1 NM_001269043.1 NM_001269044.1 NM_001269045.1 NM_001269046.1 NM_001269047.1 NM_001269048.1 NM_001269049.1 NM_001269050.1 NM_001269054.1 NM_001269055.1 NM_080618.3 NR_072975.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4675316 c.1570C>T p.R524* Chr.20: 57508710-57508710 on Build GRCh38
5529359 c.1623C>T p.F541F Chr.20: 57508657-57508657 on Build GRCh38
1028372 c.1529G>C p.G510A Chr.20: 57508751-57508751 on Build GRCh38
4865698 c.1517G>A p.R506H Chr.20: 57508763-57508763 on Build GRCh38
6637321 c.1553C>A p.S518Y Chr.20: 57508727-57508727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010130113 Chr.20: 57508846-57508846 on Build GRCh38
rs758083037 Chr.20: 57508840-57508840 on Build GRCh38
rs376165241 Chr.20: 57508709-57508709 on Build GRCh38
rs750141556 Chr.20: 57508735-57508735 on Build GRCh38
rs755993131 Chr.20: 57508586-57508586 on Build GRCh38
rs542745691 Chr.20: 57508923-57508923 on Build GRCh38
rs145270601 Chr.20: 57508651-57508651 on Build GRCh38
rs764104357 Chr.20: 57508811-57508811 on Build GRCh38
rs747605658 Chr.20: 57508652-57508652 on Build GRCh38
rs770156635 Chr.20: 57508687-57508687 on Build GRCh38
rs1044219352 Chr.20: 57508595-57508595 on Build GRCh38
rs974384203 Chr.20: 57508715-57508715 on Build GRCh38
rs548199457 Chr.20: 57508853-57508853 on Build GRCh38
rs368864885 Chr.20: 57508729-57508729 on Build GRCh38
rs545059271 Chr.20: 57508610-57508610 on Build GRCh38
rs773504302 Chr.20: 57508689-57508689 on Build GRCh38
rs781496791 Chr.20: 57508764-57508764 on Build GRCh38
rs766627448 Chr.20: 57508701-57508701 on Build GRCh38
rs373069243 Chr.20: 57508774-57508774 on Build GRCh38
rs749570710 Chr.20: 57508785-57508785 on Build GRCh38
rs748531204 Chr.20: 57508773-57508773 on Build GRCh38
rs201864645 Chr.20: 57508728-57508728 on Build GRCh38
rs748728619 Chr.20: 57508942-57508942 on Build GRCh38
rs1018766451 Chr.20: 57508915-57508915 on Build GRCh38
rs202196411 Chr.20: 57508669-57508669 on Build GRCh38
rs750192478 Chr.20: 57508839-57508839 on Build GRCh38
rs188483123 Chr.20: 57508668-57508668 on Build GRCh38
rs758031990 Chr.20: 57508736-57508736 on Build GRCh38
rs571598928 Chr.20: 57508558-57508559 on Build GRCh38
rs765522244 Chr.20: 57508577-57508577 on Build GRCh38
rs528127737 Chr.20: 57508829-57508829 on Build GRCh38
rs118045133 Chr.20: 57508924-57508924 on Build GRCh38
rs754543077 Chr.20: 57508836-57508837 on Build GRCh38
rs773100098 Chr.20: 57508557-57508557 on Build GRCh38
rs537826940 Chr.20: 57508809-57508809 on Build GRCh38
rs766399688 Chr.20: 57508562-57508562 on Build GRCh38
rs761733126 Chr.20: 57508837-57508837 on Build GRCh38
rs776798507 Chr.20: 57508649-57508649 on Build GRCh38
rs143715840 Chr.20: 57508838-57508838 on Build GRCh38
rs376316532 Chr.20: 57508799-57508799 on Build GRCh38
rs976205670 Chr.20: 57508969-57508969 on Build GRCh38
rs1023401648 Chr.20: 57508565-57508565 on Build GRCh38
rs61754540 Chr.20: 57508611-57508611 on Build GRCh38
rs553400354 Chr.20: 57508939-57508939 on Build GRCh38
rs771028507 Chr.20: 57508795-57508795 on Build GRCh38
rs1032420895 Chr.20: 57508917-57508917 on Build GRCh38
rs755537284 Chr.20: 57508763-57508763 on Build GRCh38
rs922040561 Chr.20: 57508723-57508723 on Build GRCh38
rs767570664 Chr.20: 57508582-57508582 on Build GRCh38
rs752751540 Chr.20: 57508585-57508585 on Build GRCh38
rs895550042 Chr.20: 57508576-57508576 on Build GRCh38
rs573376986 Chr.20: 57508946-57508946 on Build GRCh38
rs891825694 Chr.20: 57508666-57508666 on Build GRCh38
rs779659992 Chr.20: 57508737-57508737 on Build GRCh38
rs755954544 Chr.20: 57508906-57508906 on Build GRCh38
rs1014760876 Chr.20: 57508547-57508547 on Build GRCh38
rs891092845 Chr.20: 57508864-57508864 on Build GRCh38
rs73625093 Chr.20: 57508717-57508717 on Build GRCh38
rs369048841 Chr.20: 57508797-57508797 on Build GRCh38
rs983095777 Chr.20: 57508743-57508743 on Build GRCh38
rs140188763 Chr.20: 57508781-57508781 on Build GRCh38
rs992334627 Chr.20: 57508980-57508980 on Build GRCh38
rs565083368 Chr.20: 57508619-57508619 on Build GRCh38
rs552301120 Chr.20: 57508754-57508754 on Build GRCh38
rs546579292 Chr.20: 57508673-57508673 on Build GRCh38
rs756732065 Chr.20: 57508869-57508869 on Build GRCh38
rs1051104986 Chr.20: 57508857-57508857 on Build GRCh38
rs55924405 Chr.20: 57508550-57508550 on Build GRCh38
rs916914876 Chr.20: 57508983-57508983 on Build GRCh38
rs762141672 Chr.20: 57508562-57508562 on Build GRCh38
rs1010223704 Chr.20: 57508677-57508677 on Build GRCh38
rs971032921 Chr.20: 57508988-57508988 on Build GRCh38
rs142133416 Chr.20: 57508690-57508690 on Build GRCh38
rs753717262 Chr.20: 57508593-57508596 on Build GRCh38
rs1025785959 Chr.20: 57508961-57508961 on Build GRCh38
rs750556671 Chr.20: 57508578-57508578 on Build GRCh38
rs6070122 Chr.20: 57508707-57508707 on Build GRCh38
rs61754539 Chr.20: 57508767-57508767 on Build GRCh38
rs745395497 Chr.20: 57508639-57508639 on Build GRCh38
rs754550079 Chr.20: 57508962-57508962 on Build GRCh38
rs61746345 Chr.20: 57508664-57508664 on Build GRCh38
rs771502445 Chr.20: 57508640-57508640 on Build GRCh38
rs763072684 Chr.20: 57508580-57508580 on Build GRCh38
rs201732065 Chr.20: 57508803-57508803 on Build GRCh38
rs41304417 Chr.20: 57508817-57508817 on Build GRCh38

View Full Product Details