Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59720823-59721321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHACTR3
Entrez Gene ID
116154
Gene Name
phosphatase and actin regulator 3
Gene Aliases
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
UniGene
Hs.473218
Gene Chromosome Location
Chr.20: 59577497-59849829 on Build GRCh38
Transcripts
NM_001199505.1 NM_001199506.1 NM_001281507.1 NM_080672.4 NM_183244.1 NM_183246.1 XM_011528525.2 XM_011528526.2 XM_017027626.1 XM_017027627.1 XM_017027628.1 XM_017027629.1 XM_017027630.1 XM_017027631.1

SNP Detail

SNP ID Chromosome Location
rs1027248333 Chr.20: 59721241-59721241 on Build GRCh38
rs559274021 Chr.20: 59720915-59720915 on Build GRCh38
rs975562323 Chr.20: 59720880-59720880 on Build GRCh38
rs115377497 Chr.20: 59721153-59721153 on Build GRCh38
rs559794200 Chr.20: 59720852-59720853 on Build GRCh38
rs201940351 Chr.20: 59721097-59721097 on Build GRCh38
rs952815619 Chr.20: 59721274-59721274 on Build GRCh38
rs569679810 Chr.20: 59721238-59721238 on Build GRCh38
rs982093667 Chr.20: 59721214-59721214 on Build GRCh38
rs142594948 Chr.20: 59721009-59721009 on Build GRCh38
rs189659414 Chr.20: 59721121-59721121 on Build GRCh38
rs377670246 Chr.20: 59720938-59720938 on Build GRCh38
rs982986760 Chr.20: 59721286-59721286 on Build GRCh38
rs903115615 Chr.20: 59721073-59721073 on Build GRCh38
rs937320207 Chr.20: 59721287-59721287 on Build GRCh38
rs139542309 Chr.20: 59721180-59721180 on Build GRCh38
rs548286581 Chr.20: 59721235-59721235 on Build GRCh38
rs8124718 Chr.20: 59720955-59720955 on Build GRCh38
rs1049236721 Chr.20: 59720899-59720899 on Build GRCh38
rs574795804 Chr.20: 59720982-59720982 on Build GRCh38
rs1008222194 Chr.20: 59721165-59721165 on Build GRCh38
rs766056343 Chr.20: 59721099-59721099 on Build GRCh38
rs1057233820 Chr.20: 59721076-59721076 on Build GRCh38
rs928681802 Chr.20: 59721271-59721271 on Build GRCh38
rs987210561 Chr.20: 59720915-59720943 on Build GRCh38
rs529719904 Chr.20: 59721205-59721205 on Build GRCh38
rs112366866 Chr.20: 59721188-59721188 on Build GRCh38
rs922154678 Chr.20: 59720885-59720885 on Build GRCh38
rs78748349 Chr.20: 59720898-59720898 on Build GRCh38
rs530713580 Chr.20: 59721257-59721257 on Build GRCh38
rs908290827 Chr.20: 59721058-59721058 on Build GRCh38
rs1020044835 Chr.20: 59721039-59721039 on Build GRCh38
rs150560174 Chr.20: 59721137-59721137 on Build GRCh38
rs940961119 Chr.20: 59721173-59721173 on Build GRCh38
rs528927669 Chr.20: 59720897-59720897 on Build GRCh38
rs11698337 Chr.20: 59721295-59721295 on Build GRCh38
rs940889525 Chr.20: 59720927-59720927 on Build GRCh38
rs185737506 Chr.20: 59721118-59721118 on Build GRCh38
rs1027170733 Chr.20: 59721124-59721124 on Build GRCh38
rs892585529 Chr.20: 59721154-59721154 on Build GRCh38
rs984965755 Chr.20: 59721147-59721147 on Build GRCh38
rs931481905 Chr.20: 59720983-59720983 on Build GRCh38
rs73303186 Chr.20: 59721103-59721103 on Build GRCh38
rs907195210 Chr.20: 59720941-59720941 on Build GRCh38
rs931344551 Chr.20: 59721119-59721119 on Build GRCh38
rs79150986 Chr.20: 59720970-59720970 on Build GRCh38
rs975566733 Chr.20: 59720977-59720977 on Build GRCh38
rs201947368 Chr.20: 59720927-59720955 on Build GRCh38
rs546941731 Chr.20: 59721177-59721177 on Build GRCh38
rs760453534 Chr.20: 59720990-59720990 on Build GRCh38

View Full Product Details