Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62122487-62123022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSM14B
Entrez Gene ID
149986
Gene Name
LSM family member 14B
Gene Aliases
C20orf40, FAM61B, FT005, LSM13, RAP55B, bA11M20.3
UniGene
Hs.105379
Gene Chromosome Location
Chr.20: 62122454-62135378 on Build GRCh38
Transcripts
NM_144703.2 XM_005260302.3 XM_011528605.2 XM_011528606.2 XM_011528607.2 XM_011528608.2 XM_011528609.2 XM_011528611.2 XM_017027688.1 XM_017027689.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3405245 c.68G>A p.R23H Chr.20: 62122734-62122734 on Build GRCh38
7118754 c.120C>A p.L40L Chr.20: 62122786-62122786 on Build GRCh38
4851414 c.120C>T p.L40L Chr.20: 62122786-62122786 on Build GRCh38
4851413 c.120C>T p.L40L Chr.20: 62122786-62122786 on Build GRCh38
4851412 c.120C>T p.L40L Chr.20: 62122786-62122786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532983124 Chr.20: 62122963-62122963 on Build GRCh38
rs769000563 Chr.20: 62122814-62122814 on Build GRCh38
rs534724127 Chr.20: 62122900-62122900 on Build GRCh38
rs1053909285 Chr.20: 62122518-62122518 on Build GRCh38
rs942531525 Chr.20: 62122531-62122531 on Build GRCh38
rs1040885426 Chr.20: 62122536-62122536 on Build GRCh38
rs373503997 Chr.20: 62122836-62122836 on Build GRCh38
rs574861264 Chr.20: 62122934-62122934 on Build GRCh38
rs762227916 Chr.20: 62122829-62122829 on Build GRCh38
rs994300417 Chr.20: 62122965-62122965 on Build GRCh38
rs557685496 Chr.20: 62122966-62122966 on Build GRCh38
rs553300646 Chr.20: 62122901-62122901 on Build GRCh38
rs371351995 Chr.20: 62122807-62122807 on Build GRCh38
rs749926243 Chr.20: 62122724-62122724 on Build GRCh38
rs1030186800 Chr.20: 62122632-62122632 on Build GRCh38
rs952490721 Chr.20: 62122983-62122983 on Build GRCh38
rs901353032 Chr.20: 62122964-62122964 on Build GRCh38
rs922412571 Chr.20: 62122867-62122867 on Build GRCh38
rs775733891 Chr.20: 62122805-62122805 on Build GRCh38
rs911011749 Chr.20: 62122522-62122522 on Build GRCh38
rs971265170 Chr.20: 62122981-62122981 on Build GRCh38
rs1012308809 Chr.20: 62122956-62122956 on Build GRCh38
rs751216951 Chr.20: 62122750-62122750 on Build GRCh38
rs1016349420 Chr.20: 62122620-62122620 on Build GRCh38
rs754306347 Chr.20: 62122699-62122699 on Build GRCh38
rs1025122479 Chr.20: 62122657-62122657 on Build GRCh38
rs748030501 Chr.20: 62122774-62122774 on Build GRCh38
rs769866424 Chr.20: 62122797-62122797 on Build GRCh38
rs955392231 Chr.20: 62122824-62122824 on Build GRCh38
rs373282522 Chr.20: 62122661-62122661 on Build GRCh38
rs999739820 Chr.20: 62122982-62122982 on Build GRCh38
rs994468691 Chr.20: 62122568-62122569 on Build GRCh38
rs978786076 Chr.20: 62122614-62122637 on Build GRCh38
rs1046988892 Chr.20: 62122576-62122576 on Build GRCh38
rs776606579 Chr.20: 62122617-62122617 on Build GRCh38
rs368859449 Chr.20: 62122770-62122770 on Build GRCh38
rs375594851 Chr.20: 62122664-62122664 on Build GRCh38
rs774575602 Chr.20: 62122626-62122626 on Build GRCh38
rs958344329 Chr.20: 62122997-62122997 on Build GRCh38
rs753741719 Chr.20: 62122799-62122822 on Build GRCh38
rs780878614 Chr.20: 62122771-62122771 on Build GRCh38
rs762315898 Chr.20: 62122715-62122715 on Build GRCh38
rs1025400891 Chr.20: 62122973-62122973 on Build GRCh38
rs757908261 Chr.20: 62122726-62122726 on Build GRCh38
rs767775489 Chr.20: 62122652-62122652 on Build GRCh38
rs772093584 Chr.20: 62122803-62122803 on Build GRCh38
rs568285767 Chr.20: 62122582-62122582 on Build GRCh38
rs970931361 Chr.20: 62122693-62122693 on Build GRCh38
rs1023288334 Chr.20: 62122999-62122999 on Build GRCh38
rs766235825 Chr.20: 62122674-62122675 on Build GRCh38
rs895166786 Chr.20: 62122953-62122953 on Build GRCh38
rs978713481 Chr.20: 62122799-62122799 on Build GRCh38
rs754882836 Chr.20: 62122818-62122819 on Build GRCh38
rs550446145 Chr.20: 62122655-62122655 on Build GRCh38
rs1007123822 Chr.20: 62122955-62122955 on Build GRCh38
rs200772568 Chr.20: 62122777-62122777 on Build GRCh38
rs759774182 Chr.20: 62122637-62122637 on Build GRCh38
rs200295431 Chr.20: 62122800-62122800 on Build GRCh38
rs1031663964 Chr.20: 62122994-62122994 on Build GRCh38
rs535389394 Chr.20: 62122623-62122623 on Build GRCh38
rs1005665769 Chr.20: 62122984-62122984 on Build GRCh38
rs764672388 Chr.20: 62122722-62122722 on Build GRCh38
rs910960791 Chr.20: 62122815-62122815 on Build GRCh38
rs936360044 Chr.20: 62122509-62122509 on Build GRCh38
rs765826530 Chr.20: 62122842-62122842 on Build GRCh38
rs901079451 Chr.20: 62122559-62122559 on Build GRCh38
rs775615755 Chr.20: 62122653-62122653 on Build GRCh38
rs745911984 Chr.20: 62122802-62122802 on Build GRCh38
rs369734064 Chr.20: 62122830-62122830 on Build GRCh38
rs776996639 Chr.20: 62122825-62122825 on Build GRCh38
rs779722984 Chr.20: 62122729-62122729 on Build GRCh38
rs374825244 Chr.20: 62122813-62122813 on Build GRCh38
rs750964319 Chr.20: 62122831-62122831 on Build GRCh38
rs924588308 Chr.20: 62122889-62122899 on Build GRCh38
rs1006619155 Chr.20: 62122910-62122910 on Build GRCh38

View Full Product Details