Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85029018-85029526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCOLN3
Entrez Gene ID
55283
Gene Name
mucolipin 3
Gene Aliases
TRP-ML3, TRPML3
UniGene
Hs.535239
Gene Chromosome Location
Chr.1: 85018082-85048902 on Build GRCh38
Transcripts
NM_001253693.1 NM_018298.10 XM_005271003.1 XM_006710750.1 XM_011541739.2 XM_011541740.2 XM_011541741.2 XM_011541742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4560032 c.827G>A p.G276E Chr.1: 85029111-85029111 on Build GRCh38
7271041 c.733-10G>A p.? Chr.1: 85029215-85029215 on Build GRCh38
6710038 c.802G>T p.E268* Chr.1: 85029136-85029136 on Build GRCh38
6064116 c.735A>G p.I245M Chr.1: 85029203-85029203 on Build GRCh38
3361023 c.779T>A p.L260* Chr.1: 85029159-85029159 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576879678 Chr.1: 85029046-85029046 on Build GRCh38
rs371360127 Chr.1: 85029241-85029241 on Build GRCh38
rs148257730 Chr.1: 85029153-85029153 on Build GRCh38
rs367567148 Chr.1: 85029143-85029143 on Build GRCh38
rs1010848988 Chr.1: 85029051-85029051 on Build GRCh38
rs575592773 Chr.1: 85029135-85029135 on Build GRCh38
rs761409584 Chr.1: 85029234-85029234 on Build GRCh38
rs950986919 Chr.1: 85029347-85029347 on Build GRCh38
rs890958181 Chr.1: 85029166-85029166 on Build GRCh38
rs762721552 Chr.1: 85029219-85029219 on Build GRCh38
rs763781070 Chr.1: 85029224-85029224 on Build GRCh38
rs775419157 Chr.1: 85029203-85029203 on Build GRCh38
rs776598663 Chr.1: 85029073-85029073 on Build GRCh38
rs902354906 Chr.1: 85029130-85029130 on Build GRCh38
rs766468962 Chr.1: 85029065-85029065 on Build GRCh38
rs900563329 Chr.1: 85029210-85029210 on Build GRCh38
rs115330931 Chr.1: 85029409-85029409 on Build GRCh38
rs916590234 Chr.1: 85029501-85029501 on Build GRCh38
rs774693746 Chr.1: 85029173-85029173 on Build GRCh38
rs79302320 Chr.1: 85029495-85029495 on Build GRCh38
rs759530282 Chr.1: 85029185-85029185 on Build GRCh38
rs758203682 Chr.1: 85029149-85029149 on Build GRCh38
rs973632772 Chr.1: 85029298-85029298 on Build GRCh38
rs952363129 Chr.1: 85029242-85029242 on Build GRCh38
rs934974801 Chr.1: 85029475-85029475 on Build GRCh38
rs561648187 Chr.1: 85029184-85029184 on Build GRCh38
rs1015135701 Chr.1: 85029328-85029328 on Build GRCh38
rs920836609 Chr.1: 85029312-85029312 on Build GRCh38
rs903812170 Chr.1: 85029155-85029155 on Build GRCh38
rs763729713 Chr.1: 85029097-85029097 on Build GRCh38
rs539492503 Chr.1: 85029052-85029052 on Build GRCh38
rs540813750 Chr.1: 85029380-85029380 on Build GRCh38
rs1052317463 Chr.1: 85029162-85029162 on Build GRCh38
rs552598289 Chr.1: 85029477-85029477 on Build GRCh38
rs752775535 Chr.1: 85029098-85029098 on Build GRCh38
rs935238826 Chr.1: 85029156-85029156 on Build GRCh38
rs758047932 Chr.1: 85029096-85029096 on Build GRCh38
rs908027288 Chr.1: 85029060-85029060 on Build GRCh38
rs758107020 Chr.1: 85029385-85029385 on Build GRCh38
rs756733902 Chr.1: 85029125-85029125 on Build GRCh38
rs748279796 Chr.1: 85029177-85029177 on Build GRCh38
rs751123917 Chr.1: 85029100-85029100 on Build GRCh38
rs571939499 Chr.1: 85029223-85029223 on Build GRCh38
rs765015755 Chr.1: 85029198-85029198 on Build GRCh38
rs916295398 Chr.1: 85029120-85029120 on Build GRCh38
rs984105437 Chr.1: 85029296-85029296 on Build GRCh38
rs749838938 Chr.1: 85029240-85029240 on Build GRCh38
rs1006563874 Chr.1: 85029154-85029154 on Build GRCh38
rs916357446 Chr.1: 85029393-85029393 on Build GRCh38
rs1017911491 Chr.1: 85029284-85029284 on Build GRCh38
rs1022184574 Chr.1: 85029095-85029095 on Build GRCh38
rs559959341 Chr.1: 85029412-85029412 on Build GRCh38
rs767072284 Chr.1: 85029239-85029239 on Build GRCh38
rs924940663 Chr.1: 85029431-85029431 on Build GRCh38
rs868615954 Chr.1: 85029186-85029186 on Build GRCh38
rs375263420 Chr.1: 85029093-85029093 on Build GRCh38
rs755363903 Chr.1: 85029252-85029252 on Build GRCh38
rs772309074 Chr.1: 85029183-85029183 on Build GRCh38
rs555890379 Chr.1: 85029094-85029094 on Build GRCh38
rs759351209 Chr.1: 85029078-85029078 on Build GRCh38
rs961165307 Chr.1: 85029365-85029365 on Build GRCh38
rs769060467 Chr.1: 85029163-85029163 on Build GRCh38
rs532501267 Chr.1: 85029476-85029476 on Build GRCh38
rs928068449 Chr.1: 85029379-85029379 on Build GRCh38
rs138092736 Chr.1: 85029136-85029136 on Build GRCh38
rs983620667 Chr.1: 85029378-85029378 on Build GRCh38
rs756523202 Chr.1: 85029150-85029150 on Build GRCh38
rs112292890 Chr.1: 85029146-85029146 on Build GRCh38
rs756193641 Chr.1: 85029128-85029128 on Build GRCh38
rs751179077 Chr.1: 85029232-85029232 on Build GRCh38

View Full Product Details