Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85678627-85679159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNHIT6
Entrez Gene ID
54680
Gene Name
zinc finger HIT-type containing 6
Gene Aliases
BCD1, C1orf181, NY-BR-75
UniGene
Hs.5111
Gene Chromosome Location
Chr.1: 85649423-85708447 on Build GRCh38
Transcripts
NM_001170670.1 NM_017953.3 XM_011541614.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912718 c.1162C>T p.R388C Chr.1: 85678708-85678708 on Build GRCh38
6287258 c.1094C>A p.P365Q Chr.1: 85678776-85678776 on Build GRCh38
4564305 c.1119_1120CC>TT p.(=) Chr.1: 85678750-85678751 on Build GRCh38
5502267 c.1150G>A p.D384N Chr.1: 85678720-85678720 on Build GRCh38
287085 c.1148C>A p.S383Y Chr.1: 85678722-85678722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573984450 Chr.1: 85678794-85678795 on Build GRCh38
rs34953954 Chr.1: 85678803-85678803 on Build GRCh38
rs754618557 Chr.1: 85678819-85678819 on Build GRCh38
rs922137539 Chr.1: 85679053-85679054 on Build GRCh38
rs149609139 Chr.1: 85678755-85678755 on Build GRCh38
rs974601446 Chr.1: 85679040-85679040 on Build GRCh38
rs375751184 Chr.1: 85678687-85678687 on Build GRCh38
rs548447099 Chr.1: 85678836-85678836 on Build GRCh38
rs894186335 Chr.1: 85678811-85678811 on Build GRCh38
rs748797137 Chr.1: 85678817-85678822 on Build GRCh38
rs913455797 Chr.1: 85678918-85678922 on Build GRCh38
rs1049376311 Chr.1: 85678714-85678714 on Build GRCh38
rs773695324 Chr.1: 85678708-85678708 on Build GRCh38
rs748897149 Chr.1: 85678663-85678663 on Build GRCh38
rs559224820 Chr.1: 85679074-85679074 on Build GRCh38
rs755991724 Chr.1: 85678787-85678787 on Build GRCh38
rs567977933 Chr.1: 85678866-85678866 on Build GRCh38
rs970099658 Chr.1: 85678845-85678845 on Build GRCh38
rs772444339 Chr.1: 85678707-85678707 on Build GRCh38
rs377356475 Chr.1: 85678795-85678796 on Build GRCh38
rs779953754 Chr.1: 85678797-85678797 on Build GRCh38
rs569420320 Chr.1: 85679077-85679077 on Build GRCh38
rs776861673 Chr.1: 85678722-85678722 on Build GRCh38
rs780119577 Chr.1: 85678875-85678875 on Build GRCh38
rs928618716 Chr.1: 85678873-85678873 on Build GRCh38
rs539536922 Chr.1: 85678982-85678982 on Build GRCh38
rs1055570716 Chr.1: 85678808-85678808 on Build GRCh38
rs1042722319 Chr.1: 85678852-85678852 on Build GRCh38
rs183709897 Chr.1: 85679085-85679085 on Build GRCh38
rs570601611 Chr.1: 85678930-85678930 on Build GRCh38
rs760919705 Chr.1: 85678711-85678711 on Build GRCh38
rs753545138 Chr.1: 85678809-85678809 on Build GRCh38
rs563812038 Chr.1: 85678926-85678926 on Build GRCh38
rs951489280 Chr.1: 85679128-85679128 on Build GRCh38
rs1018459528 Chr.1: 85678981-85678981 on Build GRCh38
rs1008359087 Chr.1: 85678966-85678966 on Build GRCh38
rs768312958 Chr.1: 85678684-85678684 on Build GRCh38
rs138848779 Chr.1: 85678768-85678768 on Build GRCh38
rs761870614 Chr.1: 85678765-85678765 on Build GRCh38
rs34136739 Chr.1: 85678842-85678843 on Build GRCh38
rs752665126 Chr.1: 85678762-85678762 on Build GRCh38
rs527250241 Chr.1: 85678913-85678913 on Build GRCh38
rs142772868 Chr.1: 85679114-85679114 on Build GRCh38
rs748701193 Chr.1: 85678696-85678696 on Build GRCh38
rs531534554 Chr.1: 85678821-85678821 on Build GRCh38
rs901540055 Chr.1: 85678779-85678796 on Build GRCh38
rs139562514 Chr.1: 85678715-85678715 on Build GRCh38
rs932399475 Chr.1: 85679072-85679072 on Build GRCh38
rs372497028 Chr.1: 85678654-85678654 on Build GRCh38
rs890777733 Chr.1: 85679110-85679110 on Build GRCh38
rs960053212 Chr.1: 85678874-85678874 on Build GRCh38
rs150480369 Chr.1: 85678738-85678738 on Build GRCh38
rs192667862 Chr.1: 85678785-85678785 on Build GRCh38
rs533977110 Chr.1: 85678795-85678795 on Build GRCh38

View Full Product Details