Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86476369-86476868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA1
Entrez Gene ID
1179
Gene Name
chloride channel accessory 1
Gene Aliases
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1
UniGene
Hs.194659
Gene Chromosome Location
Chr.1: 86468843-86500294 on Build GRCh38
Transcripts
NM_001285.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667682 c.525C>A p.F175L Chr.1: 86476521-86476521 on Build GRCh38
3806026 c.484C>T p.R162* Chr.1: 86476480-86476480 on Build GRCh38
5509702 c.486A>T p.R162R Chr.1: 86476482-86476482 on Build GRCh38
1687989 c.533C>T p.S178F Chr.1: 86476529-86476529 on Build GRCh38
6454541 c.502G>A p.E168K Chr.1: 86476498-86476498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021870332 Chr.1: 86476654-86476654 on Build GRCh38
rs907840631 Chr.1: 86476507-86476507 on Build GRCh38
rs781182283 Chr.1: 86476448-86476448 on Build GRCh38
rs189212104 Chr.1: 86476480-86476480 on Build GRCh38
rs751235286 Chr.1: 86476532-86476532 on Build GRCh38
rs78185315 Chr.1: 86476638-86476638 on Build GRCh38
rs752282419 Chr.1: 86476521-86476521 on Build GRCh38
rs776166343 Chr.1: 86476487-86476487 on Build GRCh38
rs981607574 Chr.1: 86476635-86476635 on Build GRCh38
rs764804030 Chr.1: 86476511-86476511 on Build GRCh38
rs891188898 Chr.1: 86476452-86476452 on Build GRCh38
rs773390299 Chr.1: 86476481-86476481 on Build GRCh38
rs761097870 Chr.1: 86476488-86476488 on Build GRCh38
rs558168775 Chr.1: 86476771-86476771 on Build GRCh38
rs758104855 Chr.1: 86476408-86476408 on Build GRCh38
rs34132913 Chr.1: 86476785-86476786 on Build GRCh38
rs145578580 Chr.1: 86476501-86476501 on Build GRCh38
rs780966060 Chr.1: 86476601-86476611 on Build GRCh38
rs193067742 Chr.1: 86476843-86476843 on Build GRCh38
rs750329240 Chr.1: 86476541-86476541 on Build GRCh38
rs777256896 Chr.1: 86476497-86476497 on Build GRCh38
rs1044234401 Chr.1: 86476586-86476586 on Build GRCh38
rs754714906 Chr.1: 86476597-86476598 on Build GRCh38
rs550829888 Chr.1: 86476602-86476602 on Build GRCh38
rs374915401 Chr.1: 86476828-86476828 on Build GRCh38
rs763803155 Chr.1: 86476526-86476526 on Build GRCh38
rs748853961 Chr.1: 86476673-86476673 on Build GRCh38
rs772111712 Chr.1: 86476557-86476557 on Build GRCh38
rs999332824 Chr.1: 86476405-86476405 on Build GRCh38
rs777169467 Chr.1: 86476590-86476590 on Build GRCh38
rs747246916 Chr.1: 86476577-86476577 on Build GRCh38
rs137913059 Chr.1: 86476525-86476525 on Build GRCh38
rs748329812 Chr.1: 86476477-86476477 on Build GRCh38
rs199568332 Chr.1: 86476412-86476412 on Build GRCh38
rs745859335 Chr.1: 86476454-86476454 on Build GRCh38
rs764483721 Chr.1: 86476469-86476469 on Build GRCh38
rs752370356 Chr.1: 86476403-86476403 on Build GRCh38
rs779771582 Chr.1: 86476585-86476586 on Build GRCh38
rs963376096 Chr.1: 86476807-86476807 on Build GRCh38
rs778815697 Chr.1: 86476451-86476451 on Build GRCh38
rs538340938 Chr.1: 86476660-86476660 on Build GRCh38
rs2753386 Chr.1: 86476451-86476451 on Build GRCh38
rs746837266 Chr.1: 86476419-86476419 on Build GRCh38
rs746511583 Chr.1: 86476587-86476588 on Build GRCh38
rs369276458 Chr.1: 86476464-86476464 on Build GRCh38
rs990968412 Chr.1: 86476662-86476662 on Build GRCh38
rs770442703 Chr.1: 86476601-86476601 on Build GRCh38
rs923831537 Chr.1: 86476812-86476812 on Build GRCh38
rs757057706 Chr.1: 86476533-86476533 on Build GRCh38
rs770585923 Chr.1: 86476685-86476685 on Build GRCh38
rs182278149 Chr.1: 86476429-86476429 on Build GRCh38
rs2180764 Chr.1: 86476799-86476799 on Build GRCh38
rs778042733 Chr.1: 86476567-86476567 on Build GRCh38
rs529862538 Chr.1: 86476622-86476622 on Build GRCh38
rs778914498 Chr.1: 86476547-86476547 on Build GRCh38
rs146043808 Chr.1: 86476782-86476783 on Build GRCh38
rs781010387 Chr.1: 86476535-86476535 on Build GRCh38
rs1030533848 Chr.1: 86476683-86476683 on Build GRCh38
rs763483621 Chr.1: 86476556-86476557 on Build GRCh38
rs566181879 Chr.1: 86476651-86476651 on Build GRCh38
rs754309618 Chr.1: 86476540-86476543 on Build GRCh38
rs771259669 Chr.1: 86476489-86476489 on Build GRCh38
rs371500349 Chr.1: 86476550-86476550 on Build GRCh38
rs397757348 Chr.1: 86476791-86476792 on Build GRCh38
rs142663356 Chr.1: 86476542-86476542 on Build GRCh38
rs946345842 Chr.1: 86476787-86476787 on Build GRCh38
rs772814592 Chr.1: 86476585-86476585 on Build GRCh38
rs771296923 Chr.1: 86476588-86476588 on Build GRCh38
rs567370251 Chr.1: 86476607-86476607 on Build GRCh38
rs935264379 Chr.1: 86476838-86476838 on Build GRCh38
rs574151353 Chr.1: 86476393-86476393 on Build GRCh38
rs759964482 Chr.1: 86476778-86476778 on Build GRCh38
rs774957420 Chr.1: 86476704-86476704 on Build GRCh38
rs760074180 Chr.1: 86476591-86476591 on Build GRCh38
rs768697165 Chr.1: 86476459-86476459 on Build GRCh38

View Full Product Details