Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64264342-64264847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCMTD2
Entrez Gene ID
55251
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Gene Aliases
C20orf36
UniGene
Hs.744845
Gene Chromosome Location
Chr.20: 64255695-64276226 on Build GRCh38
Transcripts
NM_001104925.1 NM_018257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5889400 c.311C>T p.P104L Chr.20: 64264432-64264432 on Build GRCh38
3549294 c.325C>T p.H109Y Chr.20: 64264446-64264446 on Build GRCh38
4788160 c.391A>G p.T131A Chr.20: 64264512-64264512 on Build GRCh38
5889402 c.310C>T p.P104S Chr.20: 64264431-64264431 on Build GRCh38
5889399 c.311C>T p.P104L Chr.20: 64264432-64264432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs910221331 Chr.20: 64264410-64264410 on Build GRCh38
rs17569873 Chr.20: 64264757-64264757 on Build GRCh38
rs975023032 Chr.20: 64264624-64264624 on Build GRCh38
rs1046506597 Chr.20: 64264638-64264638 on Build GRCh38
rs766591541 Chr.20: 64264487-64264487 on Build GRCh38
rs181754850 Chr.20: 64264651-64264651 on Build GRCh38
rs747116048 Chr.20: 64264512-64264513 on Build GRCh38
rs753814481 Chr.20: 64264572-64264572 on Build GRCh38
rs772493305 Chr.20: 64264474-64264475 on Build GRCh38
rs369636774 Chr.20: 64264583-64264583 on Build GRCh38
rs548482058 Chr.20: 64264786-64264786 on Build GRCh38
rs867636625 Chr.20: 64264699-64264699 on Build GRCh38
rs907530015 Chr.20: 64264679-64264679 on Build GRCh38
rs770628080 Chr.20: 64264402-64264402 on Build GRCh38
rs933565750 Chr.20: 64264649-64264649 on Build GRCh38
rs1013523916 Chr.20: 64264637-64264637 on Build GRCh38
rs1032533044 Chr.20: 64264597-64264597 on Build GRCh38
rs73327716 Chr.20: 64264817-64264817 on Build GRCh38
rs1011505493 Chr.20: 64264697-64264697 on Build GRCh38
rs762334161 Chr.20: 64264447-64264447 on Build GRCh38
rs144292423 Chr.20: 64264796-64264796 on Build GRCh38
rs966813913 Chr.20: 64264529-64264529 on Build GRCh38
rs779272653 Chr.20: 64264407-64264412 on Build GRCh38
rs115849267 Chr.20: 64264662-64264662 on Build GRCh38
rs759068648 Chr.20: 64264541-64264541 on Build GRCh38
rs1025006289 Chr.20: 64264816-64264816 on Build GRCh38
rs975743453 Chr.20: 64264764-64264764 on Build GRCh38
rs755654163 Chr.20: 64264564-64264564 on Build GRCh38
rs770339089 Chr.20: 64264466-64264466 on Build GRCh38
rs902600368 Chr.20: 64264646-64264646 on Build GRCh38
rs114092956 Chr.20: 64264809-64264809 on Build GRCh38
rs749135302 Chr.20: 64264381-64264381 on Build GRCh38
rs368444080 Chr.20: 64264445-64264445 on Build GRCh38
rs6011374 Chr.20: 64264792-64264792 on Build GRCh38
rs752303139 Chr.20: 64264552-64264552 on Build GRCh38
rs768787975 Chr.20: 64264439-64264439 on Build GRCh38
rs958421432 Chr.20: 64264604-64264604 on Build GRCh38
rs866544051 Chr.20: 64264751-64264751 on Build GRCh38
rs940460984 Chr.20: 64264700-64264700 on Build GRCh38
rs748521662 Chr.20: 64264423-64264423 on Build GRCh38
rs780338190 Chr.20: 64264400-64264400 on Build GRCh38
rs35487633 Chr.20: 64264436-64264436 on Build GRCh38
rs763429037 Chr.20: 64264480-64264480 on Build GRCh38
rs771053347 Chr.20: 64264574-64264575 on Build GRCh38
rs372047375 Chr.20: 64264485-64264485 on Build GRCh38
rs1037828055 Chr.20: 64264725-64264725 on Build GRCh38
rs1014168931 Chr.20: 64264810-64264810 on Build GRCh38
rs1055748012 Chr.20: 64264622-64264622 on Build GRCh38
rs747144687 Chr.20: 64264424-64264424 on Build GRCh38
rs749216337 Chr.20: 64264644-64264644 on Build GRCh38
rs780382481 Chr.20: 64264422-64264422 on Build GRCh38
rs895150725 Chr.20: 64264470-64264470 on Build GRCh38
rs778338886 Chr.20: 64264499-64264501 on Build GRCh38
rs199660373 Chr.20: 64264510-64264510 on Build GRCh38
rs777925346 Chr.20: 64264566-64264566 on Build GRCh38
rs773700031 Chr.20: 64264476-64264476 on Build GRCh38
rs957409113 Chr.20: 64264664-64264664 on Build GRCh38

View Full Product Details