Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33545900-33546401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SON
Entrez Gene ID
6651
Gene Name
SON DNA binding protein
Gene Aliases
BASS1, C21orf50, DBP-5, NREBP, SON3
UniGene
Hs.517262
Gene Chromosome Location
Chr.21: 33543038-33577514 on Build GRCh38
Transcripts
NM_001291411.1 NM_001291412.1 NM_032195.2 NM_138927.2 NR_103797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4728979 c.161G>A p.R54K Chr.21: 33546296-33546296 on Build GRCh38
5343733 c.196G>C p.E66Q Chr.21: 33546331-33546331 on Build GRCh38
2843712 c.119C>A p.P40H Chr.21: 33546254-33546254 on Build GRCh38
4101317 c.148G>A p.A50T Chr.21: 33546283-33546283 on Build GRCh38
4101314 c.138G>A p.A46A Chr.21: 33546273-33546273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934215676 Chr.21: 33546195-33546195 on Build GRCh38
rs772231631 Chr.21: 33546263-33546263 on Build GRCh38
rs1053066050 Chr.21: 33546095-33546095 on Build GRCh38
rs57788567 Chr.21: 33546133-33546134 on Build GRCh38
rs548138111 Chr.21: 33545974-33545974 on Build GRCh38
rs921476123 Chr.21: 33545977-33545977 on Build GRCh38
rs946748439 Chr.21: 33545958-33545958 on Build GRCh38
rs202224123 Chr.21: 33546256-33546256 on Build GRCh38
rs779587376 Chr.21: 33546359-33546359 on Build GRCh38
rs1027778513 Chr.21: 33546228-33546228 on Build GRCh38
rs375956865 Chr.21: 33546019-33546019 on Build GRCh38
rs746341884 Chr.21: 33546364-33546364 on Build GRCh38
rs778263375 Chr.21: 33546318-33546318 on Build GRCh38
rs774256254 Chr.21: 33546170-33546170 on Build GRCh38
rs1011450450 Chr.21: 33546111-33546111 on Build GRCh38
rs775890214 Chr.21: 33546269-33546269 on Build GRCh38
rs749932303 Chr.21: 33546194-33546194 on Build GRCh38
rs988242730 Chr.21: 33546166-33546166 on Build GRCh38
rs1003428164 Chr.21: 33546142-33546142 on Build GRCh38
rs757933581 Chr.21: 33546286-33546286 on Build GRCh38
rs749860348 Chr.21: 33546250-33546250 on Build GRCh38
rs536267719 Chr.21: 33546273-33546273 on Build GRCh38
rs765348954 Chr.21: 33546168-33546168 on Build GRCh38
rs1050707255 Chr.21: 33546113-33546113 on Build GRCh38
rs751273876 Chr.21: 33546309-33546309 on Build GRCh38
rs775689868 Chr.21: 33546368-33546368 on Build GRCh38
rs201007642 Chr.21: 33546204-33546204 on Build GRCh38
rs761016170 Chr.21: 33546369-33546369 on Build GRCh38
rs147194840 Chr.21: 33546375-33546375 on Build GRCh38
rs761878500 Chr.21: 33546274-33546274 on Build GRCh38
rs771335229 Chr.21: 33546251-33546251 on Build GRCh38
rs774321034 Chr.21: 33546071-33546071 on Build GRCh38
rs1022837790 Chr.21: 33546169-33546169 on Build GRCh38
rs867628631 Chr.21: 33546304-33546304 on Build GRCh38
rs767444460 Chr.21: 33546178-33546178 on Build GRCh38
rs1018603383 Chr.21: 33546174-33546174 on Build GRCh38
rs956773048 Chr.21: 33546161-33546161 on Build GRCh38
rs898822630 Chr.21: 33546122-33546122 on Build GRCh38
rs774929003 Chr.21: 33546253-33546253 on Build GRCh38
rs145186427 Chr.21: 33546314-33546314 on Build GRCh38
rs771360849 Chr.21: 33546358-33546358 on Build GRCh38
rs750309161 Chr.21: 33546281-33546281 on Build GRCh38
rs74772689 Chr.21: 33546193-33546193 on Build GRCh38
rs569665458 Chr.21: 33545980-33545980 on Build GRCh38
rs1051233924 Chr.21: 33546021-33546021 on Build GRCh38
rs757119701 Chr.21: 33546171-33546171 on Build GRCh38
rs534769704 Chr.21: 33546225-33546225 on Build GRCh38
rs190703163 Chr.21: 33546035-33546035 on Build GRCh38
rs951921467 Chr.21: 33546240-33546240 on Build GRCh38
rs377031623 Chr.21: 33546372-33546372 on Build GRCh38
rs905096413 Chr.21: 33546141-33546141 on Build GRCh38
rs767349906 Chr.21: 33546189-33546190 on Build GRCh38
rs1036559926 Chr.21: 33546101-33546101 on Build GRCh38
rs746623397 Chr.21: 33546211-33546211 on Build GRCh38
rs779507097 Chr.21: 33546298-33546298 on Build GRCh38
rs369046723 Chr.21: 33546170-33546170 on Build GRCh38
rs576338135 Chr.21: 33546337-33546337 on Build GRCh38
rs761693573 Chr.21: 33546171-33546173 on Build GRCh38
rs765166211 Chr.21: 33546279-33546279 on Build GRCh38
rs1023261221 Chr.21: 33546138-33546138 on Build GRCh38
rs1034847986 Chr.21: 33546155-33546155 on Build GRCh38
rs760780913 Chr.21: 33546272-33546272 on Build GRCh38
rs11544794 Chr.21: 33546287-33546287 on Build GRCh38
rs770233859 Chr.21: 33546234-33546234 on Build GRCh38
rs994427236 Chr.21: 33546127-33546127 on Build GRCh38
rs76600419 Chr.21: 33546246-33546246 on Build GRCh38
rs779701724 Chr.21: 33546206-33546206 on Build GRCh38
rs574910734 Chr.21: 33546344-33546344 on Build GRCh38

View Full Product Details