Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33603325-33603727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772064 c.339delA p.K113fs*37 Chr.21: 33603530-33603530 on Build GRCh38
6451609 c.392G>A p.R131H Chr.21: 33603477-33603477 on Build GRCh38
3550420 c.434G>A p.G145E Chr.21: 33603435-33603435 on Build GRCh38
6441100 c.334C>T p.H112Y Chr.21: 33603535-33603535 on Build GRCh38
6161394 c.454G>A p.D152N Chr.21: 33603415-33603415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747461530 Chr.21: 33603409-33603409 on Build GRCh38
rs937103830 Chr.21: 33603577-33603577 on Build GRCh38
rs748576078 Chr.21: 33603417-33603417 on Build GRCh38
rs11702633 Chr.21: 33603544-33603544 on Build GRCh38
rs375050334 Chr.21: 33603372-33603372 on Build GRCh38
rs200366314 Chr.21: 33603510-33603510 on Build GRCh38
rs138891371 Chr.21: 33603540-33603540 on Build GRCh38
rs773512379 Chr.21: 33603534-33603534 on Build GRCh38
rs770289619 Chr.21: 33603520-33603520 on Build GRCh38
rs760105547 Chr.21: 33603439-33603440 on Build GRCh38
rs1036123893 Chr.21: 33603495-33603495 on Build GRCh38
rs144536410 Chr.21: 33603629-33603629 on Build GRCh38
rs753702638 Chr.21: 33603566-33603566 on Build GRCh38
rs756789044 Chr.21: 33603399-33603399 on Build GRCh38
rs766384406 Chr.21: 33603459-33603459 on Build GRCh38
rs749507174 Chr.21: 33603538-33603538 on Build GRCh38
rs752604352 Chr.21: 33603375-33603375 on Build GRCh38
rs755921026 Chr.21: 33603477-33603477 on Build GRCh38
rs143241458 Chr.21: 33603490-33603490 on Build GRCh38
rs776834306 Chr.21: 33603414-33603414 on Build GRCh38
rs775432984 Chr.21: 33603554-33603554 on Build GRCh38
rs185016106 Chr.21: 33603643-33603643 on Build GRCh38
rs771852201 Chr.21: 33603552-33603552 on Build GRCh38
rs745427523 Chr.21: 33603502-33603502 on Build GRCh38
rs368362573 Chr.21: 33603373-33603373 on Build GRCh38
rs559571829 Chr.21: 33603389-33603389 on Build GRCh38
rs868005845 Chr.21: 33603690-33603690 on Build GRCh38
rs867543284 Chr.21: 33603445-33603445 on Build GRCh38
rs1023717013 Chr.21: 33603551-33603551 on Build GRCh38
rs761343486 Chr.21: 33603574-33603574 on Build GRCh38
rs199612151 Chr.21: 33603561-33603561 on Build GRCh38
rs773404393 Chr.21: 33603367-33603367 on Build GRCh38
rs371681090 Chr.21: 33603397-33603397 on Build GRCh38
rs763694929 Chr.21: 33603478-33603478 on Build GRCh38
rs780539959 Chr.21: 33603408-33603408 on Build GRCh38
rs905226155 Chr.21: 33603619-33603619 on Build GRCh38
rs200401225 Chr.21: 33603483-33603483 on Build GRCh38
rs769274461 Chr.21: 33603413-33603413 on Build GRCh38
rs748484256 Chr.21: 33603515-33603515 on Build GRCh38
rs148269698 Chr.21: 33603509-33603509 on Build GRCh38
rs1056760833 Chr.21: 33603606-33603606 on Build GRCh38
rs752409601 Chr.21: 33603471-33603471 on Build GRCh38
rs530660636 Chr.21: 33603368-33603368 on Build GRCh38
rs534562132 Chr.21: 33603437-33603437 on Build GRCh38
rs374956495 Chr.21: 33603443-33603443 on Build GRCh38
rs570419689 Chr.21: 33603416-33603416 on Build GRCh38
rs778285191 Chr.21: 33603501-33603501 on Build GRCh38
rs765599614 Chr.21: 33603364-33603364 on Build GRCh38
rs753490551 Chr.21: 33603489-33603489 on Build GRCh38
rs759385737 Chr.21: 33603467-33603467 on Build GRCh38
rs553893063 Chr.21: 33603468-33603468 on Build GRCh38
rs940544395 Chr.21: 33603480-33603480 on Build GRCh38
rs770093300 Chr.21: 33603428-33603428 on Build GRCh38
rs763062343 Chr.21: 33603455-33603455 on Build GRCh38
rs372356394 Chr.21: 33603542-33603542 on Build GRCh38
rs192465556 Chr.21: 33603392-33603392 on Build GRCh38
rs774545898 Chr.21: 33603460-33603460 on Build GRCh38
rs777447701 Chr.21: 33603396-33603396 on Build GRCh38
rs765856540 Chr.21: 33603439-33603442 on Build GRCh38

View Full Product Details