Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 39405638-39406173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCA5L
Entrez Gene ID
150082
Gene Name
LCA5L, lebercilin like
Gene Aliases
C21orf13
UniGene
Hs.517284
Gene Chromosome Location
Chr.21: 39405844-39445796 on Build GRCh38
Transcripts
NM_152505.3 XM_005260926.1 XM_006723967.1 XM_006723972.1 XM_006723973.1 XM_011529458.1 XM_011529459.1 XM_011529460.1 XM_011529461.1 XM_011529462.1 XM_011529463.2 XM_011529464.1 XM_011529465.1 XM_011529466.1 XM_011529467.1 XM_011529468.1 XM_011529469.2 XM_011529470.1 XM_011529471.1 XM_017028274.1 XM_017028275.1 XM_017028276.1 XM_017028277.1 XM_017028278.1 NM_001317744.1

Gene Symbol
WRB-SH3BGR
Entrez Gene ID
106865373
Gene Name
WRB-SH3BGR readthrough
Gene Aliases
-
UniGene
Hs.473847
Gene Chromosome Location
Chr.21: 39380244-39515506 on Build GRCh38
Transcripts
NM_152505.3 XM_005260926.1 XM_006723967.1 XM_006723972.1 XM_006723973.1 XM_011529458.1 XM_011529459.1 XM_011529460.1 XM_011529461.1 XM_011529462.1 XM_011529463.2 XM_011529464.1 XM_011529465.1 XM_011529466.1 XM_011529467.1 XM_011529468.1 XM_011529469.2 XM_011529470.1 XM_011529471.1 XM_017028274.1 XM_017028275.1 XM_017028276.1 XM_017028277.1 XM_017028278.1 NM_001317744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304711 c.1921A>C p.T641P Chr.21: 39405974-39405974 on Build GRCh38
5392896 c.1862G>A p.G621D Chr.21: 39406033-39406033 on Build GRCh38
1030737 c.1801G>T p.E601* Chr.21: 39406094-39406094 on Build GRCh38
6581590 c.1775G>A p.R592K Chr.21: 39406120-39406120 on Build GRCh38
1751642 c.1867G>A p.E623K Chr.21: 39406028-39406028 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989661772 Chr.21: 39405731-39405731 on Build GRCh38
rs200434082 Chr.21: 39406074-39406074 on Build GRCh38
rs763644598 Chr.21: 39406047-39406047 on Build GRCh38
rs766031527 Chr.21: 39405880-39405880 on Build GRCh38
rs767052502 Chr.21: 39406030-39406030 on Build GRCh38
rs140751076 Chr.21: 39406002-39406002 on Build GRCh38
rs779475051 Chr.21: 39405899-39405899 on Build GRCh38
rs541735399 Chr.21: 39405732-39405735 on Build GRCh38
rs138510761 Chr.21: 39406032-39406032 on Build GRCh38
rs748330039 Chr.21: 39406080-39406080 on Build GRCh38
rs771856856 Chr.21: 39406113-39406113 on Build GRCh38
rs770758889 Chr.21: 39406100-39406100 on Build GRCh38
rs747262941 Chr.21: 39405978-39405978 on Build GRCh38
rs765018970 Chr.21: 39405862-39405862 on Build GRCh38
rs1005363541 Chr.21: 39405717-39405717 on Build GRCh38
rs774011644 Chr.21: 39406013-39406013 on Build GRCh38
rs1022702799 Chr.21: 39405674-39405674 on Build GRCh38
rs180865456 Chr.21: 39406056-39406056 on Build GRCh38
rs780510799 Chr.21: 39405889-39405889 on Build GRCh38
rs777762689 Chr.21: 39406089-39406089 on Build GRCh38
rs199513705 Chr.21: 39406001-39406001 on Build GRCh38
rs143233766 Chr.21: 39406099-39406099 on Build GRCh38
rs371039624 Chr.21: 39405693-39405693 on Build GRCh38
rs751237165 Chr.21: 39405943-39405943 on Build GRCh38
rs747281653 Chr.21: 39405736-39405736 on Build GRCh38
rs769708081 Chr.21: 39405931-39405931 on Build GRCh38
rs376768499 Chr.21: 39405729-39405729 on Build GRCh38
rs184085067 Chr.21: 39405888-39405888 on Build GRCh38
rs772449001 Chr.21: 39406086-39406087 on Build GRCh38
rs192250283 Chr.21: 39405831-39405831 on Build GRCh38
rs761677740 Chr.21: 39405842-39405842 on Build GRCh38
rs768759449 Chr.21: 39405980-39405980 on Build GRCh38
rs1057349551 Chr.21: 39405929-39405929 on Build GRCh38
rs776279254 Chr.21: 39406138-39406138 on Build GRCh38
rs748205431 Chr.21: 39405986-39405986 on Build GRCh38
rs966903998 Chr.21: 39406107-39406107 on Build GRCh38
rs537211892 Chr.21: 39405694-39405694 on Build GRCh38
rs754372619 Chr.21: 39405894-39405894 on Build GRCh38
rs776511481 Chr.21: 39405918-39405918 on Build GRCh38
rs759289340 Chr.21: 39406026-39406026 on Build GRCh38
rs758817274 Chr.21: 39405975-39405975 on Build GRCh38
rs202032441 Chr.21: 39405868-39405868 on Build GRCh38
rs766049126 Chr.21: 39405942-39405942 on Build GRCh38
rs769764729 Chr.21: 39405989-39405989 on Build GRCh38
rs372573617 Chr.21: 39406053-39406053 on Build GRCh38
rs772459450 Chr.21: 39405902-39405902 on Build GRCh38
rs142554637 Chr.21: 39405953-39405953 on Build GRCh38
rs775299650 Chr.21: 39406123-39406123 on Build GRCh38
rs773041334 Chr.21: 39405938-39405938 on Build GRCh38
rs760350193 Chr.21: 39406131-39406131 on Build GRCh38
rs760288961 Chr.21: 39406046-39406046 on Build GRCh38
rs780349892 Chr.21: 39405904-39405904 on Build GRCh38
rs200757955 Chr.21: 39406139-39406139 on Build GRCh38
rs757833776 Chr.21: 39405898-39405898 on Build GRCh38
rs188814692 Chr.21: 39405947-39405947 on Build GRCh38
rs201684934 Chr.21: 39405974-39405974 on Build GRCh38
rs976672564 Chr.21: 39405736-39405739 on Build GRCh38
rs773355307 Chr.21: 39406089-39406089 on Build GRCh38
rs145985071 Chr.21: 39406007-39406007 on Build GRCh38
rs768798095 Chr.21: 39405912-39405912 on Build GRCh38
rs755278588 Chr.21: 39406059-39406059 on Build GRCh38
rs959698074 Chr.21: 39406144-39406144 on Build GRCh38
rs779527952 Chr.21: 39405970-39405970 on Build GRCh38
rs756208932 Chr.21: 39406082-39406082 on Build GRCh38
rs896046023 Chr.21: 39405934-39405934 on Build GRCh38
rs201718230 Chr.21: 39405919-39405919 on Build GRCh38
rs1044021126 Chr.21: 39405966-39405966 on Build GRCh38
rs780228277 Chr.21: 39405977-39405977 on Build GRCh38
rs897685259 Chr.21: 39405925-39405925 on Build GRCh38
rs576997134 Chr.21: 39405800-39405800 on Build GRCh38
rs759042256 Chr.21: 39405950-39405950 on Build GRCh38
rs545842016 Chr.21: 39405824-39405824 on Build GRCh38
rs200492515 Chr.21: 39405910-39405910 on Build GRCh38
rs781393455 Chr.21: 39405982-39405982 on Build GRCh38
rs370552152 Chr.21: 39405940-39405940 on Build GRCh38
rs187408707 Chr.21: 39405726-39405726 on Build GRCh38
rs1030755435 Chr.21: 39405757-39405757 on Build GRCh38
rs767876735 Chr.21: 39405797-39405797 on Build GRCh38
rs544366531 Chr.21: 39405960-39405960 on Build GRCh38
rs150754131 Chr.21: 39406110-39406110 on Build GRCh38
rs750984933 Chr.21: 39406051-39406051 on Build GRCh38
rs752239600 Chr.21: 39405959-39405959 on Build GRCh38
rs766837129 Chr.21: 39406054-39406054 on Build GRCh38
rs370925430 Chr.21: 39406132-39406132 on Build GRCh38
rs1015400089 Chr.21: 39406068-39406068 on Build GRCh38
rs144695121 Chr.21: 39406003-39406003 on Build GRCh38
rs774249732 Chr.21: 39406108-39406108 on Build GRCh38
rs552928783 Chr.21: 39405867-39405867 on Build GRCh38

View Full Product Details