Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43658937-43659426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF2BP
Entrez Gene ID
11077
Gene Name
heat shock transcription factor 2 binding protein
Gene Aliases
-
UniGene
Hs.406157
Gene Chromosome Location
Chr.21: 43453987-43659499 on Build GRCh38
Transcripts
NM_007031.1 XM_011529443.2 XM_011529445.2 XM_011529446.2 XM_011529447.2 XM_017028267.1 XM_017028268.1 XM_017028269.1 XM_017028270.1 XM_017028271.1 XM_017028272.1

SNP Detail

SNP ID Chromosome Location
rs982280635 Chr.21: 43659028-43659028 on Build GRCh38
rs1034454512 Chr.21: 43659329-43659329 on Build GRCh38
rs1054538021 Chr.21: 43659166-43659166 on Build GRCh38
rs984958755 Chr.21: 43659125-43659125 on Build GRCh38
rs1033263290 Chr.21: 43658959-43658959 on Build GRCh38
rs1040498242 Chr.21: 43659151-43659151 on Build GRCh38
rs985459582 Chr.21: 43659362-43659362 on Build GRCh38
rs940725957 Chr.21: 43659147-43659147 on Build GRCh38
rs953977017 Chr.21: 43658980-43658980 on Build GRCh38
rs1028244395 Chr.21: 43658958-43658958 on Build GRCh38
rs537600856 Chr.21: 43659224-43659224 on Build GRCh38
rs971003605 Chr.21: 43659295-43659295 on Build GRCh38
rs940840353 Chr.21: 43659379-43659379 on Build GRCh38
rs568133748 Chr.21: 43659263-43659263 on Build GRCh38
rs916885704 Chr.21: 43659062-43659062 on Build GRCh38
rs1034809641 Chr.21: 43659333-43659333 on Build GRCh38
rs952315705 Chr.21: 43659359-43659359 on Build GRCh38
rs969466194 Chr.21: 43659300-43659300 on Build GRCh38
rs398101651 Chr.21: 43659148-43659149 on Build GRCh38
rs868590720 Chr.21: 43659115-43659115 on Build GRCh38
rs1033301171 Chr.21: 43658996-43658996 on Build GRCh38
rs75734677 Chr.21: 43659145-43659146 on Build GRCh38
rs957610119 Chr.21: 43658976-43658976 on Build GRCh38
rs553667979 Chr.21: 43659165-43659165 on Build GRCh38
rs539354032 Chr.21: 43659401-43659401 on Build GRCh38
rs969055702 Chr.21: 43659026-43659026 on Build GRCh38
rs1032833578 Chr.21: 43659235-43659235 on Build GRCh38
rs183170300 Chr.21: 43659302-43659302 on Build GRCh38
rs372539880 Chr.21: 43659047-43659047 on Build GRCh38
rs989189428 Chr.21: 43658988-43658988 on Build GRCh38
rs893411645 Chr.21: 43659189-43659189 on Build GRCh38
rs191099710 Chr.21: 43659127-43659127 on Build GRCh38
rs1050448778 Chr.21: 43659173-43659173 on Build GRCh38
rs1000547591 Chr.21: 43659226-43659226 on Build GRCh38
rs889710368 Chr.21: 43659202-43659202 on Build GRCh38
rs971921560 Chr.21: 43659399-43659399 on Build GRCh38
rs971057508 Chr.21: 43659066-43659066 on Build GRCh38
rs529267599 Chr.21: 43659049-43659049 on Build GRCh38
rs575829691 Chr.21: 43658966-43658966 on Build GRCh38
rs547449224 Chr.21: 43659076-43659076 on Build GRCh38
rs549916764 Chr.21: 43659243-43659243 on Build GRCh38
rs748886408 Chr.21: 43659029-43659029 on Build GRCh38
rs903236926 Chr.21: 43659150-43659150 on Build GRCh38
rs535507266 Chr.21: 43659290-43659290 on Build GRCh38
rs920547636 Chr.21: 43659101-43659101 on Build GRCh38
rs111732134 Chr.21: 43659406-43659406 on Build GRCh38
rs186225916 Chr.21: 43658995-43658995 on Build GRCh38
rs2051407 Chr.21: 43659364-43659364 on Build GRCh38
rs570264300 Chr.21: 43659190-43659190 on Build GRCh38
rs910563512 Chr.21: 43659119-43659119 on Build GRCh38
rs572798096 Chr.21: 43659118-43659118 on Build GRCh38
rs561807379 Chr.21: 43659030-43659030 on Build GRCh38
rs931975908 Chr.21: 43659171-43659171 on Build GRCh38
rs771854563 Chr.21: 43659231-43659231 on Build GRCh38
rs571966330 Chr.21: 43659400-43659400 on Build GRCh38
rs140892572 Chr.21: 43659350-43659350 on Build GRCh38
rs1009918577 Chr.21: 43659269-43659269 on Build GRCh38
rs559680085 Chr.21: 43659105-43659105 on Build GRCh38

View Full Product Details