Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23763988-23764485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf15
Entrez Gene ID
150248
Gene Name
chromosome 22 open reading frame 15
Gene Aliases
N27C7-3
UniGene
Hs.116254
Gene Chromosome Location
Chr.22: 23762522-23765863 on Build GRCh38
Transcripts
NM_182520.2 XM_011529907.2 XM_011529908.2 XM_011529912.2 XM_017028602.1 XM_017028603.1 XM_017028604.1 XM_017028605.1 XM_017028606.1 XM_017028607.1 XM_017028608.1 XM_017028609.1 XM_017028610.1 XM_017028611.1 XM_017028612.1 XM_017028613.1 XM_017028614.1 XM_017028615.1 XM_017028616.1 XM_017028617.1 XM_017028618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587347 c.86G>A p.R29K Chr.22: 23764147-23764147 on Build GRCh38
5627103 c.127C>A p.L43M Chr.22: 23764274-23764274 on Build GRCh38
5587348 c.86G>A p.R29K Chr.22: 23764147-23764147 on Build GRCh38
5627105 c.128T>C p.L43P Chr.22: 23764275-23764275 on Build GRCh38
5644297 c.59G>T p.R20M Chr.22: 23764120-23764120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569128310 Chr.22: 23764161-23764161 on Build GRCh38
rs766665749 Chr.22: 23764384-23764384 on Build GRCh38
rs1037817239 Chr.22: 23764426-23764426 on Build GRCh38
rs868349679 Chr.22: 23764368-23764368 on Build GRCh38
rs114546436 Chr.22: 23764357-23764357 on Build GRCh38
rs776914289 Chr.22: 23764450-23764450 on Build GRCh38
rs531244604 Chr.22: 23764410-23764410 on Build GRCh38
rs779941168 Chr.22: 23764437-23764437 on Build GRCh38
rs989285930 Chr.22: 23764349-23764349 on Build GRCh38
rs755979823 Chr.22: 23764432-23764432 on Build GRCh38
rs1003808195 Chr.22: 23764204-23764204 on Build GRCh38
rs776696250 Chr.22: 23764385-23764385 on Build GRCh38
rs1047658065 Chr.22: 23764402-23764402 on Build GRCh38
rs765301842 Chr.22: 23764389-23764389 on Build GRCh38
rs369504865 Chr.22: 23764331-23764331 on Build GRCh38
rs553969002 Chr.22: 23764266-23764266 on Build GRCh38
rs202059299 Chr.22: 23764233-23764233 on Build GRCh38
rs554608942 Chr.22: 23764229-23764229 on Build GRCh38
rs745480957 Chr.22: 23764424-23764424 on Build GRCh38
rs910270148 Chr.22: 23764100-23764100 on Build GRCh38
rs749086029 Chr.22: 23764439-23764439 on Build GRCh38
rs780674358 Chr.22: 23764337-23764337 on Build GRCh38
rs906352397 Chr.22: 23764181-23764181 on Build GRCh38
rs546796565 Chr.22: 23764455-23764455 on Build GRCh38
rs765516563 Chr.22: 23764390-23764390 on Build GRCh38
rs143257161 Chr.22: 23764319-23764319 on Build GRCh38
rs763269485 Chr.22: 23764463-23764463 on Build GRCh38
rs780917800 Chr.22: 23764423-23764423 on Build GRCh38
rs766497620 Chr.22: 23764125-23764125 on Build GRCh38
rs769666206 Chr.22: 23764365-23764365 on Build GRCh38
rs2298375 Chr.22: 23764261-23764261 on Build GRCh38
rs374808343 Chr.22: 23764260-23764260 on Build GRCh38
rs775895018 Chr.22: 23764459-23764459 on Build GRCh38
rs947710051 Chr.22: 23764064-23764064 on Build GRCh38
rs538706829 Chr.22: 23764097-23764097 on Build GRCh38
rs1005004159 Chr.22: 23764217-23764217 on Build GRCh38
rs181949101 Chr.22: 23764222-23764222 on Build GRCh38
rs943147059 Chr.22: 23764112-23764112 on Build GRCh38
rs561470788 Chr.22: 23764448-23764448 on Build GRCh38
rs201685549 Chr.22: 23764375-23764375 on Build GRCh38
rs773400859 Chr.22: 23764377-23764377 on Build GRCh38
rs1021591639 Chr.22: 23764034-23764034 on Build GRCh38
rs924776569 Chr.22: 23764092-23764092 on Build GRCh38
rs541798369 Chr.22: 23764418-23764418 on Build GRCh38
rs770869070 Chr.22: 23764449-23764449 on Build GRCh38
rs373636672 Chr.22: 23764405-23764405 on Build GRCh38
rs752383841 Chr.22: 23764099-23764100 on Build GRCh38
rs1031907612 Chr.22: 23764309-23764309 on Build GRCh38
rs933516282 Chr.22: 23764096-23764096 on Build GRCh38
rs3216082 Chr.22: 23764344-23764345 on Build GRCh38
rs372494840 Chr.22: 23764251-23764251 on Build GRCh38
rs370671273 Chr.22: 23764379-23764379 on Build GRCh38
rs756696554 Chr.22: 23764422-23764422 on Build GRCh38
rs758387734 Chr.22: 23764220-23764220 on Build GRCh38
rs1037338074 Chr.22: 23764137-23764137 on Build GRCh38
rs201792455 Chr.22: 23764149-23764149 on Build GRCh38
rs768706003 Chr.22: 23764443-23764443 on Build GRCh38
rs991873943 Chr.22: 23764027-23764027 on Build GRCh38
rs1047710090 Chr.22: 23764163-23764163 on Build GRCh38
rs777278205 Chr.22: 23764264-23764264 on Build GRCh38
rs758961216 Chr.22: 23764413-23764413 on Build GRCh38
rs987308712 Chr.22: 23764267-23764267 on Build GRCh38
rs886568694 Chr.22: 23764215-23764215 on Build GRCh38
rs899256991 Chr.22: 23764444-23764444 on Build GRCh38
rs1044678023 Chr.22: 23764080-23764080 on Build GRCh38
rs977684663 Chr.22: 23764091-23764091 on Build GRCh38
rs977630736 Chr.22: 23764299-23764299 on Build GRCh38
rs775602012 Chr.22: 23764367-23764367 on Build GRCh38
rs566624602 Chr.22: 23764039-23764039 on Build GRCh38
rs530281324 Chr.22: 23764453-23764453 on Build GRCh38
rs759923935 Chr.22: 23764388-23764388 on Build GRCh38
rs558924782 Chr.22: 23764131-23764131 on Build GRCh38

View Full Product Details