Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24585989-24586483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT1
Entrez Gene ID
2678
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
CD224, D22S672, D22S732, GGT, GGT 1, GTG
UniGene
Hs.595809
Gene Chromosome Location
Chr.22: 24583750-24629005 on Build GRCh38
Transcripts
NM_013430.2 NM_207644.2

Gene Symbol
LRRC75B
Entrez Gene ID
388886
Gene Name
leucine rich repeat containing 75B
Gene Aliases
C22orf36, FAM211B
UniGene
Hs.113314
Gene Chromosome Location
Chr.22: 24585620-24593104 on Build GRCh38
Transcripts
NM_013430.2 NM_207644.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044240 c.539C>A p.A180D Chr.22: 24586295-24586295 on Build GRCh38
1236350 c.625C>T p.R209C Chr.22: 24586209-24586209 on Build GRCh38
5558799 c.544C>T p.L182L Chr.22: 24586290-24586290 on Build GRCh38
1415257 c.838C>A p.L280M Chr.22: 24585996-24585996 on Build GRCh38
6040240 c.728G>A p.W243* Chr.22: 24586106-24586106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763748632 Chr.22: 24586201-24586201 on Build GRCh38
rs781489695 Chr.22: 24586259-24586259 on Build GRCh38
rs749128361 Chr.22: 24586170-24586170 on Build GRCh38
rs200740419 Chr.22: 24586186-24586186 on Build GRCh38
rs753558973 Chr.22: 24586308-24586308 on Build GRCh38
rs745778486 Chr.22: 24586023-24586023 on Build GRCh38
rs111248278 Chr.22: 24586213-24586213 on Build GRCh38
rs769285417 Chr.22: 24586280-24586280 on Build GRCh38
rs376304947 Chr.22: 24586316-24586316 on Build GRCh38
rs201635194 Chr.22: 24586135-24586135 on Build GRCh38
rs767777626 Chr.22: 24586028-24586028 on Build GRCh38
rs370955070 Chr.22: 24586171-24586171 on Build GRCh38
rs746794016 Chr.22: 24586017-24586017 on Build GRCh38
rs558891096 Chr.22: 24586291-24586291 on Build GRCh38
rs199729018 Chr.22: 24586037-24586037 on Build GRCh38
rs998943312 Chr.22: 24586456-24586457 on Build GRCh38
rs926596214 Chr.22: 24586196-24586196 on Build GRCh38
rs200284399 Chr.22: 24586050-24586050 on Build GRCh38
rs142329748 Chr.22: 24586043-24586043 on Build GRCh38
rs369810020 Chr.22: 24586237-24586237 on Build GRCh38
rs755802773 Chr.22: 24586165-24586165 on Build GRCh38
rs746150678 Chr.22: 24586324-24586324 on Build GRCh38
rs755962423 Chr.22: 24586221-24586221 on Build GRCh38
rs1024763203 Chr.22: 24586302-24586302 on Build GRCh38
rs773442492 Chr.22: 24586099-24586099 on Build GRCh38
rs756154971 Chr.22: 24586399-24586399 on Build GRCh38
rs189889936 Chr.22: 24586265-24586265 on Build GRCh38
rs767725445 Chr.22: 24586112-24586120 on Build GRCh38
rs369178498 Chr.22: 24586323-24586323 on Build GRCh38
rs201525843 Chr.22: 24586157-24586157 on Build GRCh38
rs779688498 Chr.22: 24586232-24586232 on Build GRCh38
rs748629418 Chr.22: 24586338-24586338 on Build GRCh38
rs766242740 Chr.22: 24586292-24586292 on Build GRCh38
rs773358848 Chr.22: 24586033-24586033 on Build GRCh38
rs767695838 Chr.22: 24586374-24586374 on Build GRCh38
rs981249363 Chr.22: 24586326-24586326 on Build GRCh38
rs1020020732 Chr.22: 24586086-24586086 on Build GRCh38
rs747794525 Chr.22: 24586010-24586010 on Build GRCh38
rs201901553 Chr.22: 24586333-24586333 on Build GRCh38
rs781103130 Chr.22: 24586066-24586066 on Build GRCh38
rs201864155 Chr.22: 24586403-24586403 on Build GRCh38
rs367666780 Chr.22: 24586179-24586179 on Build GRCh38
rs747084853 Chr.22: 24586242-24586242 on Build GRCh38
rs750230451 Chr.22: 24586163-24586163 on Build GRCh38
rs200554342 Chr.22: 24586025-24586025 on Build GRCh38
rs559720233 Chr.22: 24586073-24586073 on Build GRCh38
rs34602598 Chr.22: 24586011-24586012 on Build GRCh38
rs1041495697 Chr.22: 24586225-24586225 on Build GRCh38
rs973345991 Chr.22: 24586344-24586344 on Build GRCh38
rs924614918 Chr.22: 24586184-24586184 on Build GRCh38
rs753078171 Chr.22: 24586131-24586131 on Build GRCh38
rs565446062 Chr.22: 24586041-24586041 on Build GRCh38
rs372731342 Chr.22: 24586329-24586329 on Build GRCh38
rs1047552768 Chr.22: 24586236-24586236 on Build GRCh38
rs752672322 Chr.22: 24586369-24586369 on Build GRCh38
rs763658650 Chr.22: 24586139-24586139 on Build GRCh38
rs772442486 Chr.22: 24586436-24586436 on Build GRCh38
rs748709259 Chr.22: 24586435-24586435 on Build GRCh38
rs749870028 Chr.22: 24586269-24586269 on Build GRCh38
rs774605663 Chr.22: 24586332-24586332 on Build GRCh38
rs182487665 Chr.22: 24586285-24586285 on Build GRCh38
rs746612172 Chr.22: 24586051-24586051 on Build GRCh38
rs776733478 Chr.22: 24586021-24586021 on Build GRCh38
rs1013102670 Chr.22: 24586412-24586412 on Build GRCh38
rs577321478 Chr.22: 24586317-24586317 on Build GRCh38
rs754974407 Chr.22: 24586309-24586309 on Build GRCh38
rs201574647 Chr.22: 24586095-24586095 on Build GRCh38
rs77407491 Chr.22: 24586169-24586169 on Build GRCh38
rs559186944 Chr.22: 24586022-24586022 on Build GRCh38
rs774470745 Chr.22: 24586026-24586026 on Build GRCh38
rs376853405 Chr.22: 24586263-24586263 on Build GRCh38
rs542238538 Chr.22: 24586372-24586372 on Build GRCh38
rs1031841551 Chr.22: 24586457-24586457 on Build GRCh38
rs771322335 Chr.22: 24586106-24586106 on Build GRCh38
rs772944925 Chr.22: 24586290-24586290 on Build GRCh38
rs1051972366 Chr.22: 24586392-24586392 on Build GRCh38
rs41277317 Chr.22: 24586015-24586015 on Build GRCh38
rs140576121 Chr.22: 24586427-24586428 on Build GRCh38
rs777594887 Chr.22: 24586038-24586038 on Build GRCh38
rs767286236 Chr.22: 24586216-24586216 on Build GRCh38
rs373358711 Chr.22: 24586452-24586452 on Build GRCh38
rs777797143 Chr.22: 24586172-24586172 on Build GRCh38
rs763522678 Chr.22: 24586195-24586195 on Build GRCh38
rs757151379 Chr.22: 24586057-24586057 on Build GRCh38
rs185762510 Chr.22: 24586174-24586174 on Build GRCh38
rs755429130 Chr.22: 24586429-24586429 on Build GRCh38
rs774935055 Chr.22: 24586282-24586282 on Build GRCh38
rs201174249 Chr.22: 24586367-24586367 on Build GRCh38
rs369099942 Chr.22: 24586349-24586349 on Build GRCh38
rs756337986 Chr.22: 24586385-24586386 on Build GRCh38
rs41277319 Chr.22: 24586286-24586286 on Build GRCh38
rs368977922 Chr.22: 24586044-24586044 on Build GRCh38
rs375545821 Chr.22: 24586178-24586178 on Build GRCh38
rs760017541 Chr.22: 24586126-24586126 on Build GRCh38
rs541407518 Chr.22: 24586348-24586348 on Build GRCh38
rs200007713 Chr.22: 24586377-24586377 on Build GRCh38
rs759339445 Chr.22: 24586360-24586360 on Build GRCh38
rs776288075 Chr.22: 24586458-24586458 on Build GRCh38
rs757661772 Chr.22: 24586087-24586089 on Build GRCh38
rs367928623 Chr.22: 24586208-24586208 on Build GRCh38
rs369496594 Chr.22: 24586264-24586264 on Build GRCh38
rs570699672 Chr.22: 24586460-24586460 on Build GRCh38
rs201882935 Chr.22: 24586342-24586342 on Build GRCh38
rs939134912 Chr.22: 24586385-24586385 on Build GRCh38
rs16978737 Chr.22: 24586158-24586158 on Build GRCh38
rs754068863 Chr.22: 24586405-24586405 on Build GRCh38
rs746531808 Chr.22: 24586241-24586241 on Build GRCh38
rs373932134 Chr.22: 24586040-24586040 on Build GRCh38
rs778931197 Chr.22: 24586081-24586081 on Build GRCh38
rs370633505 Chr.22: 24586337-24586337 on Build GRCh38
rs776957554 Chr.22: 24586110-24586110 on Build GRCh38
rs746504984 Chr.22: 24586457-24586457 on Build GRCh38
rs780110739 Chr.22: 24586435-24586436 on Build GRCh38
rs750706509 Chr.22: 24586302-24586302 on Build GRCh38
rs1039836722 Chr.22: 24586439-24586439 on Build GRCh38
rs201243203 Chr.22: 24586226-24586226 on Build GRCh38
rs770792630 Chr.22: 24586018-24586018 on Build GRCh38
rs371457436 Chr.22: 24586185-24586185 on Build GRCh38
rs202090927 Chr.22: 24586013-24586013 on Build GRCh38
rs376541606 Chr.22: 24586027-24586027 on Build GRCh38
rs1017968803 Chr.22: 24586255-24586255 on Build GRCh38
rs531315896 Chr.22: 24586459-24586459 on Build GRCh38
rs202119783 Chr.22: 24586130-24586130 on Build GRCh38
rs769761626 Chr.22: 24586074-24586074 on Build GRCh38
rs751299319 Chr.22: 24586143-24586143 on Build GRCh38
rs548708079 Chr.22: 24586102-24586102 on Build GRCh38
rs371589501 Chr.22: 24586258-24586258 on Build GRCh38
rs144754113 Chr.22: 24586087-24586087 on Build GRCh38
rs760431197 Chr.22: 24586343-24586343 on Build GRCh38
rs771648579 Chr.22: 24586340-24586340 on Build GRCh38
rs776573764 Chr.22: 24586138-24586138 on Build GRCh38
rs201884365 Chr.22: 24586430-24586430 on Build GRCh38
rs369950570 Chr.22: 24586217-24586217 on Build GRCh38

View Full Product Details