Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30121851-30122350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HORMAD2
Entrez Gene ID
150280
Gene Name
HORMA domain containing 2
Gene Aliases
CT46.2
UniGene
Hs.120391
Gene Chromosome Location
Chr.22: 30080069-30207425 on Build GRCh38
Transcripts
XM_011529914.2 XM_011529917.2 XM_011529918.1 XM_017028621.1 XM_017028622.1 XM_017028624.1 XM_017028625.1 XM_017028626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3553246 c.640G>A p.E214K Chr.22: 30122035-30122035 on Build GRCh38
5423417 c.808T>C p.C270R Chr.22: 30122203-30122203 on Build GRCh38
7002827 c.756C>G p.N252K Chr.22: 30122151-30122151 on Build GRCh38
3553247 c.782G>A p.G261D Chr.22: 30122177-30122177 on Build GRCh38
6406412 c.577C>T p.H193Y Chr.22: 30121972-30121972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139118242 Chr.22: 30122318-30122318 on Build GRCh38
rs9620950 Chr.22: 30122278-30122278 on Build GRCh38
rs544667629 Chr.22: 30122037-30122037 on Build GRCh38
rs994732663 Chr.22: 30121883-30121883 on Build GRCh38
rs781102280 Chr.22: 30122023-30122023 on Build GRCh38
rs778426997 Chr.22: 30122015-30122015 on Build GRCh38
rs377699164 Chr.22: 30121971-30121971 on Build GRCh38
rs746417049 Chr.22: 30122033-30122035 on Build GRCh38
rs918907667 Chr.22: 30122279-30122279 on Build GRCh38
rs1004134984 Chr.22: 30122287-30122287 on Build GRCh38
rs761939996 Chr.22: 30122176-30122176 on Build GRCh38
rs768049903 Chr.22: 30122079-30122079 on Build GRCh38
rs750845659 Chr.22: 30122080-30122080 on Build GRCh38
rs753504565 Chr.22: 30122111-30122111 on Build GRCh38
rs562288347 Chr.22: 30122254-30122254 on Build GRCh38
rs868369820 Chr.22: 30121936-30121936 on Build GRCh38
rs746953399 Chr.22: 30122245-30122245 on Build GRCh38
rs370648839 Chr.22: 30122158-30122158 on Build GRCh38
rs8141471 Chr.22: 30122244-30122244 on Build GRCh38
rs944442321 Chr.22: 30122241-30122241 on Build GRCh38
rs200673594 Chr.22: 30121960-30121960 on Build GRCh38
rs768896132 Chr.22: 30122172-30122172 on Build GRCh38
rs776521322 Chr.22: 30122166-30122166 on Build GRCh38
rs571663410 Chr.22: 30122224-30122224 on Build GRCh38
rs751619978 Chr.22: 30122059-30122059 on Build GRCh38
rs746479925 Chr.22: 30122019-30122019 on Build GRCh38
rs780331899 Chr.22: 30122107-30122107 on Build GRCh38
rs374354454 Chr.22: 30122160-30122160 on Build GRCh38
rs775847470 Chr.22: 30122045-30122045 on Build GRCh38
rs757736771 Chr.22: 30121965-30121965 on Build GRCh38
rs1027646568 Chr.22: 30122249-30122249 on Build GRCh38
rs759867884 Chr.22: 30121917-30121917 on Build GRCh38
rs544508760 Chr.22: 30122265-30122265 on Build GRCh38
rs761113672 Chr.22: 30122181-30122181 on Build GRCh38
rs377080525 Chr.22: 30122046-30122046 on Build GRCh38
rs762271431 Chr.22: 30122068-30122068 on Build GRCh38
rs529588747 Chr.22: 30122261-30122261 on Build GRCh38
rs1034843609 Chr.22: 30122070-30122070 on Build GRCh38
rs181903593 Chr.22: 30122177-30122177 on Build GRCh38
rs764919673 Chr.22: 30122220-30122220 on Build GRCh38
rs200230533 Chr.22: 30122144-30122144 on Build GRCh38
rs374023457 Chr.22: 30121908-30121908 on Build GRCh38
rs563082244 Chr.22: 30122295-30122295 on Build GRCh38
rs772919728 Chr.22: 30122033-30122033 on Build GRCh38
rs765521931 Chr.22: 30121929-30121929 on Build GRCh38
rs750772616 Chr.22: 30121985-30121985 on Build GRCh38
rs556300456 Chr.22: 30121951-30121951 on Build GRCh38
rs888481111 Chr.22: 30122267-30122267 on Build GRCh38
rs752869153 Chr.22: 30121930-30121930 on Build GRCh38
rs368563029 Chr.22: 30122167-30122167 on Build GRCh38
rs960114360 Chr.22: 30122084-30122084 on Build GRCh38
rs757916924 Chr.22: 30122234-30122234 on Build GRCh38
rs755814054 Chr.22: 30121990-30121990 on Build GRCh38
rs375998548 Chr.22: 30122147-30122147 on Build GRCh38
rs952695632 Chr.22: 30121907-30121907 on Build GRCh38
rs373673977 Chr.22: 30122230-30122230 on Build GRCh38
rs766750542 Chr.22: 30122186-30122186 on Build GRCh38
rs757118057 Chr.22: 30122008-30122008 on Build GRCh38
rs745872662 Chr.22: 30122073-30122073 on Build GRCh38
rs747662890 Chr.22: 30122143-30122143 on Build GRCh38
rs930574012 Chr.22: 30122294-30122294 on Build GRCh38
rs374167647 Chr.22: 30121959-30121959 on Build GRCh38
rs770193283 Chr.22: 30122199-30122200 on Build GRCh38
rs926152711 Chr.22: 30122211-30122211 on Build GRCh38
rs754574814 Chr.22: 30122121-30122121 on Build GRCh38
rs372564784 Chr.22: 30122041-30122041 on Build GRCh38
rs370003836 Chr.22: 30122030-30122030 on Build GRCh38
rs770465970 Chr.22: 30122022-30122023 on Build GRCh38
rs374552122 Chr.22: 30122146-30122146 on Build GRCh38
rs756466677 Chr.22: 30122099-30122099 on Build GRCh38
rs777169658 Chr.22: 30122024-30122025 on Build GRCh38
rs760519985 Chr.22: 30122034-30122034 on Build GRCh38
rs778315866 Chr.22: 30122136-30122136 on Build GRCh38
rs748056519 Chr.22: 30122020-30122020 on Build GRCh38
rs748865572 Chr.22: 30121995-30121995 on Build GRCh38
rs751369307 Chr.22: 30121991-30121991 on Build GRCh38
rs370498674 Chr.22: 30122047-30122047 on Build GRCh38
rs779651508 Chr.22: 30122215-30122215 on Build GRCh38

View Full Product Details