Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31768431-31768929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC5
Entrez Gene ID
9681
Gene Name
DEP domain containing 5
Gene Aliases
DEP.5, FFEVF
UniGene
Hs.435022
Gene Chromosome Location
Chr.22: 31753951-31907034 on Build GRCh38
Transcripts
NM_001007188.2 NM_001136029.2 NM_001242896.1 NM_001242897.1 NM_014662.4 NR_110988.1 XM_005261862.1 XM_011530557.2 XM_011530558.2 XM_011530559.2 XM_011530560.2 XM_011530561.2 XM_011530562.2 XM_011530563.2 XM_011530565.2 XM_011530568.2 XM_017029109.1 XM_017029110.1 XM_017029111.1 XM_017029112.1 XM_017029113.1 XM_017029114.1 XM_017029115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6369569 c.412A>G p.R138G Chr.22: 31768862-31768862 on Build GRCh38
7003376 c.399G>A p.E133E Chr.22: 31768849-31768849 on Build GRCh38
7003378 c.399G>A p.E133E Chr.22: 31768849-31768849 on Build GRCh38
6369566 c.412A>G p.R138G Chr.22: 31768862-31768862 on Build GRCh38
5019714 c.364-10C>T p.? Chr.22: 31768804-31768804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016417394 Chr.22: 31768704-31768704 on Build GRCh38
rs374099895 Chr.22: 31768788-31768788 on Build GRCh38
rs1028798905 Chr.22: 31768509-31768509 on Build GRCh38
rs372091064 Chr.22: 31768892-31768892 on Build GRCh38
rs1018789848 Chr.22: 31768529-31768529 on Build GRCh38
rs753858770 Chr.22: 31768777-31768777 on Build GRCh38
rs116263417 Chr.22: 31768804-31768804 on Build GRCh38
rs951982957 Chr.22: 31768891-31768891 on Build GRCh38
rs758593350 Chr.22: 31768887-31768887 on Build GRCh38
rs769733038 Chr.22: 31768769-31768770 on Build GRCh38
rs757280636 Chr.22: 31768893-31768893 on Build GRCh38
rs182106402 Chr.22: 31768734-31768734 on Build GRCh38
rs958272141 Chr.22: 31768862-31768862 on Build GRCh38
rs748450091 Chr.22: 31768869-31768869 on Build GRCh38
rs1027326000 Chr.22: 31768603-31768603 on Build GRCh38
rs778986487 Chr.22: 31768823-31768823 on Build GRCh38
rs761285561 Chr.22: 31768871-31768871 on Build GRCh38
rs773427300 Chr.22: 31768868-31768868 on Build GRCh38
rs1005912191 Chr.22: 31768684-31768684 on Build GRCh38
rs774061172 Chr.22: 31768882-31768884 on Build GRCh38
rs991583453 Chr.22: 31768906-31768906 on Build GRCh38
rs199587061 Chr.22: 31768884-31768884 on Build GRCh38
rs745359708 Chr.22: 31768844-31768844 on Build GRCh38
rs752492298 Chr.22: 31768800-31768800 on Build GRCh38
rs9606877 Chr.22: 31768835-31768835 on Build GRCh38
rs200399584 Chr.22: 31768901-31768901 on Build GRCh38
rs996942832 Chr.22: 31768490-31768490 on Build GRCh38
rs775219058 Chr.22: 31768852-31768852 on Build GRCh38
rs759543820 Chr.22: 31768880-31768880 on Build GRCh38
rs1008310202 Chr.22: 31768826-31768826 on Build GRCh38
rs201677987 Chr.22: 31768846-31768846 on Build GRCh38
rs762300886 Chr.22: 31768860-31768860 on Build GRCh38
rs981614448 Chr.22: 31768723-31768723 on Build GRCh38
rs777477751 Chr.22: 31768480-31768480 on Build GRCh38
rs368531839 Chr.22: 31768766-31768766 on Build GRCh38
rs766116050 Chr.22: 31768763-31768763 on Build GRCh38
rs951776586 Chr.22: 31768612-31768612 on Build GRCh38
rs750931089 Chr.22: 31768516-31768516 on Build GRCh38
rs186787523 Chr.22: 31768890-31768890 on Build GRCh38
rs200039638 Chr.22: 31768799-31768799 on Build GRCh38
rs755250431 Chr.22: 31768786-31768786 on Build GRCh38
rs938612254 Chr.22: 31768582-31768582 on Build GRCh38
rs749671626 Chr.22: 31768782-31768782 on Build GRCh38
rs776784907 Chr.22: 31768874-31768874 on Build GRCh38
rs934920632 Chr.22: 31768742-31768742 on Build GRCh38
rs1017884255 Chr.22: 31768827-31768827 on Build GRCh38
rs745841156 Chr.22: 31768770-31768773 on Build GRCh38
rs1057519230 Chr.22: 31768870-31768870 on Build GRCh38
rs1030463430 Chr.22: 31768747-31768750 on Build GRCh38
rs770738326 Chr.22: 31768816-31768816 on Build GRCh38
rs961991833 Chr.22: 31768550-31768550 on Build GRCh38
rs749024976 Chr.22: 31768781-31768782 on Build GRCh38
rs1055077675 Chr.22: 31768761-31768761 on Build GRCh38
rs775287852 Chr.22: 31768775-31768776 on Build GRCh38
rs954936116 Chr.22: 31768759-31768759 on Build GRCh38
rs897618159 Chr.22: 31768726-31768726 on Build GRCh38
rs971942668 Chr.22: 31768597-31768597 on Build GRCh38
rs761516610 Chr.22: 31768908-31768908 on Build GRCh38
rs897624790 Chr.22: 31768879-31768879 on Build GRCh38
rs558751608 Chr.22: 31768608-31768608 on Build GRCh38
rs34212007 Chr.22: 31768623-31768623 on Build GRCh38
rs766781814 Chr.22: 31768873-31768873 on Build GRCh38
rs543364711 Chr.22: 31768882-31768882 on Build GRCh38
rs893150813 Chr.22: 31768762-31768762 on Build GRCh38
rs150837731 Chr.22: 31768527-31768527 on Build GRCh38
rs370881336 Chr.22: 31768830-31768830 on Build GRCh38
rs768302447 Chr.22: 31768790-31768791 on Build GRCh38
rs915672737 Chr.22: 31768654-31768654 on Build GRCh38
rs949784247 Chr.22: 31768677-31768677 on Build GRCh38

View Full Product Details