Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32359827-32360337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFPL3
Entrez Gene ID
10738
Gene Name
ret finger protein like 3
Gene Aliases
-
UniGene
Hs.558455
Gene Chromosome Location
Chr.22: 32354885-32361161 on Build GRCh38
Transcripts
NM_001098535.1 NM_006604.2 NR_001450.3 NR_002596.2 NR_104232.1

Gene Symbol
RFPL3S
Entrez Gene ID
10737
Gene Name
RFPL3 antisense
Gene Aliases
NCRNA00005, RFPL3-AS, RFPL3-AS1, RFPL3ANT, RFPL3AS
UniGene
Hs.274285
Gene Chromosome Location
Chr.22: 32359906-32371264 on Build GRCh38
Transcripts
NM_001098535.1 NM_006604.2 NR_001450.3 NR_002596.2 NR_104232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3708223 c.412C>T p.L138F Chr.22: 32360290-32360290 on Build GRCh38
1033607 c.356G>A p.R119Q Chr.22: 32360321-32360321 on Build GRCh38
3553736 c.324C>T p.F108F Chr.22: 32360289-32360289 on Build GRCh38
1684966 c.364C>T p.L122F Chr.22: 32360329-32360329 on Build GRCh38
726542 c.307G>T p.D103Y Chr.22: 32360272-32360272 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201696385 Chr.22: 32360247-32360247 on Build GRCh38
rs564534728 Chr.22: 32359907-32359907 on Build GRCh38
rs780270312 Chr.22: 32360307-32360307 on Build GRCh38
rs1002203903 Chr.22: 32360113-32360113 on Build GRCh38
rs766159082 Chr.22: 32360257-32360257 on Build GRCh38
rs758420446 Chr.22: 32359917-32359919 on Build GRCh38
rs148702469 Chr.22: 32360054-32360054 on Build GRCh38
rs538388667 Chr.22: 32360148-32360148 on Build GRCh38
rs56317784 Chr.22: 32360114-32360114 on Build GRCh38
rs879192342 Chr.22: 32360025-32360025 on Build GRCh38
rs750550231 Chr.22: 32360291-32360291 on Build GRCh38
rs570264949 Chr.22: 32360065-32360065 on Build GRCh38
rs566034001 Chr.22: 32360180-32360180 on Build GRCh38
rs776353522 Chr.22: 32360262-32360262 on Build GRCh38
rs971901662 Chr.22: 32360194-32360194 on Build GRCh38
rs767866339 Chr.22: 32360279-32360279 on Build GRCh38
rs756122944 Chr.22: 32360304-32360304 on Build GRCh38
rs952811155 Chr.22: 32360122-32360122 on Build GRCh38
rs746127749 Chr.22: 32360208-32360208 on Build GRCh38
rs200359803 Chr.22: 32360305-32360305 on Build GRCh38
rs778737395 Chr.22: 32360315-32360315 on Build GRCh38
rs9619258 Chr.22: 32360254-32360254 on Build GRCh38
rs892656383 Chr.22: 32359998-32359998 on Build GRCh38
rs560837699 Chr.22: 32359956-32359956 on Build GRCh38
rs181339206 Chr.22: 32360317-32360317 on Build GRCh38
rs1016613746 Chr.22: 32360160-32360160 on Build GRCh38
rs369448389 Chr.22: 32360211-32360211 on Build GRCh38
rs201533287 Chr.22: 32360271-32360271 on Build GRCh38
rs146565766 Chr.22: 32360272-32360272 on Build GRCh38
rs752932215 Chr.22: 32360243-32360243 on Build GRCh38
rs764536235 Chr.22: 32360264-32360264 on Build GRCh38
rs375349779 Chr.22: 32360202-32360202 on Build GRCh38
rs774331056 Chr.22: 32360235-32360235 on Build GRCh38
rs754027922 Chr.22: 32360017-32360017 on Build GRCh38
rs1054431236 Chr.22: 32359997-32359997 on Build GRCh38
rs778012167 Chr.22: 32359975-32359975 on Build GRCh38
rs527292492 Chr.22: 32359910-32359910 on Build GRCh38
rs888778803 Chr.22: 32359965-32359966 on Build GRCh38
rs531087281 Chr.22: 32359959-32359959 on Build GRCh38
rs145079502 Chr.22: 32359930-32359930 on Build GRCh38
rs754834278 Chr.22: 32360313-32360313 on Build GRCh38
rs201369932 Chr.22: 32360218-32360218 on Build GRCh38
rs971867557 Chr.22: 32359867-32359867 on Build GRCh38
rs191299198 Chr.22: 32359897-32359897 on Build GRCh38
rs748112640 Chr.22: 32360236-32360236 on Build GRCh38
rs933025297 Chr.22: 32359940-32359940 on Build GRCh38
rs149085402 Chr.22: 32360083-32360083 on Build GRCh38
rs752050399 Chr.22: 32360294-32360294 on Build GRCh38
rs780211630 Chr.22: 32360219-32360219 on Build GRCh38
rs906634798 Chr.22: 32360062-32360062 on Build GRCh38
rs530290122 Chr.22: 32359961-32359961 on Build GRCh38
rs146767842 Chr.22: 32360316-32360316 on Build GRCh38
rs749087259 Chr.22: 32360222-32360222 on Build GRCh38
rs373266612 Chr.22: 32360224-32360224 on Build GRCh38
rs961959506 Chr.22: 32360188-32360188 on Build GRCh38
rs552952614 Chr.22: 32360162-32360162 on Build GRCh38
rs759787770 Chr.22: 32360090-32360090 on Build GRCh38
rs936315046 Chr.22: 32360289-32360289 on Build GRCh38

View Full Product Details