Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32541392-32541792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYN3
Entrez Gene ID
8224
Gene Name
synapsin III
Gene Aliases
-
UniGene
Hs.608750
Gene Chromosome Location
Chr.22: 32508022-33058391 on Build GRCh38
Transcripts
NM_001135774.1 NM_003490.3 NM_133633.2 XM_011530405.2 XM_011530406.2 XM_011530407.2 XM_011530408.1 XM_011530410.2 XM_017028960.1 XM_017028961.1 XM_017028962.1 XM_017028963.1 XM_017028964.1 XM_017028965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3972938 c.823G>A p.A275T Chr.22: 32541665-32541665 on Build GRCh38
6485205 c.822C>T p.V274V Chr.22: 32541666-32541666 on Build GRCh38
1600116 c.826A>T p.M276L Chr.22: 32541662-32541662 on Build GRCh38
6855943 c.844A>G p.T282A Chr.22: 32541644-32541644 on Build GRCh38
1228121 c.841G>A p.A281T Chr.22: 32541647-32541647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142547526 Chr.22: 32541444-32541444 on Build GRCh38
rs34165870 Chr.22: 32541651-32541651 on Build GRCh38
rs1022255187 Chr.22: 32541441-32541441 on Build GRCh38
rs577444070 Chr.22: 32541485-32541485 on Build GRCh38
rs761387900 Chr.22: 32541643-32541643 on Build GRCh38
rs996106078 Chr.22: 32541422-32541422 on Build GRCh38
rs135479 Chr.22: 32541754-32541754 on Build GRCh38
rs34270213 Chr.22: 32541661-32541661 on Build GRCh38
rs928016031 Chr.22: 32541456-32541456 on Build GRCh38
rs775238041 Chr.22: 32541638-32541638 on Build GRCh38
rs150712448 Chr.22: 32541693-32541693 on Build GRCh38
rs368619954 Chr.22: 32541556-32541556 on Build GRCh38
rs1031636656 Chr.22: 32541673-32541673 on Build GRCh38
rs765645809 Chr.22: 32541579-32541579 on Build GRCh38
rs769827986 Chr.22: 32541659-32541659 on Build GRCh38
rs771757179 Chr.22: 32541743-32541743 on Build GRCh38
rs761425481 Chr.22: 32541540-32541540 on Build GRCh38
rs760380900 Chr.22: 32541649-32541649 on Build GRCh38
rs200361200 Chr.22: 32541647-32541647 on Build GRCh38
rs772816610 Chr.22: 32541747-32541747 on Build GRCh38
rs767209253 Chr.22: 32541546-32541546 on Build GRCh38
rs570661835 Chr.22: 32541433-32541433 on Build GRCh38
rs549391257 Chr.22: 32541669-32541669 on Build GRCh38
rs762929046 Chr.22: 32541524-32541524 on Build GRCh38
rs141940631 Chr.22: 32541734-32541735 on Build GRCh38
rs2213407 Chr.22: 32541415-32541415 on Build GRCh38
rs756321358 Chr.22: 32541697-32541697 on Build GRCh38
rs780407268 Chr.22: 32541538-32541539 on Build GRCh38
rs886114363 Chr.22: 32541569-32541569 on Build GRCh38
rs574847032 Chr.22: 32541426-32541426 on Build GRCh38
rs200854450 Chr.22: 32541615-32541615 on Build GRCh38
rs964905468 Chr.22: 32541421-32541421 on Build GRCh38
rs758941248 Chr.22: 32541607-32541607 on Build GRCh38
rs944432651 Chr.22: 32541580-32541580 on Build GRCh38
rs564978453 Chr.22: 32541498-32541498 on Build GRCh38
rs777775255 Chr.22: 32541608-32541608 on Build GRCh38
rs781427098 Chr.22: 32541626-32541626 on Build GRCh38
rs894016102 Chr.22: 32541495-32541495 on Build GRCh38
rs779877666 Chr.22: 32541700-32541700 on Build GRCh38
rs141167959 Chr.22: 32541665-32541665 on Build GRCh38
rs548269854 Chr.22: 32541752-32541752 on Build GRCh38
rs186550097 Chr.22: 32541457-32541457 on Build GRCh38
rs547334487 Chr.22: 32541519-32541519 on Build GRCh38
rs866467836 Chr.22: 32541660-32541660 on Build GRCh38
rs769628418 Chr.22: 32541629-32541629 on Build GRCh38
rs763549894 Chr.22: 32541526-32541526 on Build GRCh38
rs774093694 Chr.22: 32541642-32541642 on Build GRCh38
rs746046047 Chr.22: 32541627-32541627 on Build GRCh38
rs765735742 Chr.22: 32541652-32541652 on Build GRCh38
rs747984992 Chr.22: 32541742-32541742 on Build GRCh38
rs778939750 Chr.22: 32541738-32541738 on Build GRCh38
rs763169451 Chr.22: 32541662-32541662 on Build GRCh38
rs757382879 Chr.22: 32541671-32541671 on Build GRCh38
rs527482833 Chr.22: 32541516-32541516 on Build GRCh38
rs768266861 Chr.22: 32541724-32541724 on Build GRCh38
rs148217218 Chr.22: 32541648-32541648 on Build GRCh38
rs980307501 Chr.22: 32541512-32541512 on Build GRCh38
rs545172818 Chr.22: 32541487-32541487 on Build GRCh38
rs1009078548 Chr.22: 32541417-32541417 on Build GRCh38
rs956114081 Chr.22: 32541466-32541466 on Build GRCh38
rs749248197 Chr.22: 32541639-32541639 on Build GRCh38
rs773905738 Chr.22: 32541533-32541533 on Build GRCh38
rs750029312 Chr.22: 32541553-32541553 on Build GRCh38
rs757217891 Chr.22: 32541621-32541621 on Build GRCh38
rs760462715 Chr.22: 32541751-32541751 on Build GRCh38
rs200163576 Chr.22: 32541753-32541753 on Build GRCh38
rs371435439 Chr.22: 32541589-32541589 on Build GRCh38
rs749119465 Chr.22: 32541722-32541722 on Build GRCh38
rs189882761 Chr.22: 32541672-32541672 on Build GRCh38
rs565468246 Chr.22: 32541504-32541504 on Build GRCh38
rs970235213 Chr.22: 32541749-32541749 on Build GRCh38
rs111594551 Chr.22: 32541467-32541467 on Build GRCh38

View Full Product Details