Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37372613-37373120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653275 c.2457G>T p.K819N Chr.22: 37373078-37373078 on Build GRCh38
3842629 c.2434A>G p.K812E Chr.22: 37373101-37373101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs901393018 Chr.22: 37372768-37372768 on Build GRCh38
rs1013157862 Chr.22: 37372695-37372695 on Build GRCh38
rs942716865 Chr.22: 37372928-37372935 on Build GRCh38
rs373874492 Chr.22: 37373046-37373046 on Build GRCh38
rs1037321628 Chr.22: 37373070-37373070 on Build GRCh38
rs890219309 Chr.22: 37372939-37372939 on Build GRCh38
rs1005690969 Chr.22: 37372712-37372712 on Build GRCh38
rs990775114 Chr.22: 37372659-37372659 on Build GRCh38
rs1005826697 Chr.22: 37372888-37372888 on Build GRCh38
rs993239384 Chr.22: 37372932-37372932 on Build GRCh38
rs1031455885 Chr.22: 37372995-37372995 on Build GRCh38
rs1011887391 Chr.22: 37372871-37372871 on Build GRCh38
rs952571889 Chr.22: 37372919-37372920 on Build GRCh38
rs868020674 Chr.22: 37372857-37372857 on Build GRCh38
rs746122481 Chr.22: 37372827-37372827 on Build GRCh38
rs917290035 Chr.22: 37373009-37373009 on Build GRCh38
rs542697822 Chr.22: 37372754-37372754 on Build GRCh38
rs561334919 Chr.22: 37372761-37372761 on Build GRCh38
rs968586436 Chr.22: 37372958-37372958 on Build GRCh38
rs959030442 Chr.22: 37372859-37372859 on Build GRCh38
rs970596764 Chr.22: 37372682-37372682 on Build GRCh38
rs780122345 Chr.22: 37372982-37372982 on Build GRCh38
rs978541098 Chr.22: 37372793-37372793 on Build GRCh38
rs1040432009 Chr.22: 37372733-37372733 on Build GRCh38
rs879907711 Chr.22: 37372994-37372994 on Build GRCh38
rs185611479 Chr.22: 37372885-37372885 on Build GRCh38
rs934730603 Chr.22: 37372854-37372854 on Build GRCh38
rs745841330 Chr.22: 37372976-37372977 on Build GRCh38
rs948692040 Chr.22: 37372876-37372876 on Build GRCh38
rs983890678 Chr.22: 37373054-37373054 on Build GRCh38
rs760203893 Chr.22: 37373085-37373085 on Build GRCh38
rs550103003 Chr.22: 37373063-37373064 on Build GRCh38
rs1044411108 Chr.22: 37372884-37372884 on Build GRCh38
rs768670879 Chr.22: 37373064-37373064 on Build GRCh38
rs373169281 Chr.22: 37372686-37372686 on Build GRCh38
rs985816007 Chr.22: 37372771-37372771 on Build GRCh38
rs529749515 Chr.22: 37372822-37372822 on Build GRCh38
rs904677766 Chr.22: 37372784-37372784 on Build GRCh38
rs753211802 Chr.22: 37373096-37373096 on Build GRCh38
rs377321643 Chr.22: 37372758-37372758 on Build GRCh38
rs994529397 Chr.22: 37373031-37373032 on Build GRCh38
rs910426390 Chr.22: 37372772-37372772 on Build GRCh38
rs144435849 Chr.22: 37373004-37373004 on Build GRCh38
rs565340904 Chr.22: 37373003-37373003 on Build GRCh38
rs372219343 Chr.22: 37373066-37373066 on Build GRCh38
rs778062441 Chr.22: 37373027-37373027 on Build GRCh38
rs1041967438 Chr.22: 37372935-37372935 on Build GRCh38
rs781442509 Chr.22: 37373057-37373057 on Build GRCh38
rs917257303 Chr.22: 37372872-37372872 on Build GRCh38
rs972659349 Chr.22: 37372897-37372897 on Build GRCh38
rs34957013 Chr.22: 37372718-37372718 on Build GRCh38
rs924669559 Chr.22: 37372641-37372641 on Build GRCh38
rs375521751 Chr.22: 37373084-37373084 on Build GRCh38
rs28706310 Chr.22: 37372692-37372692 on Build GRCh38
rs1017288249 Chr.22: 37372957-37372957 on Build GRCh38
rs897357111 Chr.22: 37372931-37372931 on Build GRCh38
rs1025517734 Chr.22: 37373037-37373037 on Build GRCh38
rs569458468 Chr.22: 37373069-37373069 on Build GRCh38
rs368464304 Chr.22: 37373065-37373065 on Build GRCh38
rs1029985473 Chr.22: 37372760-37372760 on Build GRCh38
rs895831737 Chr.22: 37372694-37372694 on Build GRCh38
rs936116199 Chr.22: 37372943-37372943 on Build GRCh38
rs897326378 Chr.22: 37373071-37373071 on Build GRCh38
rs61737924 Chr.22: 37373089-37373089 on Build GRCh38
rs911271775 Chr.22: 37372927-37372928 on Build GRCh38
rs1045084245 Chr.22: 37372698-37372698 on Build GRCh38
rs528609285 Chr.22: 37372816-37372816 on Build GRCh38
rs200269334 Chr.22: 37373090-37373090 on Build GRCh38
rs888709704 Chr.22: 37372704-37372704 on Build GRCh38
rs1015785755 Chr.22: 37372727-37372727 on Build GRCh38
rs1037185980 Chr.22: 37372889-37372889 on Build GRCh38
rs1025931885 Chr.22: 37372906-37372906 on Build GRCh38
rs961768712 Chr.22: 37372732-37372732 on Build GRCh38
rs547521310 Chr.22: 37372717-37372717 on Build GRCh38
rs1053542856 Chr.22: 37372978-37372978 on Build GRCh38
rs369093489 Chr.22: 37373038-37373038 on Build GRCh38
rs182751132 Chr.22: 37372678-37372678 on Build GRCh38
rs759034259 Chr.22: 37372936-37372936 on Build GRCh38
rs1007608179 Chr.22: 37372945-37372945 on Build GRCh38
rs398037078 Chr.22: 37372723-37372723 on Build GRCh38
rs968676440 Chr.22: 37372773-37372773 on Build GRCh38

View Full Product Details