Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41459626-41460125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHF5A
Entrez Gene ID
84844
Gene Name
PHD finger protein 5A
Gene Aliases
INI, Rds3, SAP14b, SF3B7, SF3b14b, bK223H9.2
UniGene
Hs.474980
Gene Chromosome Location
Chr.22: 41459717-41468704 on Build GRCh38
Transcripts
NM_032758.3

SNP Detail

SNP ID Chromosome Location
rs918695905 Chr.22: 41459676-41459676 on Build GRCh38
rs769031354 Chr.22: 41459821-41459821 on Build GRCh38
rs780059894 Chr.22: 41459919-41459919 on Build GRCh38
rs894936274 Chr.22: 41460050-41460050 on Build GRCh38
rs982556503 Chr.22: 41460010-41460010 on Build GRCh38
rs902963427 Chr.22: 41459837-41459837 on Build GRCh38
rs1042972 Chr.22: 41459945-41459945 on Build GRCh38
rs891781644 Chr.22: 41459996-41459996 on Build GRCh38
rs751852522 Chr.22: 41459900-41459901 on Build GRCh38
rs951420876 Chr.22: 41460033-41460033 on Build GRCh38
rs5758364 Chr.22: 41459908-41459908 on Build GRCh38
rs975531805 Chr.22: 41460058-41460058 on Build GRCh38
rs569192944 Chr.22: 41460055-41460055 on Build GRCh38
rs942839814 Chr.22: 41459702-41459702 on Build GRCh38
rs35777353 Chr.22: 41459895-41459896 on Build GRCh38
rs186918522 Chr.22: 41460036-41460036 on Build GRCh38
rs1033504926 Chr.22: 41460049-41460049 on Build GRCh38
rs559991943 Chr.22: 41459892-41459892 on Build GRCh38
rs906408572 Chr.22: 41460030-41460030 on Build GRCh38
rs765030725 Chr.22: 41460095-41460095 on Build GRCh38
rs192793191 Chr.22: 41460005-41460005 on Build GRCh38
rs13766 Chr.22: 41460059-41460059 on Build GRCh38
rs1027319200 Chr.22: 41459746-41459746 on Build GRCh38
rs574734405 Chr.22: 41459688-41459693 on Build GRCh38
rs865871235 Chr.22: 41459918-41459918 on Build GRCh38
rs76744137 Chr.22: 41459926-41459926 on Build GRCh38
rs922055533 Chr.22: 41459979-41459979 on Build GRCh38
rs557479641 Chr.22: 41460047-41460048 on Build GRCh38
rs998280365 Chr.22: 41459802-41459802 on Build GRCh38
rs1041368420 Chr.22: 41459809-41459811 on Build GRCh38
rs973099143 Chr.22: 41459727-41459727 on Build GRCh38
rs974848757 Chr.22: 41459962-41459962 on Build GRCh38
rs1033431298 Chr.22: 41459678-41459678 on Build GRCh38
rs774216439 Chr.22: 41459813-41459813 on Build GRCh38
rs17002469 Chr.22: 41460037-41460037 on Build GRCh38
rs1042993 Chr.22: 41459803-41459803 on Build GRCh38
rs9423 Chr.22: 41459919-41459919 on Build GRCh38
rs936086315 Chr.22: 41459984-41459984 on Build GRCh38
rs183695146 Chr.22: 41460034-41460034 on Build GRCh38
rs57259696 Chr.22: 41460048-41460049 on Build GRCh38
rs944893073 Chr.22: 41459942-41459942 on Build GRCh38
rs999945259 Chr.22: 41459880-41459880 on Build GRCh38
rs946009443 Chr.22: 41459997-41459997 on Build GRCh38
rs1025643536 Chr.22: 41460027-41460027 on Build GRCh38
rs1054889689 Chr.22: 41459985-41459985 on Build GRCh38
rs1027761031 Chr.22: 41460071-41460071 on Build GRCh38
rs958535984 Chr.22: 41459913-41459913 on Build GRCh38
rs761187666 Chr.22: 41459706-41459706 on Build GRCh38
rs113800666 Chr.22: 41459847-41459847 on Build GRCh38
rs1048585645 Chr.22: 41459681-41459681 on Build GRCh38
rs535977723 Chr.22: 41460008-41460008 on Build GRCh38
rs951055697 Chr.22: 41459764-41459764 on Build GRCh38

View Full Product Details