Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41507657-41508167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACO2
Entrez Gene ID
50
Gene Name
aconitase 2
Gene Aliases
ACONM, HEL-S-284, ICRD, OCA8, OPA9
UniGene
Hs.643610
Gene Chromosome Location
Chr.22: 41469125-41528989 on Build GRCh38
Transcripts
NM_001098.2 XM_017028812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1034633 c.284G>A p.R95H Chr.22: 41507901-41507901 on Build GRCh38
4651460 c.233C>T p.A78V Chr.22: 41507850-41507850 on Build GRCh38
1034634 c.339C>T p.S113S Chr.22: 41507956-41507956 on Build GRCh38
1682295 c.330C>A p.F110L Chr.22: 41507947-41507947 on Build GRCh38
1181608 c.317C>T p.A106V Chr.22: 41507934-41507934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764337627 Chr.22: 41507902-41507902 on Build GRCh38
rs373973502 Chr.22: 41507886-41507886 on Build GRCh38
rs763171140 Chr.22: 41507959-41507959 on Build GRCh38
rs375439516 Chr.22: 41508063-41508063 on Build GRCh38
rs11705367 Chr.22: 41508057-41508057 on Build GRCh38
rs779868128 Chr.22: 41508041-41508041 on Build GRCh38
rs186922643 Chr.22: 41507854-41507854 on Build GRCh38
rs387907389 Chr.22: 41507867-41507867 on Build GRCh38
rs150593227 Chr.22: 41507848-41507848 on Build GRCh38
rs573877104 Chr.22: 41508081-41508081 on Build GRCh38
rs948581542 Chr.22: 41507778-41507778 on Build GRCh38
rs776871654 Chr.22: 41508065-41508065 on Build GRCh38
rs763769484 Chr.22: 41508086-41508086 on Build GRCh38
rs990154816 Chr.22: 41507698-41507698 on Build GRCh38
rs763516614 Chr.22: 41508072-41508072 on Build GRCh38
rs773229850 Chr.22: 41507798-41507798 on Build GRCh38
rs576127948 Chr.22: 41508022-41508022 on Build GRCh38
rs745579534 Chr.22: 41507891-41507891 on Build GRCh38
rs777420895 Chr.22: 41507877-41507877 on Build GRCh38
rs891484856 Chr.22: 41507844-41507844 on Build GRCh38
rs915177221 Chr.22: 41507712-41507712 on Build GRCh38
rs141772938 Chr.22: 41507837-41507837 on Build GRCh38
rs1030205453 Chr.22: 41507687-41507687 on Build GRCh38
rs753985150 Chr.22: 41507835-41507835 on Build GRCh38
rs184501012 Chr.22: 41507729-41507729 on Build GRCh38
rs748850283 Chr.22: 41507878-41507878 on Build GRCh38
rs137831 Chr.22: 41507809-41507809 on Build GRCh38
rs863223879 Chr.22: 41508042-41508042 on Build GRCh38
rs11554469 Chr.22: 41507908-41507908 on Build GRCh38
rs142651093 Chr.22: 41507899-41507899 on Build GRCh38
rs377384540 Chr.22: 41507944-41507944 on Build GRCh38
rs939137020 Chr.22: 41507745-41507745 on Build GRCh38
rs370417583 Chr.22: 41508039-41508039 on Build GRCh38
rs766837794 Chr.22: 41508073-41508073 on Build GRCh38
rs565805751 Chr.22: 41507872-41507872 on Build GRCh38
rs769434569 Chr.22: 41507783-41507783 on Build GRCh38
rs939190705 Chr.22: 41508054-41508054 on Build GRCh38
rs1020287670 Chr.22: 41508023-41508023 on Build GRCh38
rs543394003 Chr.22: 41508025-41508025 on Build GRCh38
rs200362666 Chr.22: 41508068-41508068 on Build GRCh38
rs760882521 Chr.22: 41507901-41507901 on Build GRCh38
rs887178970 Chr.22: 41507827-41507827 on Build GRCh38
rs775780883 Chr.22: 41507807-41507807 on Build GRCh38
rs771060439 Chr.22: 41507799-41507799 on Build GRCh38
rs755799401 Chr.22: 41508018-41508018 on Build GRCh38
rs886729439 Chr.22: 41507815-41507815 on Build GRCh38
rs754958998 Chr.22: 41507760-41507760 on Build GRCh38
rs531571885 Chr.22: 41507751-41507751 on Build GRCh38
rs765694409 Chr.22: 41507950-41507950 on Build GRCh38
rs138373868 Chr.22: 41507956-41507956 on Build GRCh38
rs753549443 Chr.22: 41508095-41508095 on Build GRCh38
rs376995122 Chr.22: 41508070-41508070 on Build GRCh38
rs773989143 Chr.22: 41507800-41507800 on Build GRCh38
rs764145213 Chr.22: 41507830-41507830 on Build GRCh38
rs746854004 Chr.22: 41508049-41508049 on Build GRCh38
rs776668867 Chr.22: 41507923-41507923 on Build GRCh38
rs376128073 Chr.22: 41508096-41508096 on Build GRCh38
rs759393809 Chr.22: 41507802-41507802 on Build GRCh38
rs200692540 Chr.22: 41507862-41507862 on Build GRCh38
rs955574428 Chr.22: 41507704-41507704 on Build GRCh38
rs747571957 Chr.22: 41507773-41507773 on Build GRCh38
rs2076199 Chr.22: 41507739-41507739 on Build GRCh38
rs552187968 Chr.22: 41507737-41507737 on Build GRCh38
rs767592074 Chr.22: 41507803-41507803 on Build GRCh38
rs199671283 Chr.22: 41508093-41508093 on Build GRCh38
rs539524933 Chr.22: 41507885-41507885 on Build GRCh38
rs778653392 Chr.22: 41508026-41508026 on Build GRCh38
rs766757344 Chr.22: 41507964-41507964 on Build GRCh38
rs750705704 Chr.22: 41507849-41507849 on Build GRCh38
rs751939728 Chr.22: 41507984-41507984 on Build GRCh38
rs757819166 Chr.22: 41507747-41507747 on Build GRCh38
rs144204883 Chr.22: 41507861-41507861 on Build GRCh38
rs778480089 Chr.22: 41508097-41508097 on Build GRCh38
rs751953368 Chr.22: 41508075-41508075 on Build GRCh38
rs1048044477 Chr.22: 41507818-41507818 on Build GRCh38
rs147763800 Chr.22: 41507892-41507892 on Build GRCh38
rs745801074 Chr.22: 41508038-41508038 on Build GRCh38
rs915599554 Chr.22: 41507763-41507763 on Build GRCh38
rs978373607 Chr.22: 41507736-41507736 on Build GRCh38
rs786200924 Chr.22: 41507953-41507953 on Build GRCh38
rs374163200 Chr.22: 41507748-41507748 on Build GRCh38
rs749213072 Chr.22: 41508089-41508090 on Build GRCh38
rs115181917 Chr.22: 41508109-41508109 on Build GRCh38
rs758823518 Chr.22: 41507853-41507853 on Build GRCh38

View Full Product Details