Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41730440-41730941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEI1
Entrez Gene ID
150365
Gene Name
meiotic double-stranded break formation protein 1
Gene Aliases
SPATA38
UniGene
Hs.116419
Gene Chromosome Location
Chr.22: 41699503-41799456 on Build GRCh38
Transcripts
NM_152513.3 XM_011529935.1 XM_011529936.1 XM_011529937.1 XM_011529938.1 XM_011529939.1 XM_011529940.1 XM_011529941.1 XM_011529942.2 XM_011529943.1 XM_011529944.1 XM_011529945.2 XM_011529946.1 XM_011529947.1 XM_011529948.2 XM_011529949.1 XM_011529950.1 XM_011529952.1 XM_011529955.1 XM_011529956.2 XM_017028633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6301860 c.1011T>C p.S337S Chr.22: 41730552-41730552 on Build GRCh38
3153640 c.1096G>T p.G366W Chr.22: 41730637-41730637 on Build GRCh38
6441116 c.1004C>T p.S335F Chr.22: 41730545-41730545 on Build GRCh38
4753666 c.307A>G p.T103A Chr.22: 41730628-41730628 on Build GRCh38
6301861 c.231T>C p.S77S Chr.22: 41730552-41730552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771653488 Chr.22: 41730676-41730676 on Build GRCh38
rs958737039 Chr.22: 41730705-41730705 on Build GRCh38
rs368049574 Chr.22: 41730645-41730645 on Build GRCh38
rs538590188 Chr.22: 41730701-41730701 on Build GRCh38
rs778047977 Chr.22: 41730588-41730588 on Build GRCh38
rs12171085 Chr.22: 41730679-41730679 on Build GRCh38
rs1003292336 Chr.22: 41730834-41730834 on Build GRCh38
rs758929622 Chr.22: 41730475-41730475 on Build GRCh38
rs745778270 Chr.22: 41730548-41730548 on Build GRCh38
rs771776312 Chr.22: 41730589-41730589 on Build GRCh38
rs41528750 Chr.22: 41730815-41730815 on Build GRCh38
rs760598720 Chr.22: 41730489-41730489 on Build GRCh38
rs567432422 Chr.22: 41730516-41730516 on Build GRCh38
rs970512681 Chr.22: 41730684-41730684 on Build GRCh38
rs1036668804 Chr.22: 41730707-41730707 on Build GRCh38
rs73161379 Chr.22: 41730704-41730704 on Build GRCh38
rs908726897 Chr.22: 41730767-41730767 on Build GRCh38
rs755865001 Chr.22: 41730561-41730561 on Build GRCh38
rs767031327 Chr.22: 41730477-41730477 on Build GRCh38
rs991937836 Chr.22: 41730716-41730716 on Build GRCh38
rs371001211 Chr.22: 41730506-41730506 on Build GRCh38
rs919619641 Chr.22: 41730911-41730913 on Build GRCh38
rs774913078 Chr.22: 41730606-41730606 on Build GRCh38
rs367838848 Chr.22: 41730571-41730571 on Build GRCh38
rs779701477 Chr.22: 41730677-41730677 on Build GRCh38
rs533464959 Chr.22: 41730853-41730853 on Build GRCh38
rs200162893 Chr.22: 41730669-41730669 on Build GRCh38
rs554734752 Chr.22: 41730870-41730870 on Build GRCh38
rs755133035 Chr.22: 41730651-41730651 on Build GRCh38
rs979689302 Chr.22: 41730847-41730847 on Build GRCh38
rs755128389 Chr.22: 41730474-41730474 on Build GRCh38
rs768414868 Chr.22: 41730848-41730848 on Build GRCh38
rs577862497 Chr.22: 41730687-41730687 on Build GRCh38
rs764022140 Chr.22: 41730507-41730507 on Build GRCh38
rs78712535 Chr.22: 41730817-41730817 on Build GRCh38
rs368356860 Chr.22: 41730680-41730680 on Build GRCh38
rs1003428060 Chr.22: 41730694-41730694 on Build GRCh38
rs374534859 Chr.22: 41730569-41730569 on Build GRCh38
rs944157528 Chr.22: 41730773-41730773 on Build GRCh38
rs982867963 Chr.22: 41730766-41730766 on Build GRCh38
rs746844132 Chr.22: 41730824-41730824 on Build GRCh38
rs767790261 Chr.22: 41730483-41730483 on Build GRCh38
rs549543913 Chr.22: 41730466-41730466 on Build GRCh38
rs574145432 Chr.22: 41730823-41730823 on Build GRCh38
rs780272809 Chr.22: 41730527-41730527 on Build GRCh38
rs756637560 Chr.22: 41730665-41730665 on Build GRCh38
rs906384401 Chr.22: 41730833-41730833 on Build GRCh38
rs1042093129 Chr.22: 41730825-41730825 on Build GRCh38
rs889664872 Chr.22: 41730742-41730742 on Build GRCh38
rs1019028450 Chr.22: 41730631-41730631 on Build GRCh38
rs770435537 Chr.22: 41730471-41730471 on Build GRCh38
rs917671950 Chr.22: 41730872-41730872 on Build GRCh38
rs982875721 Chr.22: 41730901-41730901 on Build GRCh38
rs952936776 Chr.22: 41730729-41730729 on Build GRCh38
rs770227541 Chr.22: 41730585-41730585 on Build GRCh38
rs944294815 Chr.22: 41730570-41730570 on Build GRCh38
rs199767157 Chr.22: 41730663-41730663 on Build GRCh38
rs574942651 Chr.22: 41730782-41730782 on Build GRCh38
rs182455386 Chr.22: 41730835-41730835 on Build GRCh38
rs751757895 Chr.22: 41730558-41730558 on Build GRCh38
rs926823848 Chr.22: 41730692-41730692 on Build GRCh38
rs376561902 Chr.22: 41730649-41730649 on Build GRCh38
rs745815591 Chr.22: 41730799-41730799 on Build GRCh38
rs559946950 Chr.22: 41730850-41730850 on Build GRCh38
rs921476057 Chr.22: 41730809-41730809 on Build GRCh38
rs370146597 Chr.22: 41730629-41730629 on Build GRCh38
rs1027423699 Chr.22: 41730868-41730868 on Build GRCh38
rs994750414 Chr.22: 41730863-41730863 on Build GRCh38
rs190646469 Chr.22: 41730648-41730648 on Build GRCh38
rs368271605 Chr.22: 41730650-41730650 on Build GRCh38
rs375353368 Chr.22: 41730515-41730515 on Build GRCh38
rs760187909 Chr.22: 41730616-41730616 on Build GRCh38
rs765063052 Chr.22: 41730643-41730643 on Build GRCh38
rs768529707 Chr.22: 41730622-41730622 on Build GRCh38
rs1049659079 Chr.22: 41730739-41730739 on Build GRCh38
rs748412324 Chr.22: 41730562-41730562 on Build GRCh38
rs371108479 Chr.22: 41730522-41730522 on Build GRCh38
rs755478052 Chr.22: 41730559-41730559 on Build GRCh38
rs781750811 Chr.22: 41730656-41730656 on Build GRCh38
rs753568812 Chr.22: 41730508-41730508 on Build GRCh38
rs185660661 Chr.22: 41730877-41730877 on Build GRCh38
rs959534079 Chr.22: 41730854-41730854 on Build GRCh38
rs781129196 Chr.22: 41730494-41730496 on Build GRCh38
rs972875074 Chr.22: 41730907-41730907 on Build GRCh38
rs775039635 Chr.22: 41730480-41730480 on Build GRCh38
rs1026977529 Chr.22: 41730723-41730723 on Build GRCh38
rs373604685 Chr.22: 41730641-41730641 on Build GRCh38
rs774074834 Chr.22: 41730472-41730472 on Build GRCh38
rs66636807 Chr.22: 41730712-41730713 on Build GRCh38

View Full Product Details