Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41800557-41801056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC134
Entrez Gene ID
79879
Gene Name
coiled-coil domain containing 134
Gene Aliases
-
UniGene
Hs.474991
Gene Chromosome Location
Chr.22: 41800610-41832164 on Build GRCh38
Transcripts
NM_001304797.1 NM_024821.3 XM_005261748.3

SNP Detail

SNP ID Chromosome Location
rs972226911 Chr.22: 41800917-41800917 on Build GRCh38
rs941478037 Chr.22: 41800651-41800651 on Build GRCh38
rs949837121 Chr.22: 41800727-41800727 on Build GRCh38
rs1036419651 Chr.22: 41800619-41800619 on Build GRCh38
rs1052247423 Chr.22: 41800946-41800946 on Build GRCh38
rs910493569 Chr.22: 41801010-41801010 on Build GRCh38
rs892403820 Chr.22: 41800728-41800728 on Build GRCh38
rs943276878 Chr.22: 41801036-41801036 on Build GRCh38
rs1013907165 Chr.22: 41800739-41800739 on Build GRCh38
rs897785358 Chr.22: 41800621-41800621 on Build GRCh38
rs368060219 Chr.22: 41800640-41800640 on Build GRCh38
rs1039820486 Chr.22: 41800709-41800709 on Build GRCh38
rs1020817455 Chr.22: 41800952-41800952 on Build GRCh38
rs1024708076 Chr.22: 41800611-41800611 on Build GRCh38
rs958930691 Chr.22: 41800875-41800875 on Build GRCh38
rs1000625746 Chr.22: 41800714-41800714 on Build GRCh38
rs58505525 Chr.22: 41800741-41800741 on Build GRCh38
rs139562 Chr.22: 41800680-41800680 on Build GRCh38
rs903212530 Chr.22: 41800858-41800858 on Build GRCh38
rs1020699603 Chr.22: 41800662-41800662 on Build GRCh38
rs116251441 Chr.22: 41800865-41800865 on Build GRCh38
rs980592810 Chr.22: 41800636-41800636 on Build GRCh38
rs962891680 Chr.22: 41800843-41800843 on Build GRCh38
rs925007276 Chr.22: 41800914-41800914 on Build GRCh38
rs1001635836 Chr.22: 41800811-41800811 on Build GRCh38
rs1049024561 Chr.22: 41800746-41800746 on Build GRCh38
rs943243816 Chr.22: 41800828-41800828 on Build GRCh38
rs912231340 Chr.22: 41800874-41800874 on Build GRCh38
rs1000740090 Chr.22: 41800578-41800578 on Build GRCh38
rs890779834 Chr.22: 41800684-41800684 on Build GRCh38
rs930538107 Chr.22: 41800623-41800623 on Build GRCh38
rs951704305 Chr.22: 41800936-41800936 on Build GRCh38
rs73163349 Chr.22: 41800839-41800839 on Build GRCh38
rs991711983 Chr.22: 41800892-41800892 on Build GRCh38
rs544847161 Chr.22: 41800674-41800674 on Build GRCh38
rs988802135 Chr.22: 41800595-41800595 on Build GRCh38
rs1052319350 Chr.22: 41800717-41800717 on Build GRCh38
rs998218748 Chr.22: 41800660-41800660 on Build GRCh38
rs747715350 Chr.22: 41801035-41801035 on Build GRCh38
rs971767175 Chr.22: 41800775-41800775 on Build GRCh38
rs987455994 Chr.22: 41800872-41800872 on Build GRCh38
rs1030841615 Chr.22: 41800582-41800582 on Build GRCh38
rs942502948 Chr.22: 41800686-41800686 on Build GRCh38
rs892291169 Chr.22: 41800966-41800966 on Build GRCh38
rs578096458 Chr.22: 41800657-41800657 on Build GRCh38
rs918819413 Chr.22: 41800672-41800672 on Build GRCh38
rs974262630 Chr.22: 41800849-41800849 on Build GRCh38
rs889006479 Chr.22: 41800780-41800780 on Build GRCh38
rs942326234 Chr.22: 41800895-41800895 on Build GRCh38
rs546629848 Chr.22: 41800985-41800985 on Build GRCh38
rs2076018 Chr.22: 41800907-41800907 on Build GRCh38
rs971532934 Chr.22: 41800626-41800626 on Build GRCh38
rs951644047 Chr.22: 41800871-41800871 on Build GRCh38
rs578050710 Chr.22: 41800655-41800655 on Build GRCh38
rs553008660 Chr.22: 41800652-41800652 on Build GRCh38
rs1040591305 Chr.22: 41800832-41800832 on Build GRCh38
rs1046712115 Chr.22: 41800999-41800999 on Build GRCh38
rs936404619 Chr.22: 41800929-41800929 on Build GRCh38
rs918984695 Chr.22: 41800921-41800921 on Build GRCh38
rs1013508449 Chr.22: 41800987-41800987 on Build GRCh38
rs978988787 Chr.22: 41800668-41800668 on Build GRCh38
rs927488499 Chr.22: 41800641-41800641 on Build GRCh38
rs903704312 Chr.22: 41800710-41800710 on Build GRCh38
rs971791324 Chr.22: 41800729-41800729 on Build GRCh38
rs541075215 Chr.22: 41800583-41800583 on Build GRCh38
rs749282520 Chr.22: 41800776-41800776 on Build GRCh38
rs1024886868 Chr.22: 41800904-41800904 on Build GRCh38

View Full Product Details