Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43973148-43973625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMM50
Entrez Gene ID
25813
Gene Name
SAMM50 sorting and assembly machinery component
Gene Aliases
CGI-51, OMP85, SAM50, TOB55, TRG-3, YNL026W
UniGene
Hs.505824
Gene Chromosome Location
Chr.22: 43955381-43996532 on Build GRCh38
Transcripts
NM_015380.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7193436 c.643T>C p.Y215H Chr.22: 43973318-43973318 on Build GRCh38
5790682 c.609A>T p.S203S Chr.22: 43973284-43973284 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772249410 Chr.22: 43973262-43973262 on Build GRCh38
rs781692606 Chr.22: 43973375-43973375 on Build GRCh38
rs943536317 Chr.22: 43973454-43973454 on Build GRCh38
rs1013974835 Chr.22: 43973350-43973350 on Build GRCh38
rs368022961 Chr.22: 43973545-43973545 on Build GRCh38
rs150292452 Chr.22: 43973288-43973288 on Build GRCh38
rs1020975570 Chr.22: 43973439-43973439 on Build GRCh38
rs777798922 Chr.22: 43973246-43973246 on Build GRCh38
rs550259267 Chr.22: 43973283-43973283 on Build GRCh38
rs538219063 Chr.22: 43973437-43973437 on Build GRCh38
rs374982740 Chr.22: 43973257-43973257 on Build GRCh38
rs117177740 Chr.22: 43973411-43973411 on Build GRCh38
rs888281746 Chr.22: 43973390-43973390 on Build GRCh38
rs766633209 Chr.22: 43973477-43973477 on Build GRCh38
rs77442627 Chr.22: 43973357-43973357 on Build GRCh38
rs987591779 Chr.22: 43973448-43973448 on Build GRCh38
rs537659442 Chr.22: 43973516-43973516 on Build GRCh38
rs772011624 Chr.22: 43973190-43973190 on Build GRCh38
rs987886830 Chr.22: 43973515-43973515 on Build GRCh38
rs753742813 Chr.22: 43973189-43973189 on Build GRCh38
rs768680870 Chr.22: 43973367-43973367 on Build GRCh38
rs766076381 Chr.22: 43973214-43973214 on Build GRCh38
rs149006642 Chr.22: 43973323-43973323 on Build GRCh38
rs975348574 Chr.22: 43973472-43973472 on Build GRCh38
rs1005830801 Chr.22: 43973410-43973410 on Build GRCh38
rs777074079 Chr.22: 43973281-43973281 on Build GRCh38
rs573589246 Chr.22: 43973367-43973368 on Build GRCh38
rs753798267 Chr.22: 43973302-43973302 on Build GRCh38
rs535199716 Chr.22: 43973568-43973568 on Build GRCh38
rs774389937 Chr.22: 43973185-43973185 on Build GRCh38
rs6006466 Chr.22: 43973549-43973549 on Build GRCh38
rs756968475 Chr.22: 43973198-43973201 on Build GRCh38
rs2235772 Chr.22: 43973449-43973449 on Build GRCh38
rs749122766 Chr.22: 43973248-43973248 on Build GRCh38
rs34677401 Chr.22: 43973296-43973296 on Build GRCh38
rs1029334111 Chr.22: 43973198-43973198 on Build GRCh38
rs761152620 Chr.22: 43973202-43973202 on Build GRCh38
rs764505285 Chr.22: 43973203-43973203 on Build GRCh38
rs758550338 Chr.22: 43973334-43973334 on Build GRCh38
rs779806276 Chr.22: 43973382-43973382 on Build GRCh38
rs747021327 Chr.22: 43973267-43973267 on Build GRCh38
rs757079940 Chr.22: 43973251-43973251 on Build GRCh38
rs746673447 Chr.22: 43973389-43973389 on Build GRCh38
rs180928342 Chr.22: 43973252-43973252 on Build GRCh38
rs776111889 Chr.22: 43973192-43973192 on Build GRCh38
rs956383559 Chr.22: 43973508-43973508 on Build GRCh38
rs752270121 Chr.22: 43973290-43973290 on Build GRCh38
rs930897752 Chr.22: 43973498-43973498 on Build GRCh38
rs562320137 Chr.22: 43973536-43973536 on Build GRCh38
rs762107533 Chr.22: 43973210-43973210 on Build GRCh38
rs757207629 Chr.22: 43973243-43973243 on Build GRCh38
rs766925672 Chr.22: 43973231-43973231 on Build GRCh38
rs141154080 Chr.22: 43973263-43973263 on Build GRCh38
rs9614301 Chr.22: 43973524-43973524 on Build GRCh38
rs920841366 Chr.22: 43973478-43973478 on Build GRCh38
rs549377504 Chr.22: 43973324-43973324 on Build GRCh38
rs769163632 Chr.22: 43973273-43973273 on Build GRCh38
rs77764932 Chr.22: 43973535-43973535 on Build GRCh38
rs113373406 Chr.22: 43973289-43973289 on Build GRCh38
rs750719541 Chr.22: 43973329-43973329 on Build GRCh38
rs11912752 Chr.22: 43973500-43973500 on Build GRCh38
rs1001373691 Chr.22: 43973601-43973601 on Build GRCh38
rs200502238 Chr.22: 43973295-43973295 on Build GRCh38
rs754566377 Chr.22: 43973226-43973226 on Build GRCh38
rs1022498193 Chr.22: 43973534-43973534 on Build GRCh38
rs557170969 Chr.22: 43973539-43973539 on Build GRCh38
rs773639710 Chr.22: 43973317-43973317 on Build GRCh38
rs949747741 Chr.22: 43973547-43973547 on Build GRCh38
rs780263929 Chr.22: 43973338-43973338 on Build GRCh38
rs759640367 Chr.22: 43973559-43973559 on Build GRCh38
rs751131222 Chr.22: 43973218-43973218 on Build GRCh38

View Full Product Details