Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46255571-46256067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PKDREJ
Entrez Gene ID
10343
Gene Name
polycystin (PKD) family receptor for egg jelly
Gene Aliases
-
UniGene
Hs.241383
Gene Chromosome Location
Chr.22: 46255663-46263322 on Build GRCh38
Transcripts
NM_006071.1

SNP Detail

SNP ID Chromosome Location
rs777823744 Chr.22: 46255751-46255751 on Build GRCh38
rs1036776525 Chr.22: 46255670-46255670 on Build GRCh38
rs1025183177 Chr.22: 46255814-46255814 on Build GRCh38
rs576869960 Chr.22: 46255705-46255705 on Build GRCh38
rs376427092 Chr.22: 46255806-46255806 on Build GRCh38
rs932618200 Chr.22: 46255984-46255984 on Build GRCh38
rs999717342 Chr.22: 46255931-46255931 on Build GRCh38
rs1044033932 Chr.22: 46255922-46255922 on Build GRCh38
rs932462537 Chr.22: 46255641-46255641 on Build GRCh38
rs1019432471 Chr.22: 46256000-46256000 on Build GRCh38
rs8135363 Chr.22: 46255816-46255816 on Build GRCh38
rs1015255789 Chr.22: 46255843-46255843 on Build GRCh38
rs755834120 Chr.22: 46255781-46255781 on Build GRCh38
rs984005367 Chr.22: 46255826-46255826 on Build GRCh38
rs939113092 Chr.22: 46255897-46255897 on Build GRCh38
rs41279831 Chr.22: 46255726-46255726 on Build GRCh38
rs373001167 Chr.22: 46256034-46256034 on Build GRCh38
rs527979058 Chr.22: 46255873-46255873 on Build GRCh38
rs527986703 Chr.22: 46255829-46255829 on Build GRCh38
rs999601221 Chr.22: 46255746-46255746 on Build GRCh38
rs906188987 Chr.22: 46255681-46255681 on Build GRCh38
rs764549031 Chr.22: 46255711-46255711 on Build GRCh38
rs1003235279 Chr.22: 46255689-46255689 on Build GRCh38
rs542024482 Chr.22: 46255813-46255813 on Build GRCh38
rs540956993 Chr.22: 46255680-46255680 on Build GRCh38
rs966421150 Chr.22: 46255766-46255766 on Build GRCh38
rs754585870 Chr.22: 46255867-46255867 on Build GRCh38
rs778688064 Chr.22: 46255877-46255877 on Build GRCh38
rs779970610 Chr.22: 46255905-46255907 on Build GRCh38
rs555552312 Chr.22: 46255652-46255652 on Build GRCh38
rs549600328 Chr.22: 46256033-46256034 on Build GRCh38
rs9615870 Chr.22: 46255872-46255872 on Build GRCh38
rs1034363219 Chr.22: 46255798-46255798 on Build GRCh38
rs966874245 Chr.22: 46255727-46255727 on Build GRCh38
rs921184203 Chr.22: 46255972-46255972 on Build GRCh38
rs902678220 Chr.22: 46255923-46255923 on Build GRCh38
rs986592981 Chr.22: 46256032-46256032 on Build GRCh38
rs763196051 Chr.22: 46255653-46255653 on Build GRCh38
rs1011182578 Chr.22: 46255708-46255708 on Build GRCh38
rs909712405 Chr.22: 46255833-46255833 on Build GRCh38
rs572229816 Chr.22: 46255661-46255661 on Build GRCh38
rs553749100 Chr.22: 46255702-46255702 on Build GRCh38
rs1054777025 Chr.22: 46255902-46255904 on Build GRCh38
rs562795892 Chr.22: 46255786-46255786 on Build GRCh38
rs927765045 Chr.22: 46255893-46255893 on Build GRCh38
rs1011130435 Chr.22: 46255922-46255923 on Build GRCh38
rs939733825 Chr.22: 46255665-46255667 on Build GRCh38

View Full Product Details