Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10126569-10127067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
UniGene
Hs.649307
Gene Chromosome Location
Chr.3: 10115649-10127190 on Build GRCh38
Transcripts
NM_018462.4

SNP Detail

SNP ID Chromosome Location
rs527506605 Chr.3: 10126615-10126615 on Build GRCh38
rs942309656 Chr.3: 10126642-10126642 on Build GRCh38
rs986697244 Chr.3: 10126995-10126995 on Build GRCh38
rs995134814 Chr.3: 10126966-10126966 on Build GRCh38
rs1033870873 Chr.3: 10126590-10126590 on Build GRCh38
rs528520969 Chr.3: 10126959-10126959 on Build GRCh38
rs542117291 Chr.3: 10126821-10126821 on Build GRCh38
rs371552157 Chr.3: 10126791-10126791 on Build GRCh38
rs1025092835 Chr.3: 10126620-10126620 on Build GRCh38
rs867298658 Chr.3: 10126787-10126787 on Build GRCh38
rs1003650516 Chr.3: 10126941-10126941 on Build GRCh38
rs895406040 Chr.3: 10126591-10126592 on Build GRCh38
rs962375829 Chr.3: 10126739-10126739 on Build GRCh38
rs1013790503 Chr.3: 10126895-10126895 on Build GRCh38
rs768646335 Chr.3: 10126624-10126625 on Build GRCh38
rs932561636 Chr.3: 10126777-10126777 on Build GRCh38
rs190197748 Chr.3: 10126691-10126691 on Build GRCh38
rs574500515 Chr.3: 10126717-10126717 on Build GRCh38
rs752530126 Chr.3: 10127033-10127033 on Build GRCh38
rs113700441 Chr.3: 10126994-10126994 on Build GRCh38
rs6785976 Chr.3: 10126963-10126963 on Build GRCh38
rs748069870 Chr.3: 10126942-10126942 on Build GRCh38
rs1013674120 Chr.3: 10126599-10126599 on Build GRCh38
rs181379563 Chr.3: 10126880-10126880 on Build GRCh38
rs921141210 Chr.3: 10126771-10126771 on Build GRCh38
rs774778873 Chr.3: 10126680-10126680 on Build GRCh38
rs912394484 Chr.3: 10126798-10126798 on Build GRCh38
rs766082896 Chr.3: 10126709-10126709 on Build GRCh38
rs978334048 Chr.3: 10127025-10127025 on Build GRCh38
rs768841577 Chr.3: 10126629-10126629 on Build GRCh38
rs912498236 Chr.3: 10127016-10127016 on Build GRCh38
rs374406993 Chr.3: 10126838-10126838 on Build GRCh38
rs893098207 Chr.3: 10126786-10126786 on Build GRCh38
rs936637144 Chr.3: 10126885-10126885 on Build GRCh38
rs763535451 Chr.3: 10126619-10126619 on Build GRCh38
rs371722534 Chr.3: 10127028-10127028 on Build GRCh38
rs185958915 Chr.3: 10126993-10126993 on Build GRCh38
rs886597918 Chr.3: 10126933-10126933 on Build GRCh38
rs972963519 Chr.3: 10126605-10126605 on Build GRCh38
rs199877068 Chr.3: 10126892-10126892 on Build GRCh38
rs973751226 Chr.3: 10126753-10126753 on Build GRCh38
rs773198915 Chr.3: 10127019-10127019 on Build GRCh38
rs907015056 Chr.3: 10126884-10126884 on Build GRCh38
rs560156910 Chr.3: 10126926-10126926 on Build GRCh38
rs1027984282 Chr.3: 10126978-10126978 on Build GRCh38
rs1055055792 Chr.3: 10126889-10126889 on Build GRCh38
rs986936194 Chr.3: 10126628-10126628 on Build GRCh38
rs3087695 Chr.3: 10126607-10126607 on Build GRCh38
rs933784889 Chr.3: 10126623-10126623 on Build GRCh38

View Full Product Details