Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11839213-11839542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAMM41
Entrez Gene ID
132001
Gene Name
TAM41 mitochondrial translocator assembly and maintenance homolog
Gene Aliases
C3orf31, RAM41, TAM41
UniGene
Hs.475472
Gene Chromosome Location
Chr.3: 11721896-11846919 on Build GRCh38
Transcripts
NM_001284401.1 NM_001321294.1 NM_001321295.1 NM_138807.3 NR_104314.1 NR_135609.1 NR_135610.1 NR_135611.1 XM_005264873.3 XM_005264875.3 XM_017005725.1 XM_017005726.1 XM_017005727.1 XM_017005728.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4002404 c.347A>G p.N116S Chr.3: 11839286-11839286 on Build GRCh38
159603 c.411G>C p.P137P Chr.3: 11839222-11839222 on Build GRCh38
368139 c.321T>A p.L107L Chr.3: 11839312-11839312 on Build GRCh38
4112493 c.410C>T p.P137L Chr.3: 11839223-11839223 on Build GRCh38
5582714 c.394G>A p.G132R Chr.3: 11839239-11839239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113087354 Chr.3: 11839395-11839395 on Build GRCh38
rs376351296 Chr.3: 11839285-11839285 on Build GRCh38
rs750367417 Chr.3: 11839353-11839353 on Build GRCh38
rs997746870 Chr.3: 11839387-11839387 on Build GRCh38
rs928991899 Chr.3: 11839305-11839305 on Build GRCh38
rs769048276 Chr.3: 11839236-11839236 on Build GRCh38
rs777512768 Chr.3: 11839276-11839288 on Build GRCh38
rs527736395 Chr.3: 11839390-11839390 on Build GRCh38
rs1015371512 Chr.3: 11839518-11839518 on Build GRCh38
rs1001146081 Chr.3: 11839481-11839481 on Build GRCh38
rs303862 Chr.3: 11839342-11839342 on Build GRCh38
rs747219108 Chr.3: 11839235-11839235 on Build GRCh38
rs891552132 Chr.3: 11839372-11839372 on Build GRCh38
rs760607469 Chr.3: 11839369-11839369 on Build GRCh38
rs776821168 Chr.3: 11839245-11839245 on Build GRCh38
rs549788576 Chr.3: 11839520-11839520 on Build GRCh38
rs763439520 Chr.3: 11839307-11839307 on Build GRCh38
rs922476310 Chr.3: 11839281-11839281 on Build GRCh38
rs778287472 Chr.3: 11839348-11839348 on Build GRCh38
rs80019078 Chr.3: 11839462-11839462 on Build GRCh38
rs761903294 Chr.3: 11839284-11839284 on Build GRCh38
rs760293785 Chr.3: 11839331-11839331 on Build GRCh38
rs768961533 Chr.3: 11839337-11839337 on Build GRCh38
rs1034618922 Chr.3: 11839508-11839508 on Build GRCh38
rs951987496 Chr.3: 11839513-11839513 on Build GRCh38
rs1020934934 Chr.3: 11839452-11839452 on Build GRCh38
rs900358954 Chr.3: 11839381-11839381 on Build GRCh38
rs757024775 Chr.3: 11839321-11839322 on Build GRCh38
rs573594197 Chr.3: 11839440-11839440 on Build GRCh38
rs751972820 Chr.3: 11839327-11839327 on Build GRCh38
rs972582394 Chr.3: 11839248-11839248 on Build GRCh38
rs1006229508 Chr.3: 11839517-11839517 on Build GRCh38
rs7641243 Chr.3: 11839286-11839286 on Build GRCh38
rs763701073 Chr.3: 11839333-11839333 on Build GRCh38
rs34626354 Chr.3: 11839501-11839502 on Build GRCh38
rs564104394 Chr.3: 11839396-11839396 on Build GRCh38
rs377637536 Chr.3: 11839318-11839318 on Build GRCh38
rs753581636 Chr.3: 11839276-11839281 on Build GRCh38
rs891194609 Chr.3: 11839434-11839434 on Build GRCh38
rs907394119 Chr.3: 11839428-11839428 on Build GRCh38
rs561202953 Chr.3: 11839414-11839414 on Build GRCh38
rs1009576398 Chr.3: 11839448-11839448 on Build GRCh38

View Full Product Details