Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 20001283-20001829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PP2D1
Entrez Gene ID
151649
Gene Name
protein phosphatase 2C like domain containing 1
Gene Aliases
C3orf48
UniGene
Hs.733161
Gene Chromosome Location
Chr.3: 19979961-20012273 on Build GRCh38
Transcripts
NM_001252657.1 NR_027694.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6019978 c.641C>T p.S214L Chr.3: 20001479-20001479 on Build GRCh38
3945369 c.435A>G p.A145A Chr.3: 20001685-20001685 on Build GRCh38
3945368 c.570T>A p.T190T Chr.3: 20001550-20001550 on Build GRCh38
1177218 c.769G>T p.E257* Chr.3: 20001351-20001351 on Build GRCh38
6215030 c.793C>T p.L265F Chr.3: 20001327-20001327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753748406 Chr.3: 20001482-20001482 on Build GRCh38
rs550751104 Chr.3: 20001507-20001508 on Build GRCh38
rs772388599 Chr.3: 20001553-20001553 on Build GRCh38
rs4103004 Chr.3: 20001342-20001342 on Build GRCh38
rs7652446 Chr.3: 20001751-20001751 on Build GRCh38
rs533784654 Chr.3: 20001358-20001358 on Build GRCh38
rs556321248 Chr.3: 20001611-20001611 on Build GRCh38
rs774558040 Chr.3: 20001771-20001771 on Build GRCh38
rs763566019 Chr.3: 20001742-20001742 on Build GRCh38
rs868398287 Chr.3: 20001755-20001755 on Build GRCh38
rs773667408 Chr.3: 20001740-20001740 on Build GRCh38
rs773470917 Chr.3: 20001618-20001618 on Build GRCh38
rs770147141 Chr.3: 20001739-20001739 on Build GRCh38
rs184958519 Chr.3: 20001390-20001390 on Build GRCh38
rs965511460 Chr.3: 20001550-20001550 on Build GRCh38
rs776967661 Chr.3: 20001664-20001664 on Build GRCh38
rs182437145 Chr.3: 20001341-20001341 on Build GRCh38
rs1007223267 Chr.3: 20001487-20001487 on Build GRCh38
rs267599758 Chr.3: 20001561-20001561 on Build GRCh38
rs567551896 Chr.3: 20001476-20001476 on Build GRCh38
rs757795617 Chr.3: 20001579-20001579 on Build GRCh38
rs570854355 Chr.3: 20001696-20001697 on Build GRCh38
rs866512177 Chr.3: 20001623-20001623 on Build GRCh38
rs994896147 Chr.3: 20001475-20001475 on Build GRCh38
rs746607381 Chr.3: 20001641-20001641 on Build GRCh38
rs755268420 Chr.3: 20001376-20001376 on Build GRCh38
rs866318489 Chr.3: 20001328-20001328 on Build GRCh38
rs566403179 Chr.3: 20001514-20001514 on Build GRCh38
rs149789258 Chr.3: 20001343-20001343 on Build GRCh38
rs748454348 Chr.3: 20001719-20001719 on Build GRCh38
rs766720647 Chr.3: 20001357-20001358 on Build GRCh38
rs998405385 Chr.3: 20001568-20001568 on Build GRCh38
rs534129147 Chr.3: 20001434-20001434 on Build GRCh38
rs201179163 Chr.3: 20001515-20001515 on Build GRCh38
rs189616411 Chr.3: 20001753-20001753 on Build GRCh38
rs963323396 Chr.3: 20001520-20001520 on Build GRCh38
rs117216652 Chr.3: 20001369-20001369 on Build GRCh38
rs897762871 Chr.3: 20001441-20001441 on Build GRCh38
rs145726946 Chr.3: 20001658-20001658 on Build GRCh38
rs1004338455 Chr.3: 20001477-20001477 on Build GRCh38
rs1036684517 Chr.3: 20001436-20001436 on Build GRCh38
rs759664118 Chr.3: 20001774-20001774 on Build GRCh38
rs971406001 Chr.3: 20001668-20001668 on Build GRCh38
rs1032990465 Chr.3: 20001680-20001680 on Build GRCh38
rs917377060 Chr.3: 20001761-20001761 on Build GRCh38
rs979438673 Chr.3: 20001663-20001663 on Build GRCh38
rs546716751 Chr.3: 20001486-20001486 on Build GRCh38
rs745851976 Chr.3: 20001577-20001577 on Build GRCh38
rs577766718 Chr.3: 20001637-20001637 on Build GRCh38
rs577787919 Chr.3: 20001728-20001728 on Build GRCh38
rs199921992 Chr.3: 20001386-20001386 on Build GRCh38
rs199957786 Chr.3: 20001314-20001314 on Build GRCh38

View Full Product Details