Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 29897292-29897802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RBMS3
Entrez Gene ID
27303
Gene Name
RNA binding motif single stranded interacting protein 3
Gene Aliases
-
UniGene
Hs.221436
Gene Chromosome Location
Chr.3: 29280860-30010395 on Build GRCh38
Transcripts
NM_001003792.2 NM_001003793.2 NM_001177711.1 NM_001177712.1 NM_014483.3 XM_005265060.2 XM_005265061.2 XM_005265062.1 XM_005265063.1 XM_005265065.4 XM_017006178.1 XM_017006179.1 XM_017006180.1 XM_017006181.1 XM_017006182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
163963 c.807G>A p.P269P Chr.3: 29897394-29897394 on Build GRCh38
3592308 c.888C>T p.I296I Chr.3: 29897436-29897436 on Build GRCh38
6764234 c.839C>A p.S280Y Chr.3: 29897387-29897387 on Build GRCh38
1043497 c.827G>A p.R276H Chr.3: 29897414-29897414 on Build GRCh38
4116787 c.890C>T p.T297M Chr.3: 29897438-29897438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761135163 Chr.3: 29897436-29897436 on Build GRCh38
rs868740296 Chr.3: 29897392-29897392 on Build GRCh38
rs764138738 Chr.3: 29897347-29897347 on Build GRCh38
rs777794191 Chr.3: 29897518-29897518 on Build GRCh38
rs1044460115 Chr.3: 29897576-29897576 on Build GRCh38
rs13064547 Chr.3: 29897551-29897551 on Build GRCh38
rs140529175 Chr.3: 29897375-29897375 on Build GRCh38
rs376210051 Chr.3: 29897413-29897413 on Build GRCh38
rs942109222 Chr.3: 29897575-29897575 on Build GRCh38
rs977441221 Chr.3: 29897772-29897772 on Build GRCh38
rs747705395 Chr.3: 29897679-29897679 on Build GRCh38
rs765475614 Chr.3: 29897357-29897357 on Build GRCh38
rs777313346 Chr.3: 29897713-29897713 on Build GRCh38
rs905535332 Chr.3: 29897590-29897590 on Build GRCh38
rs1014627721 Chr.3: 29897563-29897564 on Build GRCh38
rs964377165 Chr.3: 29897726-29897726 on Build GRCh38
rs150456438 Chr.3: 29897707-29897707 on Build GRCh38
rs879172189 Chr.3: 29897424-29897424 on Build GRCh38
rs13064143 Chr.3: 29897560-29897560 on Build GRCh38
rs370849321 Chr.3: 29897439-29897439 on Build GRCh38
rs13082457 Chr.3: 29897481-29897481 on Build GRCh38
rs762064115 Chr.3: 29897351-29897351 on Build GRCh38
rs536319618 Chr.3: 29897776-29897776 on Build GRCh38
rs765245946 Chr.3: 29897480-29897480 on Build GRCh38
rs746699630 Chr.3: 29897501-29897501 on Build GRCh38
rs777084340 Chr.3: 29897469-29897469 on Build GRCh38
rs750653218 Chr.3: 29897493-29897493 on Build GRCh38
rs13064142 Chr.3: 29897559-29897559 on Build GRCh38
rs200990419 Chr.3: 29897373-29897373 on Build GRCh38
rs745656126 Chr.3: 29897393-29897393 on Build GRCh38
rs191759920 Chr.3: 29897579-29897579 on Build GRCh38
rs1022220142 Chr.3: 29897646-29897646 on Build GRCh38
rs923180562 Chr.3: 29897498-29897498 on Build GRCh38
rs775953185 Chr.3: 29897634-29897634 on Build GRCh38
rs755585331 Chr.3: 29897573-29897573 on Build GRCh38
rs186505428 Chr.3: 29897525-29897525 on Build GRCh38
rs758326781 Chr.3: 29897431-29897431 on Build GRCh38
rs753778967 Chr.3: 29897519-29897519 on Build GRCh38
rs929180830 Chr.3: 29897517-29897517 on Build GRCh38
rs752510414 Chr.3: 29897503-29897503 on Build GRCh38
rs547370537 Chr.3: 29897681-29897681 on Build GRCh38
rs779486787 Chr.3: 29897359-29897359 on Build GRCh38
rs968194863 Chr.3: 29897396-29897396 on Build GRCh38
rs975703575 Chr.3: 29897739-29897739 on Build GRCh38
rs761906709 Chr.3: 29897471-29897471 on Build GRCh38
rs1022875728 Chr.3: 29897564-29897564 on Build GRCh38
rs13060313 Chr.3: 29897536-29897536 on Build GRCh38
rs779831008 Chr.3: 29897486-29897486 on Build GRCh38
rs752159305 Chr.3: 29897529-29897529 on Build GRCh38
rs1002861226 Chr.3: 29897578-29897578 on Build GRCh38
rs200187522 Chr.3: 29897394-29897394 on Build GRCh38
rs377447342 Chr.3: 29897348-29897348 on Build GRCh38
rs745458478 Chr.3: 29897414-29897414 on Build GRCh38
rs968875534 Chr.3: 29897650-29897650 on Build GRCh38
rs750598571 Chr.3: 29897359-29897359 on Build GRCh38
rs373893728 Chr.3: 29897504-29897504 on Build GRCh38
rs892049404 Chr.3: 29897604-29897604 on Build GRCh38
rs756062242 Chr.3: 29897390-29897390 on Build GRCh38
rs746683984 Chr.3: 29897413-29897414 on Build GRCh38
rs13059964 Chr.3: 29897465-29897465 on Build GRCh38
rs770522581 Chr.3: 29897408-29897408 on Build GRCh38
rs13060292 Chr.3: 29897475-29897475 on Build GRCh38
rs923231410 Chr.3: 29897740-29897740 on Build GRCh38
rs748774302 Chr.3: 29897399-29897399 on Build GRCh38
rs775917846 Chr.3: 29897420-29897420 on Build GRCh38
rs905875980 Chr.3: 29897577-29897577 on Build GRCh38
rs754783380 Chr.3: 29897364-29897364 on Build GRCh38
rs763235834 Chr.3: 29897496-29897496 on Build GRCh38
rs780977358 Chr.3: 29897369-29897369 on Build GRCh38

View Full Product Details