Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39394390-39394940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A38
Entrez Gene ID
54977
Gene Name
solute carrier family 25 member 38
Gene Aliases
SIDBA2
UniGene
Hs.369615
Gene Chromosome Location
Chr.3: 39383324-39397328 on Build GRCh38
Transcripts
NM_017875.2 XM_006713214.1 XM_011533869.2 XM_017006710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780726 c.714G>A p.A238A Chr.3: 39394498-39394498 on Build GRCh38
6780724 c.760T>C p.W254R Chr.3: 39394544-39394544 on Build GRCh38
446484 c.767G>A p.G256D Chr.3: 39394551-39394551 on Build GRCh38
3593922 c.641C>T p.T214I Chr.3: 39394425-39394425 on Build GRCh38
7151463 c.663C>T p.F221F Chr.3: 39394447-39394447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746340072 Chr.3: 39394797-39394797 on Build GRCh38
rs78965445 Chr.3: 39394674-39394674 on Build GRCh38
rs1055187958 Chr.3: 39394845-39394845 on Build GRCh38
rs201190883 Chr.3: 39394536-39394536 on Build GRCh38
rs145760389 Chr.3: 39394417-39394417 on Build GRCh38
rs200152652 Chr.3: 39394486-39394486 on Build GRCh38
rs375346031 Chr.3: 39394616-39394616 on Build GRCh38
rs748574910 Chr.3: 39394490-39394490 on Build GRCh38
rs938087978 Chr.3: 39394829-39394829 on Build GRCh38
rs566939365 Chr.3: 39394713-39394713 on Build GRCh38
rs764125735 Chr.3: 39394436-39394436 on Build GRCh38
rs766387890 Chr.3: 39394578-39394578 on Build GRCh38
rs371503933 Chr.3: 39394506-39394506 on Build GRCh38
rs763845960 Chr.3: 39394564-39394564 on Build GRCh38
rs868782201 Chr.3: 39394862-39394862 on Build GRCh38
rs533169173 Chr.3: 39394824-39394824 on Build GRCh38
rs1016379309 Chr.3: 39394673-39394673 on Build GRCh38
rs766505056 Chr.3: 39394458-39394458 on Build GRCh38
rs751713207 Chr.3: 39394463-39394463 on Build GRCh38
rs886726241 Chr.3: 39394905-39394905 on Build GRCh38
rs755205622 Chr.3: 39394467-39394467 on Build GRCh38
rs778318538 Chr.3: 39394498-39394498 on Build GRCh38
rs1002284642 Chr.3: 39394446-39394446 on Build GRCh38
rs1006829737 Chr.3: 39394632-39394632 on Build GRCh38
rs756472175 Chr.3: 39394497-39394497 on Build GRCh38
rs760488714 Chr.3: 39394544-39394544 on Build GRCh38
rs770557347 Chr.3: 39394509-39394509 on Build GRCh38
rs970355195 Chr.3: 39394740-39394740 on Build GRCh38
rs1014029461 Chr.3: 39394884-39394884 on Build GRCh38
rs758451219 Chr.3: 39394445-39394445 on Build GRCh38
rs760483420 Chr.3: 39394422-39394422 on Build GRCh38
rs925070603 Chr.3: 39394817-39394817 on Build GRCh38
rs767746893 Chr.3: 39394615-39394615 on Build GRCh38
rs745464587 Chr.3: 39394528-39394528 on Build GRCh38
rs35616367 Chr.3: 39394736-39394736 on Build GRCh38
rs67035835 Chr.3: 39394747-39394747 on Build GRCh38
rs368651459 Chr.3: 39394594-39394594 on Build GRCh38
rs121918332 Chr.3: 39394574-39394574 on Build GRCh38
rs776536241 Chr.3: 39394566-39394566 on Build GRCh38
rs753815164 Chr.3: 39394439-39394439 on Build GRCh38
rs6793498 Chr.3: 39394869-39394869 on Build GRCh38
rs781372292 Chr.3: 39394449-39394462 on Build GRCh38
rs370492216 Chr.3: 39394599-39394599 on Build GRCh38
rs376371226 Chr.3: 39394472-39394472 on Build GRCh38
rs112346314 Chr.3: 39394691-39394691 on Build GRCh38
rs775110434 Chr.3: 39394543-39394543 on Build GRCh38
rs67353739 Chr.3: 39394816-39394816 on Build GRCh38
rs751652986 Chr.3: 39394569-39394569 on Build GRCh38
rs1047712252 Chr.3: 39394904-39394904 on Build GRCh38
rs1024657404 Chr.3: 39394738-39394739 on Build GRCh38
rs955189994 Chr.3: 39394724-39394724 on Build GRCh38
rs774006570 Chr.3: 39394510-39394510 on Build GRCh38
rs913834984 Chr.3: 39394738-39394738 on Build GRCh38
rs1026959948 Chr.3: 39394609-39394609 on Build GRCh38

View Full Product Details