Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39408282-39408795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPSA
Entrez Gene ID
3921
Gene Name
ribosomal protein SA
Gene Aliases
37LRP, 67LR, ICAS, LAMBR, LAMR1, LBP, LBP/p40, LRP, LRP/LR, NEM/1CHD4, SA, lamR, p40
UniGene
Hs.449909
Gene Chromosome Location
Chr.3: 39406689-39412542 on Build GRCh38
Transcripts
NM_001304288.1 NM_002295.5 NR_002325.1

Gene Symbol
SNORA6
Entrez Gene ID
574040
Gene Name
small nucleolar RNA, H/ACA box 6
Gene Aliases
ACA6
UniGene
Hs.745521
Gene Chromosome Location
Chr.3: 39408391-39408539 on Build GRCh38
Transcripts
NM_001304288.1 NM_002295.5 NR_002325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4447058 c.134-3T>A p.? Chr.3: 39408603-39408603 on Build GRCh38
4722785 c.181G>A p.A61T Chr.3: 39408653-39408653 on Build GRCh38
7087829 c.230T>C p.I77T Chr.3: 39408702-39408702 on Build GRCh38
345516 c.233C>A p.S78Y Chr.3: 39408705-39408705 on Build GRCh38
5418589 c.180G>A p.L60L Chr.3: 39408652-39408652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747622236 Chr.3: 39408462-39408462 on Build GRCh38
rs373123514 Chr.3: 39408561-39408561 on Build GRCh38
rs780348162 Chr.3: 39408663-39408663 on Build GRCh38
rs893556660 Chr.3: 39408702-39408702 on Build GRCh38
rs1052138765 Chr.3: 39408676-39408676 on Build GRCh38
rs930632791 Chr.3: 39408527-39408527 on Build GRCh38
rs397514762 Chr.3: 39408633-39408633 on Build GRCh38
rs757348877 Chr.3: 39408639-39408639 on Build GRCh38
rs190008375 Chr.3: 39408740-39408740 on Build GRCh38
rs543844278 Chr.3: 39408753-39408753 on Build GRCh38
rs141963705 Chr.3: 39408305-39408305 on Build GRCh38
rs372380170 Chr.3: 39408594-39408594 on Build GRCh38
rs753429644 Chr.3: 39408404-39408404 on Build GRCh38
rs554479658 Chr.3: 39408659-39408659 on Build GRCh38
rs3772139 Chr.3: 39408319-39408319 on Build GRCh38
rs772596551 Chr.3: 39408373-39408373 on Build GRCh38
rs538269180 Chr.3: 39408413-39408413 on Build GRCh38
rs956175364 Chr.3: 39408349-39408349 on Build GRCh38
rs762826480 Chr.3: 39408498-39408498 on Build GRCh38
rs367800822 Chr.3: 39408355-39408355 on Build GRCh38
rs779206833 Chr.3: 39408451-39408451 on Build GRCh38
rs199984839 Chr.3: 39408389-39408389 on Build GRCh38
rs1030681038 Chr.3: 39408394-39408394 on Build GRCh38
rs748667898 Chr.3: 39408674-39408674 on Build GRCh38
rs747150713 Chr.3: 39408487-39408487 on Build GRCh38
rs767372989 Chr.3: 39408355-39408359 on Build GRCh38
rs760709940 Chr.3: 39408398-39408399 on Build GRCh38
rs369937486 Chr.3: 39408566-39408566 on Build GRCh38
rs79914243 Chr.3: 39408562-39408562 on Build GRCh38
rs765243939 Chr.3: 39408767-39408767 on Build GRCh38
rs753979701 Chr.3: 39408534-39408534 on Build GRCh38
rs759349152 Chr.3: 39408601-39408601 on Build GRCh38
rs961880947 Chr.3: 39408480-39408480 on Build GRCh38
rs779088057 Chr.3: 39408539-39408539 on Build GRCh38
rs889327212 Chr.3: 39408337-39408337 on Build GRCh38
rs771130669 Chr.3: 39408359-39408359 on Build GRCh38
rs750580550 Chr.3: 39408655-39408655 on Build GRCh38
rs759985171 Chr.3: 39408369-39408369 on Build GRCh38
rs1009105138 Chr.3: 39408346-39408346 on Build GRCh38
rs771204845 Chr.3: 39408450-39408450 on Build GRCh38
rs758761014 Chr.3: 39408557-39408557 on Build GRCh38
rs760160875 Chr.3: 39408382-39408382 on Build GRCh38
rs746204067 Chr.3: 39408455-39408455 on Build GRCh38
rs763721384 Chr.3: 39408399-39408399 on Build GRCh38
rs762698689 Chr.3: 39408592-39408592 on Build GRCh38
rs186381702 Chr.3: 39408377-39408377 on Build GRCh38
rs770875444 Chr.3: 39408499-39408499 on Build GRCh38
rs752252932 Chr.3: 39408528-39408528 on Build GRCh38
rs780115735 Chr.3: 39408419-39408419 on Build GRCh38
rs17849988 Chr.3: 39408664-39408664 on Build GRCh38
rs751475099 Chr.3: 39408500-39408500 on Build GRCh38
rs559024939 Chr.3: 39408329-39408329 on Build GRCh38
rs767985148 Chr.3: 39408467-39408467 on Build GRCh38
rs774155963 Chr.3: 39408352-39408353 on Build GRCh38
rs397514763 Chr.3: 39408644-39408644 on Build GRCh38
rs201530807 Chr.3: 39408741-39408741 on Build GRCh38
rs777216757 Chr.3: 39408505-39408505 on Build GRCh38
rs763123376 Chr.3: 39408769-39408769 on Build GRCh38
rs766379923 Chr.3: 39408481-39408481 on Build GRCh38
rs1011924200 Chr.3: 39408306-39408306 on Build GRCh38
rs761325221 Chr.3: 39408479-39408479 on Build GRCh38
rs756143134 Chr.3: 39408432-39408432 on Build GRCh38
rs757318774 Chr.3: 39408537-39408537 on Build GRCh38
rs777909736 Chr.3: 39408448-39408448 on Build GRCh38
rs755340646 Chr.3: 39408670-39408670 on Build GRCh38
rs773913524 Chr.3: 39408734-39408734 on Build GRCh38
rs758306417 Chr.3: 39408418-39408418 on Build GRCh38
rs764884464 Chr.3: 39408409-39408409 on Build GRCh38
rs1050673049 Chr.3: 39408333-39408333 on Build GRCh38
rs750525243 Chr.3: 39408768-39408768 on Build GRCh38
rs781714830 Chr.3: 39408672-39408672 on Build GRCh38
rs752628562 Chr.3: 39408614-39408614 on Build GRCh38
rs747434456 Chr.3: 39408665-39408665 on Build GRCh38
rs765372610 Chr.3: 39408650-39408650 on Build GRCh38
rs200338902 Chr.3: 39408756-39408756 on Build GRCh38
rs377192766 Chr.3: 39408460-39408460 on Build GRCh38
rs772994722 Chr.3: 39408583-39408583 on Build GRCh38
rs770735061 Chr.3: 39408721-39408721 on Build GRCh38
rs774706905 Chr.3: 39408366-39408366 on Build GRCh38
rs767454762 Chr.3: 39408607-39408607 on Build GRCh38
rs775128825 Chr.3: 39408398-39408398 on Build GRCh38
rs369941023 Chr.3: 39408549-39408549 on Build GRCh38
rs577387094 Chr.3: 39408700-39408700 on Build GRCh38
rs772481521 Chr.3: 39408330-39408330 on Build GRCh38
rs751840571 Chr.3: 39408504-39408505 on Build GRCh38
rs3772138 Chr.3: 39408574-39408574 on Build GRCh38
rs769363013 Chr.3: 39408685-39408685 on Build GRCh38
rs200885445 Chr.3: 39408486-39408486 on Build GRCh38
rs776218441 Chr.3: 39408470-39408470 on Build GRCh38
rs772476262 Chr.3: 39408459-39408459 on Build GRCh38
rs759041901 Chr.3: 39408354-39408357 on Build GRCh38
rs546066905 Chr.3: 39408706-39408706 on Build GRCh38
rs574844853 Chr.3: 39408565-39408565 on Build GRCh38
rs761531263 Chr.3: 39408405-39408405 on Build GRCh38
rs758487900 Chr.3: 39408464-39408469 on Build GRCh38
rs755107896 Chr.3: 39408581-39408582 on Build GRCh38
rs764018267 Chr.3: 39408399-39408400 on Build GRCh38
rs749408758 Chr.3: 39408449-39408449 on Build GRCh38
rs751604540 Chr.3: 39408430-39408430 on Build GRCh38
rs750563864 Chr.3: 39408449-39408449 on Build GRCh38
rs747671119 Chr.3: 39408572-39408572 on Build GRCh38

View Full Product Details