Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42502461-42502956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VIPR1
Entrez Gene ID
7433
Gene Name
vasoactive intestinal peptide receptor 1
Gene Aliases
HVR1, II, PACAP-R-2, PACAP-R2, RDC1, V1RG, VAPC1, VIP-R-1, VIPR, VIRG, VPAC1, VPAC1R, VPCAP1R
UniGene
Hs.348500
Gene Chromosome Location
Chr.3: 42489299-42537573 on Build GRCh38
Transcripts
NM_001251882.1 NM_001251883.1 NM_001251884.1 NM_001251885.1 NM_004624.3 XM_005265437.2 XM_005265438.3 XM_005265439.2 XM_011534079.1 XM_011534080.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661141 c.78+2T>G p.? Chr.3: 42502815-42502815 on Build GRCh38
1633169 c.24C>A p.P8P Chr.3: 42502759-42502759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs957622970 Chr.3: 42502654-42502654 on Build GRCh38
rs377752938 Chr.3: 42502926-42502926 on Build GRCh38
rs1043293013 Chr.3: 42502722-42502722 on Build GRCh38
rs921738974 Chr.3: 42502875-42502875 on Build GRCh38
rs565026856 Chr.3: 42502497-42502497 on Build GRCh38
rs912437483 Chr.3: 42502483-42502483 on Build GRCh38
rs922028124 Chr.3: 42502692-42502692 on Build GRCh38
rs904754223 Chr.3: 42502910-42502910 on Build GRCh38
rs891773249 Chr.3: 42502709-42502709 on Build GRCh38
rs765645789 Chr.3: 42502805-42502805 on Build GRCh38
rs904903064 Chr.3: 42502719-42502719 on Build GRCh38
rs867160911 Chr.3: 42502653-42502653 on Build GRCh38
rs1026843123 Chr.3: 42502867-42502867 on Build GRCh38
rs543516778 Chr.3: 42502649-42502649 on Build GRCh38
rs371174986 Chr.3: 42502740-42502740 on Build GRCh38
rs1034953478 Chr.3: 42502792-42502792 on Build GRCh38
rs926380228 Chr.3: 42502906-42502906 on Build GRCh38
rs1056063438 Chr.3: 42502934-42502934 on Build GRCh38
rs1046945605 Chr.3: 42502683-42502683 on Build GRCh38
rs907398588 Chr.3: 42502589-42502589 on Build GRCh38
rs1012006346 Chr.3: 42502658-42502658 on Build GRCh38
rs959411371 Chr.3: 42502614-42502614 on Build GRCh38
rs527949035 Chr.3: 42502624-42502624 on Build GRCh38
rs967368002 Chr.3: 42502669-42502669 on Build GRCh38
rs912615615 Chr.3: 42502677-42502677 on Build GRCh38
rs946029550 Chr.3: 42502902-42502902 on Build GRCh38
rs977356748 Chr.3: 42502511-42502511 on Build GRCh38
rs1002112779 Chr.3: 42502606-42502606 on Build GRCh38
rs559657095 Chr.3: 42502894-42502894 on Build GRCh38
rs933156111 Chr.3: 42502880-42502880 on Build GRCh38
rs909123144 Chr.3: 42502659-42502659 on Build GRCh38
rs556062853 Chr.3: 42502500-42502501 on Build GRCh38
rs893284159 Chr.3: 42502760-42502760 on Build GRCh38
rs949060625 Chr.3: 42502885-42502885 on Build GRCh38
rs548962550 Chr.3: 42502861-42502861 on Build GRCh38
rs886761207 Chr.3: 42502547-42502547 on Build GRCh38
rs913322517 Chr.3: 42502895-42502895 on Build GRCh38
rs991765889 Chr.3: 42502615-42502615 on Build GRCh38
rs947500783 Chr.3: 42502932-42502932 on Build GRCh38
rs1033131654 Chr.3: 42502541-42502541 on Build GRCh38
rs981721189 Chr.3: 42502905-42502905 on Build GRCh38
rs933426441 Chr.3: 42502693-42502693 on Build GRCh38
rs529298769 Chr.3: 42502831-42502831 on Build GRCh38
rs975248452 Chr.3: 42502679-42502679 on Build GRCh38
rs1020538812 Chr.3: 42502808-42502808 on Build GRCh38
rs941864492 Chr.3: 42502502-42502502 on Build GRCh38
rs190835822 Chr.3: 42502519-42502519 on Build GRCh38
rs964221665 Chr.3: 42502678-42502678 on Build GRCh38
rs986916075 Chr.3: 42502675-42502675 on Build GRCh38
rs762139215 Chr.3: 42502786-42502786 on Build GRCh38
rs1011872628 Chr.3: 42502776-42502776 on Build GRCh38
rs941870038 Chr.3: 42502662-42502662 on Build GRCh38
rs900622832 Chr.3: 42502509-42502509 on Build GRCh38
rs937526208 Chr.3: 42502721-42502721 on Build GRCh38
rs868319589 Chr.3: 42502909-42502909 on Build GRCh38
rs974586163 Chr.3: 42502868-42502868 on Build GRCh38
rs982697178 Chr.3: 42502869-42502869 on Build GRCh38
rs997520778 Chr.3: 42502512-42502512 on Build GRCh38
rs985886063 Chr.3: 42502876-42502876 on Build GRCh38
rs946039463 Chr.3: 42502710-42502710 on Build GRCh38
rs867746375 Chr.3: 42502860-42502860 on Build GRCh38
rs950400602 Chr.3: 42502646-42502646 on Build GRCh38
rs968880614 Chr.3: 42502585-42502585 on Build GRCh38
rs967728248 Chr.3: 42502500-42502500 on Build GRCh38
rs1010471084 Chr.3: 42502561-42502561 on Build GRCh38
rs1039311966 Chr.3: 42502682-42502682 on Build GRCh38
rs1033433456 Chr.3: 42502607-42502607 on Build GRCh38
rs1055941562 Chr.3: 42502758-42502758 on Build GRCh38
rs1001696349 Chr.3: 42502777-42502777 on Build GRCh38
rs903191700 Chr.3: 42502827-42502828 on Build GRCh38
rs983116912 Chr.3: 42502855-42502855 on Build GRCh38
rs1039076552 Chr.3: 42502507-42502507 on Build GRCh38
rs868167170 Chr.3: 42502569-42502569 on Build GRCh38
rs1021898639 Chr.3: 42502566-42502566 on Build GRCh38
rs750829637 Chr.3: 42502806-42502806 on Build GRCh38
rs1051870228 Chr.3: 42502701-42502701 on Build GRCh38
rs563785977 Chr.3: 42502827-42502827 on Build GRCh38
rs892036358 Chr.3: 42502557-42502557 on Build GRCh38
rs909019581 Chr.3: 42502499-42502499 on Build GRCh38
rs917558184 Chr.3: 42502623-42502623 on Build GRCh38
rs919848588 Chr.3: 42502680-42502680 on Build GRCh38
rs528717313 Chr.3: 42502917-42502917 on Build GRCh38
rs376180580 Chr.3: 42502908-42502908 on Build GRCh38

View Full Product Details