Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47017103-47017610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SETD2
Entrez Gene ID
29072
Gene Name
SET domain containing 2
Gene Aliases
HBP231, HIF-1, HIP-1, HSPC069, HYPB, KMT3A, LLS, SET2, p231HBP
UniGene
Hs.517941
Gene Chromosome Location
Chr.3: 47016408-47164109 on Build GRCh38
Transcripts
NM_014159.6 XM_011533631.2 XM_011533632.2 XM_011533633.2 XM_011533634.1 XM_017006270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625104 c.7580C>A p.P2527H Chr.3: 47017208-47017208 on Build GRCh38
6926585 c.7570T>C p.C2524R Chr.3: 47017218-47017218 on Build GRCh38
6775845 c.7602G>A p.E2534E Chr.3: 47017186-47017186 on Build GRCh38
7184984 c.7592A>T p.E2531V Chr.3: 47017196-47017196 on Build GRCh38
6836009 c.6149C>T p.A2050V Chr.3: 47017130-47017130 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs923629681 Chr.3: 47017506-47017506 on Build GRCh38
rs753072684 Chr.3: 47017272-47017273 on Build GRCh38
rs935718573 Chr.3: 47017449-47017449 on Build GRCh38
rs982052566 Chr.3: 47017394-47017394 on Build GRCh38
rs773606468 Chr.3: 47017139-47017139 on Build GRCh38
rs1033971564 Chr.3: 47017312-47017312 on Build GRCh38
rs915018338 Chr.3: 47017484-47017484 on Build GRCh38
rs759966189 Chr.3: 47017215-47017215 on Build GRCh38
rs1037161059 Chr.3: 47017569-47017569 on Build GRCh38
rs1006981047 Chr.3: 47017313-47017313 on Build GRCh38
rs924232750 Chr.3: 47017437-47017437 on Build GRCh38
rs940072528 Chr.3: 47017553-47017553 on Build GRCh38
rs199973330 Chr.3: 47017200-47017200 on Build GRCh38
rs568921177 Chr.3: 47017445-47017445 on Build GRCh38
rs371454381 Chr.3: 47017280-47017280 on Build GRCh38
rs1038486814 Chr.3: 47017583-47017583 on Build GRCh38
rs551975319 Chr.3: 47017518-47017518 on Build GRCh38
rs977177090 Chr.3: 47017399-47017399 on Build GRCh38
rs962748637 Chr.3: 47017370-47017370 on Build GRCh38
rs889966286 Chr.3: 47017279-47017279 on Build GRCh38
rs898483545 Chr.3: 47017575-47017576 on Build GRCh38
rs758897985 Chr.3: 47017513-47017513 on Build GRCh38
rs368761115 Chr.3: 47017163-47017163 on Build GRCh38
rs921215776 Chr.3: 47017496-47017496 on Build GRCh38
rs972735700 Chr.3: 47017382-47017382 on Build GRCh38
rs566691365 Chr.3: 47017560-47017560 on Build GRCh38
rs974360281 Chr.3: 47017407-47017407 on Build GRCh38
rs959248245 Chr.3: 47017479-47017479 on Build GRCh38
rs79781114 Chr.3: 47017551-47017551 on Build GRCh38
rs1042058100 Chr.3: 47017146-47017146 on Build GRCh38
rs1016991030 Chr.3: 47017337-47017337 on Build GRCh38
rs879918934 Chr.3: 47017540-47017540 on Build GRCh38
rs767954928 Chr.3: 47017246-47017246 on Build GRCh38
rs1015979303 Chr.3: 47017145-47017145 on Build GRCh38
rs112308602 Chr.3: 47017307-47017307 on Build GRCh38
rs768564000 Chr.3: 47017128-47017128 on Build GRCh38
rs933614274 Chr.3: 47017507-47017507 on Build GRCh38
rs946436786 Chr.3: 47017488-47017488 on Build GRCh38
rs890008999 Chr.3: 47017544-47017544 on Build GRCh38
rs763210716 Chr.3: 47017141-47017141 on Build GRCh38
rs990464567 Chr.3: 47017481-47017481 on Build GRCh38
rs927836319 Chr.3: 47017432-47017432 on Build GRCh38
rs1042110913 Chr.3: 47017493-47017493 on Build GRCh38
rs764413274 Chr.3: 47017293-47017293 on Build GRCh38
rs1030284173 Chr.3: 47017275-47017275 on Build GRCh38
rs550447003 Chr.3: 47017424-47017424 on Build GRCh38
rs990629496 Chr.3: 47017480-47017480 on Build GRCh38
rs533392234 Chr.3: 47017497-47017497 on Build GRCh38
rs770936612 Chr.3: 47017183-47017183 on Build GRCh38
rs950731134 Chr.3: 47017384-47017384 on Build GRCh38
rs753191399 Chr.3: 47017265-47017265 on Build GRCh38
rs753486666 Chr.3: 47017298-47017298 on Build GRCh38
rs1025590485 Chr.3: 47017383-47017383 on Build GRCh38
rs902794659 Chr.3: 47017180-47017180 on Build GRCh38
rs996160516 Chr.3: 47017309-47017309 on Build GRCh38

View Full Product Details