Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47235434-47235943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF9
Entrez Gene ID
64147
Gene Name
kinesin family member 9
Gene Aliases
-
UniGene
Hs.373947
Gene Chromosome Location
Chr.3: 47228026-47282847 on Build GRCh38
Transcripts
NM_001134878.1 NM_022342.4 NM_182902.3 XM_005265388.4 XM_005265389.4 XM_006713291.3 XM_011534003.2 XM_011534004.2 XM_011534005.2 XM_011534006.2 XM_011534007.2 XM_017007029.1 XM_017007030.1 XM_017007031.1 NR_033373.1

Gene Symbol
KIF9-AS1
Entrez Gene ID
285352
Gene Name
KIF9 antisense RNA 1
Gene Aliases
-
UniGene
Hs.570631
Gene Chromosome Location
Chr.3: 47164370-47244116 on Build GRCh38
Transcripts
NM_001134878.1 NM_022342.4 NM_182902.3 XM_005265388.4 XM_005265389.4 XM_006713291.3 XM_011534003.2 XM_011534004.2 XM_011534005.2 XM_011534006.2 XM_011534007.2 XM_017007029.1 XM_017007030.1 XM_017007031.1 NR_033373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4836216 c.2286C>T p.I762I Chr.3: 47235549-47235549 on Build GRCh38
7076461 c.2314G>C p.E772Q Chr.3: 47235521-47235521 on Build GRCh38
6251155 c.2322+7C>A p.? Chr.3: 47235506-47235506 on Build GRCh38
5040294 c.2282C>T p.S761F Chr.3: 47235553-47235553 on Build GRCh38
5043912 c.2306T>C p.V769A Chr.3: 47235529-47235529 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756912089 Chr.3: 47235644-47235644 on Build GRCh38
rs1028771086 Chr.3: 47235846-47235846 on Build GRCh38
rs763869822 Chr.3: 47235635-47235635 on Build GRCh38
rs761212270 Chr.3: 47235909-47235909 on Build GRCh38
rs779910422 Chr.3: 47235659-47235659 on Build GRCh38
rs2276854 Chr.3: 47235478-47235478 on Build GRCh38
rs918242936 Chr.3: 47235468-47235468 on Build GRCh38
rs371418079 Chr.3: 47235652-47235652 on Build GRCh38
rs551701052 Chr.3: 47235704-47235704 on Build GRCh38
rs537846834 Chr.3: 47235802-47235802 on Build GRCh38
rs950283301 Chr.3: 47235887-47235887 on Build GRCh38
rs912773323 Chr.3: 47235888-47235889 on Build GRCh38
rs534526575 Chr.3: 47235721-47235721 on Build GRCh38
rs987498924 Chr.3: 47235875-47235875 on Build GRCh38
rs1049182777 Chr.3: 47235696-47235696 on Build GRCh38
rs918883076 Chr.3: 47235876-47235876 on Build GRCh38
rs753104206 Chr.3: 47235541-47235541 on Build GRCh38
rs769491934 Chr.3: 47235883-47235883 on Build GRCh38
rs897542915 Chr.3: 47235661-47235661 on Build GRCh38
rs907074943 Chr.3: 47235761-47235761 on Build GRCh38
rs1029624732 Chr.3: 47235766-47235766 on Build GRCh38
rs753122745 Chr.3: 47235629-47235629 on Build GRCh38
rs189770175 Chr.3: 47235869-47235869 on Build GRCh38
rs539897039 Chr.3: 47235616-47235616 on Build GRCh38
rs556081983 Chr.3: 47235729-47235729 on Build GRCh38
rs773024481 Chr.3: 47235501-47235501 on Build GRCh38
rs112253265 Chr.3: 47235797-47235797 on Build GRCh38
rs769503524 Chr.3: 47235496-47235497 on Build GRCh38
rs746795067 Chr.3: 47235576-47235576 on Build GRCh38
rs777506297 Chr.3: 47235553-47235553 on Build GRCh38
rs754730396 Chr.3: 47235666-47235666 on Build GRCh38
rs778326150 Chr.3: 47235650-47235650 on Build GRCh38
rs1036071399 Chr.3: 47235620-47235620 on Build GRCh38
rs763055489 Chr.3: 47235600-47235600 on Build GRCh38
rs759969622 Chr.3: 47235523-47235523 on Build GRCh38
rs755885776 Chr.3: 47235544-47235544 on Build GRCh38
rs940187595 Chr.3: 47235598-47235598 on Build GRCh38
rs745839521 Chr.3: 47235563-47235563 on Build GRCh38
rs772014179 Chr.3: 47235569-47235569 on Build GRCh38
rs762918537 Chr.3: 47235546-47235546 on Build GRCh38
rs1031769233 Chr.3: 47235796-47235796 on Build GRCh38
rs567046208 Chr.3: 47235720-47235720 on Build GRCh38
rs375678018 Chr.3: 47235491-47235491 on Build GRCh38
rs776650017 Chr.3: 47235657-47235657 on Build GRCh38
rs776507414 Chr.3: 47235646-47235646 on Build GRCh38
rs770017686 Chr.3: 47235585-47235585 on Build GRCh38
rs753490137 Chr.3: 47235641-47235641 on Build GRCh38
rs568863417 Chr.3: 47235619-47235619 on Build GRCh38
rs143906701 Chr.3: 47235575-47235575 on Build GRCh38
rs368001559 Chr.3: 47235626-47235626 on Build GRCh38
rs539479959 Chr.3: 47235701-47235701 on Build GRCh38
rs892990670 Chr.3: 47235655-47235655 on Build GRCh38
rs368210479 Chr.3: 47235531-47235531 on Build GRCh38
rs753399921 Chr.3: 47235560-47235560 on Build GRCh38
rs773338332 Chr.3: 47235497-47235497 on Build GRCh38
rs1020188541 Chr.3: 47235700-47235700 on Build GRCh38
rs750941036 Chr.3: 47235728-47235728 on Build GRCh38
rs930323960 Chr.3: 47235674-47235674 on Build GRCh38
rs144410264 Chr.3: 47235594-47235594 on Build GRCh38
rs747662323 Chr.3: 47235801-47235801 on Build GRCh38
rs751974420 Chr.3: 47235521-47235521 on Build GRCh38
rs998649809 Chr.3: 47235777-47235777 on Build GRCh38
rs115961092 Chr.3: 47235832-47235832 on Build GRCh38
rs1017500912 Chr.3: 47235817-47235817 on Build GRCh38
rs954064020 Chr.3: 47235856-47235856 on Build GRCh38
rs762064014 Chr.3: 47235479-47235479 on Build GRCh38
rs746813158 Chr.3: 47235662-47235662 on Build GRCh38
rs962938211 Chr.3: 47235834-47235834 on Build GRCh38
rs756787749 Chr.3: 47235562-47235562 on Build GRCh38

View Full Product Details