Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48562658-48563162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UCN2
Entrez Gene ID
90226
Gene Name
urocortin 2
Gene Aliases
SRP, UCN-II, UCNI, UR, URP
UniGene
Hs.631914
Gene Chromosome Location
Chr.3: 48561718-48563768 on Build GRCh38
Transcripts
NM_033199.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4567879 c.90_91CC>TT p.R31C Chr.3: 48563034-48563035 on Build GRCh38
4509860 c.81C>A p.T27T Chr.3: 48563044-48563044 on Build GRCh38
4158111 c.209C>T p.S70L Chr.3: 48562916-48562916 on Build GRCh38
3974452 c.91C>T p.R31C Chr.3: 48563034-48563034 on Build GRCh38
3945537 c.149C>G p.A50G Chr.3: 48562976-48562976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762392342 Chr.3: 48562847-48562847 on Build GRCh38
rs1003131154 Chr.3: 48562705-48562705 on Build GRCh38
rs142687169 Chr.3: 48563126-48563126 on Build GRCh38
rs778224095 Chr.3: 48562989-48562989 on Build GRCh38
rs781418790 Chr.3: 48562959-48562959 on Build GRCh38
rs746799381 Chr.3: 48562776-48562776 on Build GRCh38
rs749118149 Chr.3: 48562803-48562803 on Build GRCh38
rs200038467 Chr.3: 48563009-48563009 on Build GRCh38
rs761790367 Chr.3: 48562786-48562786 on Build GRCh38
rs780231872 Chr.3: 48562894-48562894 on Build GRCh38
rs769732102 Chr.3: 48562841-48562841 on Build GRCh38
rs780430031 Chr.3: 48562898-48562898 on Build GRCh38
rs747293529 Chr.3: 48562825-48562825 on Build GRCh38
rs192610365 Chr.3: 48562998-48562998 on Build GRCh38
rs200780329 Chr.3: 48562915-48562915 on Build GRCh38
rs770902414 Chr.3: 48562966-48562966 on Build GRCh38
rs751664694 Chr.3: 48562948-48562948 on Build GRCh38
rs778940067 Chr.3: 48563094-48563094 on Build GRCh38
rs897149630 Chr.3: 48562805-48562805 on Build GRCh38
rs749174738 Chr.3: 48562963-48562963 on Build GRCh38
rs781323282 Chr.3: 48563003-48563003 on Build GRCh38
rs768620393 Chr.3: 48563125-48563125 on Build GRCh38
rs199600581 Chr.3: 48562819-48562819 on Build GRCh38
rs982147757 Chr.3: 48562703-48562703 on Build GRCh38
rs560143325 Chr.3: 48562783-48562783 on Build GRCh38
rs764717156 Chr.3: 48562788-48562788 on Build GRCh38
rs762298735 Chr.3: 48562928-48562928 on Build GRCh38
rs747949979 Chr.3: 48562901-48562901 on Build GRCh38
rs368657396 Chr.3: 48562837-48562837 on Build GRCh38
rs771863692 Chr.3: 48562923-48562923 on Build GRCh38
rs78159609 Chr.3: 48562712-48562712 on Build GRCh38
rs777603693 Chr.3: 48562906-48562906 on Build GRCh38
rs201700183 Chr.3: 48563134-48563134 on Build GRCh38
rs754731072 Chr.3: 48562770-48562771 on Build GRCh38
rs145141083 Chr.3: 48562995-48562995 on Build GRCh38
rs1023894959 Chr.3: 48562860-48562860 on Build GRCh38
rs370633415 Chr.3: 48562913-48562913 on Build GRCh38
rs777083265 Chr.3: 48562927-48562927 on Build GRCh38
rs755135781 Chr.3: 48562953-48562953 on Build GRCh38
rs749685260 Chr.3: 48563133-48563133 on Build GRCh38
rs374482474 Chr.3: 48562814-48562814 on Build GRCh38
rs2633965 Chr.3: 48562859-48562859 on Build GRCh38
rs989210127 Chr.3: 48562728-48562733 on Build GRCh38
rs147683560 Chr.3: 48562900-48562900 on Build GRCh38
rs750909702 Chr.3: 48562823-48562823 on Build GRCh38
rs374682370 Chr.3: 48562758-48562758 on Build GRCh38
rs563118286 Chr.3: 48562734-48562734 on Build GRCh38
rs140823487 Chr.3: 48562916-48562916 on Build GRCh38
rs144853688 Chr.3: 48562891-48562891 on Build GRCh38
rs138367529 Chr.3: 48562797-48562797 on Build GRCh38
rs765858137 Chr.3: 48562887-48562887 on Build GRCh38
rs757958640 Chr.3: 48563112-48563112 on Build GRCh38
rs766017010 Chr.3: 48562849-48562849 on Build GRCh38
rs767627359 Chr.3: 48563034-48563034 on Build GRCh38
rs763302786 Chr.3: 48563011-48563011 on Build GRCh38
rs758716106 Chr.3: 48562834-48562834 on Build GRCh38
rs773846274 Chr.3: 48562853-48562853 on Build GRCh38
rs1011777076 Chr.3: 48562714-48562714 on Build GRCh38
rs200766954 Chr.3: 48562756-48562756 on Build GRCh38
rs753677449 Chr.3: 48563083-48563083 on Build GRCh38
rs774812814 Chr.3: 48563139-48563139 on Build GRCh38
rs768595692 Chr.3: 48562781-48562781 on Build GRCh38
rs200363094 Chr.3: 48562914-48562914 on Build GRCh38
rs752288852 Chr.3: 48562914-48562916 on Build GRCh38
rs183513733 Chr.3: 48562885-48562885 on Build GRCh38
rs775292544 Chr.3: 48562771-48562771 on Build GRCh38
rs754258975 Chr.3: 48562884-48562884 on Build GRCh38
rs759647833 Chr.3: 48563021-48563021 on Build GRCh38
rs906164284 Chr.3: 48562678-48562678 on Build GRCh38
rs757089177 Chr.3: 48563089-48563089 on Build GRCh38
rs903310048 Chr.3: 48562899-48562899 on Build GRCh38
rs762599762 Chr.3: 48562798-48562798 on Build GRCh38
rs754381784 Chr.3: 48562815-48562815 on Build GRCh38
rs780820752 Chr.3: 48562775-48562776 on Build GRCh38
rs779767464 Chr.3: 48563123-48563123 on Build GRCh38
rs549051034 Chr.3: 48562818-48562818 on Build GRCh38
rs423561 Chr.3: 48562713-48562713 on Build GRCh38
rs977440961 Chr.3: 48562779-48562779 on Build GRCh38
rs368389228 Chr.3: 48562905-48562905 on Build GRCh38
rs371104367 Chr.3: 48562802-48562802 on Build GRCh38
rs763726856 Chr.3: 48563080-48563080 on Build GRCh38
rs764548408 Chr.3: 48562861-48562861 on Build GRCh38
rs750871049 Chr.3: 48562931-48562931 on Build GRCh38
rs759289349 Chr.3: 48562854-48562854 on Build GRCh38
rs745840289 Chr.3: 48563110-48563110 on Build GRCh38
rs970428314 Chr.3: 48562686-48562686 on Build GRCh38
rs751720407 Chr.3: 48562896-48562896 on Build GRCh38
rs756392975 Chr.3: 48563070-48563070 on Build GRCh38
rs779191515 Chr.3: 48562737-48562737 on Build GRCh38
rs140335326 Chr.3: 48562960-48562960 on Build GRCh38
rs372343725 Chr.3: 48562997-48562997 on Build GRCh38
rs752897108 Chr.3: 48563050-48563050 on Build GRCh38
rs368177355 Chr.3: 48562939-48562939 on Build GRCh38
rs750472578 Chr.3: 48562889-48562889 on Build GRCh38
rs765735658 Chr.3: 48562929-48562929 on Build GRCh38
rs149022309 Chr.3: 48563004-48563004 on Build GRCh38
rs748251551 Chr.3: 48563020-48563020 on Build GRCh38
rs1007570429 Chr.3: 48563120-48563120 on Build GRCh38
rs763575871 Chr.3: 48562936-48562936 on Build GRCh38
rs768420794 Chr.3: 48562830-48562830 on Build GRCh38
rs78525960 Chr.3: 48563026-48563026 on Build GRCh38
rs777675487 Chr.3: 48562961-48562961 on Build GRCh38
rs766923243 Chr.3: 48563012-48563012 on Build GRCh38
rs772651953 Chr.3: 48562845-48562845 on Build GRCh38

View Full Product Details