Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48603472-48603972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UQCRC1
Entrez Gene ID
7384
Gene Name
ubiquinol-cytochrome c reductase core protein I
Gene Aliases
D3S3191, QCR1, UQCR1
UniGene
Hs.119251
Gene Chromosome Location
Chr.3: 48598999-48609665 on Build GRCh38
Transcripts
NM_003365.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595029 c.637C>T p.R213C Chr.3: 48603633-48603633 on Build GRCh38
446661 c.688G>C p.V230L Chr.3: 48603582-48603582 on Build GRCh38
1045888 c.651C>T p.T217T Chr.3: 48603619-48603619 on Build GRCh38
5399543 c.682C>T p.R228* Chr.3: 48603588-48603588 on Build GRCh38
3683137 c.643G>C p.D215H Chr.3: 48603627-48603627 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1020094211 Chr.3: 48603698-48603698 on Build GRCh38
rs779076507 Chr.3: 48603690-48603690 on Build GRCh38
rs752346245 Chr.3: 48603671-48603671 on Build GRCh38
rs747496002 Chr.3: 48603702-48603702 on Build GRCh38
rs201771292 Chr.3: 48603673-48603673 on Build GRCh38
rs937394966 Chr.3: 48603895-48603895 on Build GRCh38
rs975832664 Chr.3: 48603742-48603742 on Build GRCh38
rs1049980317 Chr.3: 48603504-48603504 on Build GRCh38
rs564951346 Chr.3: 48603619-48603619 on Build GRCh38
rs931020019 Chr.3: 48603869-48603869 on Build GRCh38
rs766328762 Chr.3: 48603655-48603655 on Build GRCh38
rs773065384 Chr.3: 48603632-48603632 on Build GRCh38
rs768976832 Chr.3: 48603612-48603612 on Build GRCh38
rs762761417 Chr.3: 48603638-48603638 on Build GRCh38
rs950194063 Chr.3: 48603842-48603842 on Build GRCh38
rs932914305 Chr.3: 48603503-48603503 on Build GRCh38
rs17080284 Chr.3: 48603627-48603627 on Build GRCh38
rs935158580 Chr.3: 48603907-48603907 on Build GRCh38
rs1032954606 Chr.3: 48603658-48603658 on Build GRCh38
rs920162485 Chr.3: 48603863-48603863 on Build GRCh38
rs779458757 Chr.3: 48603595-48603595 on Build GRCh38
rs756208111 Chr.3: 48603679-48603679 on Build GRCh38
rs757634111 Chr.3: 48603569-48603569 on Build GRCh38
rs1010105633 Chr.3: 48603685-48603685 on Build GRCh38
rs905836922 Chr.3: 48603642-48603642 on Build GRCh38
rs780875339 Chr.3: 48603549-48603549 on Build GRCh38
rs368060850 Chr.3: 48603618-48603618 on Build GRCh38
rs2267843 Chr.3: 48603774-48603774 on Build GRCh38
rs762961906 Chr.3: 48603532-48603532 on Build GRCh38
rs965787012 Chr.3: 48603736-48603736 on Build GRCh38
rs371808760 Chr.3: 48603670-48603670 on Build GRCh38
rs750386055 Chr.3: 48603527-48603527 on Build GRCh38
rs754202987 Chr.3: 48603555-48603555 on Build GRCh38
rs1045845457 Chr.3: 48603873-48603873 on Build GRCh38
rs1029088214 Chr.3: 48603749-48603749 on Build GRCh38
rs116149309 Chr.3: 48603499-48603499 on Build GRCh38
rs536537284 Chr.3: 48603751-48603751 on Build GRCh38
rs751103384 Chr.3: 48603545-48603545 on Build GRCh38
rs1028431641 Chr.3: 48603712-48603712 on Build GRCh38
rs13085761 Chr.3: 48603764-48603764 on Build GRCh38
rs984845407 Chr.3: 48603752-48603752 on Build GRCh38
rs778051361 Chr.3: 48603687-48603690 on Build GRCh38
rs373691566 Chr.3: 48603766-48603766 on Build GRCh38
rs749786068 Chr.3: 48603689-48603690 on Build GRCh38
rs916226699 Chr.3: 48603890-48603890 on Build GRCh38
rs114461058 Chr.3: 48603935-48603935 on Build GRCh38
rs910168775 Chr.3: 48603767-48603767 on Build GRCh38
rs902359088 Chr.3: 48603903-48603903 on Build GRCh38
rs972879259 Chr.3: 48603791-48603791 on Build GRCh38
rs905927580 Chr.3: 48603891-48603891 on Build GRCh38
rs772656995 Chr.3: 48603790-48603791 on Build GRCh38
rs894887972 Chr.3: 48603500-48603500 on Build GRCh38
rs910123899 Chr.3: 48603540-48603540 on Build GRCh38
rs865824723 Chr.3: 48603707-48603707 on Build GRCh38
rs751050320 Chr.3: 48603662-48603662 on Build GRCh38
rs1046099283 Chr.3: 48603894-48603894 on Build GRCh38
rs954128005 Chr.3: 48603739-48603739 on Build GRCh38
rs746298877 Chr.3: 48603597-48603597 on Build GRCh38
rs761436568 Chr.3: 48603693-48603694 on Build GRCh38
rs973046957 Chr.3: 48603803-48603803 on Build GRCh38
rs754578893 Chr.3: 48603547-48603547 on Build GRCh38
rs765189811 Chr.3: 48603525-48603525 on Build GRCh38
rs918733100 Chr.3: 48603832-48603832 on Build GRCh38
rs374960172 Chr.3: 48603608-48603608 on Build GRCh38
rs758923653 Chr.3: 48603693-48603693 on Build GRCh38
rs780753563 Chr.3: 48603694-48603694 on Build GRCh38
rs374455325 Chr.3: 48603691-48603691 on Build GRCh38
rs755878675 Chr.3: 48603845-48603845 on Build GRCh38
rs111932041 Chr.3: 48603646-48603646 on Build GRCh38
rs549512516 Chr.3: 48603604-48603604 on Build GRCh38
rs370760243 Chr.3: 48603521-48603521 on Build GRCh38
rs769212522 Chr.3: 48603808-48603808 on Build GRCh38
rs374751960 Chr.3: 48603939-48603939 on Build GRCh38
rs564627874 Chr.3: 48603695-48603695 on Build GRCh38
rs765810610 Chr.3: 48603537-48603537 on Build GRCh38
rs759090174 Chr.3: 48603665-48603665 on Build GRCh38

View Full Product Details