Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49285906-49286427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP4
Entrez Gene ID
7375
Gene Name
ubiquitin specific peptidase 4
Gene Aliases
UNP, Unph
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49277144-49340103 on Build GRCh38
Transcripts
NM_003363.3 NM_199443.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5954160 c.2005G>A p.E669K Chr.3: 49286293-49286293 on Build GRCh38
7110968 c.2047G>C p.D683H Chr.3: 49286251-49286251 on Build GRCh38
355457 c.2177A>G p.D726G Chr.3: 49286121-49286121 on Build GRCh38
3767466 c.2029A>G p.T677A Chr.3: 49286269-49286269 on Build GRCh38
7316933 c.2062G>A p.D688N Chr.3: 49286236-49286236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139138506 Chr.3: 49286231-49286231 on Build GRCh38
rs762343417 Chr.3: 49286244-49286244 on Build GRCh38
rs200730474 Chr.3: 49286225-49286225 on Build GRCh38
rs370990799 Chr.3: 49286051-49286051 on Build GRCh38
rs769069770 Chr.3: 49286351-49286351 on Build GRCh38
rs191817413 Chr.3: 49286013-49286013 on Build GRCh38
rs541925823 Chr.3: 49286128-49286128 on Build GRCh38
rs748763135 Chr.3: 49286314-49286314 on Build GRCh38
rs776381838 Chr.3: 49286085-49286085 on Build GRCh38
rs140439154 Chr.3: 49286175-49286175 on Build GRCh38
rs370663035 Chr.3: 49286153-49286153 on Build GRCh38
rs780600441 Chr.3: 49286089-49286089 on Build GRCh38
rs369375022 Chr.3: 49286246-49286246 on Build GRCh38
rs767424577 Chr.3: 49286286-49286286 on Build GRCh38
rs769099094 Chr.3: 49286236-49286236 on Build GRCh38
rs563308930 Chr.3: 49286281-49286281 on Build GRCh38
rs1038729264 Chr.3: 49286148-49286148 on Build GRCh38
rs988960196 Chr.3: 49286123-49286123 on Build GRCh38
rs769294022 Chr.3: 49286214-49286214 on Build GRCh38
rs775870883 Chr.3: 49286125-49286125 on Build GRCh38
rs758128203 Chr.3: 49286328-49286328 on Build GRCh38
rs769508882 Chr.3: 49286177-49286177 on Build GRCh38
rs901631343 Chr.3: 49286398-49286398 on Build GRCh38
rs768260247 Chr.3: 49286057-49286057 on Build GRCh38
rs754791104 Chr.3: 49286284-49286284 on Build GRCh38
rs777201100 Chr.3: 49286216-49286216 on Build GRCh38
rs754912818 Chr.3: 49286195-49286195 on Build GRCh38
rs866935137 Chr.3: 49286325-49286325 on Build GRCh38
rs761077333 Chr.3: 49286232-49286232 on Build GRCh38
rs779929164 Chr.3: 49286057-49286057 on Build GRCh38
rs968043659 Chr.3: 49286357-49286357 on Build GRCh38
rs748688565 Chr.3: 49286369-49286369 on Build GRCh38
rs1040485924 Chr.3: 49286397-49286397 on Build GRCh38
rs979051098 Chr.3: 49286307-49286307 on Build GRCh38
rs901301454 Chr.3: 49286083-49286083 on Build GRCh38
rs748957590 Chr.3: 49286220-49286220 on Build GRCh38
rs1043409277 Chr.3: 49285992-49285992 on Build GRCh38
rs766426541 Chr.3: 49286397-49286398 on Build GRCh38
rs746934295 Chr.3: 49286067-49286067 on Build GRCh38
rs752639565 Chr.3: 49286291-49286291 on Build GRCh38
rs756905159 Chr.3: 49286327-49286327 on Build GRCh38
rs781169911 Chr.3: 49286201-49286201 on Build GRCh38
rs760980133 Chr.3: 49286112-49286112 on Build GRCh38
rs762805926 Chr.3: 49286323-49286325 on Build GRCh38
rs944335444 Chr.3: 49286138-49286138 on Build GRCh38
rs372181380 Chr.3: 49286347-49286347 on Build GRCh38
rs761114777 Chr.3: 49286127-49286127 on Build GRCh38
rs768591584 Chr.3: 49286071-49286071 on Build GRCh38
rs752337711 Chr.3: 49286288-49286288 on Build GRCh38
rs758636661 Chr.3: 49286181-49286181 on Build GRCh38
rs183009991 Chr.3: 49286188-49286188 on Build GRCh38
rs761837873 Chr.3: 49286091-49286091 on Build GRCh38
rs776802330 Chr.3: 49286356-49286356 on Build GRCh38
rs766594024 Chr.3: 49286277-49286277 on Build GRCh38
rs913824669 Chr.3: 49285974-49285974 on Build GRCh38
rs778612981 Chr.3: 49286340-49286340 on Build GRCh38
rs577634738 Chr.3: 49286092-49286092 on Build GRCh38
rs377601795 Chr.3: 49286098-49286098 on Build GRCh38
rs777291077 Chr.3: 49286301-49286301 on Build GRCh38
rs763300392 Chr.3: 49286254-49286254 on Build GRCh38
rs1040145142 Chr.3: 49286072-49286072 on Build GRCh38
rs1035119359 Chr.3: 49286194-49286194 on Build GRCh38
rs773377532 Chr.3: 49286248-49286248 on Build GRCh38
rs755992508 Chr.3: 49286300-49286300 on Build GRCh38
rs754095579 Chr.3: 49286278-49286278 on Build GRCh38
rs773000465 Chr.3: 49286313-49286315 on Build GRCh38
rs946383602 Chr.3: 49285977-49285977 on Build GRCh38
rs777190330 Chr.3: 49286240-49286240 on Build GRCh38
rs561364286 Chr.3: 49286378-49286378 on Build GRCh38
rs913814782 Chr.3: 49286258-49286258 on Build GRCh38
rs765621253 Chr.3: 49286165-49286165 on Build GRCh38
rs1012251517 Chr.3: 49285932-49285932 on Build GRCh38
rs956921077 Chr.3: 49286120-49286120 on Build GRCh38
rs1012632182 Chr.3: 49286282-49286282 on Build GRCh38
rs772842134 Chr.3: 49286101-49286101 on Build GRCh38
rs373889878 Chr.3: 49286186-49286186 on Build GRCh38
rs753912219 Chr.3: 49286133-49286133 on Build GRCh38
rs930565760 Chr.3: 49286162-49286162 on Build GRCh38
rs144324033 Chr.3: 49286202-49286202 on Build GRCh38
rs544412000 Chr.3: 49285950-49285950 on Build GRCh38

View Full Product Details